Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97584539-97585039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOGA1
Entrez Gene ID
112817
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases
C10orf65, DHDPS2, DHDPSL, HP3, NPL2
UniGene
Hs.180346
Gene Chromosome Location
Chr.10: 97584345-97612802 on Build GRCh38
Transcripts
NM_001134670.1 NM_138413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5591284 c.47G>A p.R16K Chr.10: 97584750-97584750 on Build GRCh38
4945561 c.19T>C p.W7R Chr.10: 97584722-97584722 on Build GRCh38
147017 c.90G>A p.G30G Chr.10: 97584793-97584793 on Build GRCh38
3442429 c.60G>A p.R20R Chr.10: 97584763-97584763 on Build GRCh38
7106916 c.163G>T p.G55W Chr.10: 97584866-97584866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765289362 Chr.10: 97584895-97584895 on Build GRCh38
rs775876371 Chr.10: 97584834-97584834 on Build GRCh38
rs916935304 Chr.10: 97584945-97584945 on Build GRCh38
rs558461907 Chr.10: 97584988-97584989 on Build GRCh38
rs756272372 Chr.10: 97584714-97584714 on Build GRCh38
rs954248865 Chr.10: 97584863-97584863 on Build GRCh38
rs372517912 Chr.10: 97584928-97584928 on Build GRCh38
rs768415273 Chr.10: 97584759-97584759 on Build GRCh38
rs201467858 Chr.10: 97584952-97584952 on Build GRCh38
rs764274149 Chr.10: 97584788-97584788 on Build GRCh38
rs559777676 Chr.10: 97584590-97584590 on Build GRCh38
rs985573876 Chr.10: 97584864-97584864 on Build GRCh38
rs200127255 Chr.10: 97584742-97584742 on Build GRCh38
rs369191992 Chr.10: 97584903-97584903 on Build GRCh38
rs746712236 Chr.10: 97584754-97584754 on Build GRCh38
rs763621943 Chr.10: 97584672-97584672 on Build GRCh38
rs1056874645 Chr.10: 97584741-97584741 on Build GRCh38
rs766418467 Chr.10: 97584663-97584663 on Build GRCh38
rs753970733 Chr.10: 97584791-97584791 on Build GRCh38
rs758339645 Chr.10: 97584806-97584806 on Build GRCh38
rs776736742 Chr.10: 97584760-97584760 on Build GRCh38
rs764396564 Chr.10: 97584837-97584837 on Build GRCh38
rs371585334 Chr.10: 97584674-97584674 on Build GRCh38
rs769193078 Chr.10: 97584812-97584812 on Build GRCh38
rs141993402 Chr.10: 97584871-97584871 on Build GRCh38
rs924232072 Chr.10: 97584709-97584710 on Build GRCh38
rs888278009 Chr.10: 97584987-97584987 on Build GRCh38
rs772722925 Chr.10: 97584813-97584813 on Build GRCh38
rs562415026 Chr.10: 97584620-97584620 on Build GRCh38
rs747588198 Chr.10: 97584764-97584764 on Build GRCh38
rs774752851 Chr.10: 97584779-97584779 on Build GRCh38
rs771766335 Chr.10: 97584744-97584744 on Build GRCh38
rs746419489 Chr.10: 97584820-97584820 on Build GRCh38
rs761009383 Chr.10: 97584835-97584835 on Build GRCh38
rs573292460 Chr.10: 97584809-97584809 on Build GRCh38
rs202016288 Chr.10: 97584821-97584821 on Build GRCh38
rs267606763 Chr.10: 97584912-97584912 on Build GRCh38
rs757262365 Chr.10: 97584729-97584729 on Build GRCh38
rs779935047 Chr.10: 97584753-97584753 on Build GRCh38
rs777215856 Chr.10: 97584941-97584941 on Build GRCh38
rs1044127300 Chr.10: 97584949-97584949 on Build GRCh38
rs765733570 Chr.10: 97584942-97584942 on Build GRCh38
rs986057755 Chr.10: 97584618-97584618 on Build GRCh38
rs750507401 Chr.10: 97584904-97584904 on Build GRCh38
rs934329920 Chr.10: 97584943-97584943 on Build GRCh38
rs948015722 Chr.10: 97584926-97584926 on Build GRCh38
rs758304537 Chr.10: 97584911-97584911 on Build GRCh38
rs957227758 Chr.10: 97584884-97584884 on Build GRCh38
rs796052091 Chr.10: 97584861-97584861 on Build GRCh38
rs759795758 Chr.10: 97584792-97584792 on Build GRCh38
rs751493421 Chr.10: 97584919-97584919 on Build GRCh38
rs756707266 Chr.10: 97584943-97584945 on Build GRCh38
rs780503515 Chr.10: 97584947-97584948 on Build GRCh38
rs778696295 Chr.10: 97584736-97584736 on Build GRCh38
rs948441948 Chr.10: 97584947-97584947 on Build GRCh38
rs202120436 Chr.10: 97584943-97584943 on Build GRCh38
rs201803986 Chr.10: 97584810-97584810 on Build GRCh38
rs776319977 Chr.10: 97584763-97584763 on Build GRCh38
rs750736378 Chr.10: 97584793-97584795 on Build GRCh38
rs775819365 Chr.10: 97584784-97584784 on Build GRCh38
rs760703304 Chr.10: 97584638-97584638 on Build GRCh38
rs752002204 Chr.10: 97584679-97584679 on Build GRCh38
rs529940373 Chr.10: 97584705-97584705 on Build GRCh38
rs750339099 Chr.10: 97584804-97584804 on Build GRCh38
rs760958174 Chr.10: 97584785-97584785 on Build GRCh38
rs368335660 Chr.10: 97584773-97584773 on Build GRCh38
rs781068354 Chr.10: 97584929-97584929 on Build GRCh38
rs968208573 Chr.10: 97584890-97584890 on Build GRCh38
rs552420631 Chr.10: 97584934-97584934 on Build GRCh38
rs759839443 Chr.10: 97584696-97584696 on Build GRCh38
rs747795341 Chr.10: 97584811-97584811 on Build GRCh38
rs749265107 Chr.10: 97584570-97584570 on Build GRCh38
rs767786474 Chr.10: 97584783-97584783 on Build GRCh38
rs760024469 Chr.10: 97584780-97584780 on Build GRCh38
rs116731711 Chr.10: 97584723-97584723 on Build GRCh38
rs772564071 Chr.10: 97584832-97584832 on Build GRCh38
rs755777934 Chr.10: 97584939-97584939 on Build GRCh38
rs192567315 Chr.10: 97584802-97584802 on Build GRCh38
rs375261073 Chr.10: 97584707-97584707 on Build GRCh38
rs768560477 Chr.10: 97584587-97584587 on Build GRCh38
rs544209799 Chr.10: 97584613-97584613 on Build GRCh38
rs150812556 Chr.10: 97584888-97584888 on Build GRCh38
rs930035823 Chr.10: 97584984-97584984 on Build GRCh38
rs1047258297 Chr.10: 97584577-97584577 on Build GRCh38

View Full Product Details