Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116634749-116635171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNLIPRP2
Entrez Gene ID
5408
Gene Name
pancreatic lipase related protein 2 (gene/pseudogene)
Gene Aliases
PLRP2
UniGene
Hs.423598
Gene Chromosome Location
Chr.10: 116620953-116645143 on Build GRCh38
Transcripts
NM_005396.4 NR_103727.1 XM_017016339.1

SNP Detail

SNP ID Chromosome Location
rs782502072 Chr.10: 116635055-116635055 on Build GRCh38
rs782296069 Chr.10: 116635005-116635005 on Build GRCh38
rs782618115 Chr.10: 116635121-116635121 on Build GRCh38
rs1015086172 Chr.10: 116635032-116635032 on Build GRCh38
rs781869781 Chr.10: 116634844-116634844 on Build GRCh38
rs544737732 Chr.10: 116634899-116634899 on Build GRCh38
rs782467436 Chr.10: 116634869-116634869 on Build GRCh38
rs184400248 Chr.10: 116634867-116634867 on Build GRCh38
rs781924300 Chr.10: 116634881-116634882 on Build GRCh38
rs977247637 Chr.10: 116635126-116635126 on Build GRCh38
rs782164242 Chr.10: 116634830-116634830 on Build GRCh38
rs782689759 Chr.10: 116634799-116634799 on Build GRCh38
rs1021360803 Chr.10: 116635062-116635062 on Build GRCh38
rs967553440 Chr.10: 116635119-116635119 on Build GRCh38
rs782286939 Chr.10: 116634803-116634803 on Build GRCh38
rs782761048 Chr.10: 116634861-116634861 on Build GRCh38
rs782330423 Chr.10: 116634900-116634900 on Build GRCh38
rs1049159832 Chr.10: 116634964-116634964 on Build GRCh38
rs781822444 Chr.10: 116634865-116634865 on Build GRCh38
rs782658256 Chr.10: 116634898-116634898 on Build GRCh38
rs781973501 Chr.10: 116634958-116634958 on Build GRCh38
rs541266144 Chr.10: 116634968-116634968 on Build GRCh38
rs45601737 Chr.10: 116635001-116635001 on Build GRCh38
rs782330038 Chr.10: 116634830-116634831 on Build GRCh38
rs782511459 Chr.10: 116634993-116634993 on Build GRCh38
rs782017496 Chr.10: 116634935-116634935 on Build GRCh38
rs781960259 Chr.10: 116634809-116634809 on Build GRCh38
rs781917199 Chr.10: 116634826-116634826 on Build GRCh38
rs782722888 Chr.10: 116634996-116634996 on Build GRCh38
rs547718487 Chr.10: 116635037-116635037 on Build GRCh38
rs782719454 Chr.10: 116634950-116634950 on Build GRCh38
rs192226180 Chr.10: 116634804-116634804 on Build GRCh38
rs782206708 Chr.10: 116634820-116634821 on Build GRCh38
rs782668658 Chr.10: 116635007-116635007 on Build GRCh38
rs200247568 Chr.10: 116634884-116634884 on Build GRCh38
rs782132518 Chr.10: 116634945-116634945 on Build GRCh38
rs782563884 Chr.10: 116634881-116634881 on Build GRCh38
rs534190844 Chr.10: 116635048-116635048 on Build GRCh38
rs532286435 Chr.10: 116634850-116634850 on Build GRCh38
rs558623051 Chr.10: 116634953-116634953 on Build GRCh38
rs375176437 Chr.10: 116634846-116634846 on Build GRCh38
rs368506466 Chr.10: 116634882-116634882 on Build GRCh38
rs782802363 Chr.10: 116634974-116634974 on Build GRCh38
rs902095431 Chr.10: 116635034-116635034 on Build GRCh38
rs782791395 Chr.10: 116634841-116634841 on Build GRCh38
rs1015117370 Chr.10: 116634818-116634818 on Build GRCh38
rs372119293 Chr.10: 116634988-116634988 on Build GRCh38
rs782587998 Chr.10: 116634907-116634907 on Build GRCh38
rs2286779 Chr.10: 116635040-116635040 on Build GRCh38
rs577075588 Chr.10: 116635021-116635021 on Build GRCh38
rs781922830 Chr.10: 116634960-116634960 on Build GRCh38
rs938617766 Chr.10: 116634955-116634955 on Build GRCh38

View Full Product Details