Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116856535-116857067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
NM_001242699.1 XM_005269815.3 XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275591 c.396G>T p.V132V Chr.10: 116856590-116856590 on Build GRCh38
2054863 c.392C>T p.A131V Chr.10: 116856586-116856586 on Build GRCh38
6518323 c.353C>A p.P118H Chr.10: 116856547-116856547 on Build GRCh38
6837797 c.358C>G p.L120V Chr.10: 116856552-116856552 on Build GRCh38
7322252 c.436G>A p.E146K Chr.10: 116856630-116856630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866235466 Chr.10: 116856555-116856555 on Build GRCh38
rs867172257 Chr.10: 116856797-116856797 on Build GRCh38
rs973501323 Chr.10: 116856768-116856768 on Build GRCh38
rs7083543 Chr.10: 116856599-116856599 on Build GRCh38
rs922658625 Chr.10: 116857005-116857005 on Build GRCh38
rs747107212 Chr.10: 116856602-116856602 on Build GRCh38
rs929369969 Chr.10: 116856789-116856789 on Build GRCh38
rs1001869018 Chr.10: 116856950-116856950 on Build GRCh38
rs779310798 Chr.10: 116856562-116856562 on Build GRCh38
rs537871560 Chr.10: 116856767-116856767 on Build GRCh38
rs1051926130 Chr.10: 116856594-116856594 on Build GRCh38
rs1048849958 Chr.10: 116856983-116856983 on Build GRCh38
rs984278323 Chr.10: 116856956-116856956 on Build GRCh38
rs1040690434 Chr.10: 116856651-116856651 on Build GRCh38
rs961252401 Chr.10: 116856792-116856792 on Build GRCh38
rs1033609221 Chr.10: 116856777-116856777 on Build GRCh38
rs569610808 Chr.10: 116856595-116856595 on Build GRCh38
rs1013879088 Chr.10: 116856804-116856804 on Build GRCh38
rs1681719 Chr.10: 116856882-116856882 on Build GRCh38
rs138188097 Chr.10: 116856739-116856739 on Build GRCh38
rs532822452 Chr.10: 116856684-116856684 on Build GRCh38
rs7087685 Chr.10: 116857026-116857026 on Build GRCh38
rs534463390 Chr.10: 116856967-116856967 on Build GRCh38
rs529048275 Chr.10: 116856735-116856735 on Build GRCh38
rs935676494 Chr.10: 116856611-116856611 on Build GRCh38
rs768385505 Chr.10: 116856629-116856629 on Build GRCh38
rs550523692 Chr.10: 116856600-116856600 on Build GRCh38
rs576643065 Chr.10: 116856968-116856968 on Build GRCh38
rs770989176 Chr.10: 116856605-116856605 on Build GRCh38
rs374526334 Chr.10: 116856850-116856850 on Build GRCh38
rs371072508 Chr.10: 116856626-116856626 on Build GRCh38
rs891579684 Chr.10: 116856881-116856881 on Build GRCh38
rs758855274 Chr.10: 116856771-116856771 on Build GRCh38
rs929435301 Chr.10: 116856587-116856587 on Build GRCh38
rs887837587 Chr.10: 116856753-116856753 on Build GRCh38
rs926622245 Chr.10: 116856765-116856765 on Build GRCh38
rs796487492 Chr.10: 116856991-116856991 on Build GRCh38
rs780967991 Chr.10: 116856775-116856775 on Build GRCh38
rs887872148 Chr.10: 116856999-116856999 on Build GRCh38
rs1013992017 Chr.10: 116857040-116857040 on Build GRCh38
rs7087301 Chr.10: 116856990-116856990 on Build GRCh38
rs904217069 Chr.10: 116856851-116856851 on Build GRCh38
rs1037753288 Chr.10: 116856578-116856578 on Build GRCh38
rs755450363 Chr.10: 116856596-116856596 on Build GRCh38
rs569113526 Chr.10: 116856758-116856758 on Build GRCh38
rs999843430 Chr.10: 116856861-116856861 on Build GRCh38
rs925712064 Chr.10: 116856839-116856839 on Build GRCh38
rs66621882 Chr.10: 116857004-116857004 on Build GRCh38
rs546392561 Chr.10: 116856685-116856685 on Build GRCh38
rs555692105 Chr.10: 116857005-116857005 on Build GRCh38
rs897690645 Chr.10: 116856713-116856713 on Build GRCh38
rs969880655 Chr.10: 116856931-116856931 on Build GRCh38
rs910257196 Chr.10: 116856939-116856939 on Build GRCh38
rs768416240 Chr.10: 116856989-116856989 on Build GRCh38
rs929157598 Chr.10: 116856974-116856974 on Build GRCh38
rs890592456 Chr.10: 116856795-116856795 on Build GRCh38
rs186461606 Chr.10: 116856806-116856806 on Build GRCh38
rs200907862 Chr.10: 116856586-116856586 on Build GRCh38
rs944363296 Chr.10: 116856941-116856941 on Build GRCh38
rs919148625 Chr.10: 116856961-116856961 on Build GRCh38
rs957074707 Chr.10: 116856855-116856855 on Build GRCh38
rs373776170 Chr.10: 116856868-116856868 on Build GRCh38
rs564042506 Chr.10: 116856660-116856660 on Build GRCh38
rs966934387 Chr.10: 116856821-116856821 on Build GRCh38
rs755433351 Chr.10: 116856560-116856560 on Build GRCh38
rs571775740 Chr.10: 116856805-116856805 on Build GRCh38
rs972952697 Chr.10: 116856992-116856992 on Build GRCh38
rs529586945 Chr.10: 116856597-116856597 on Build GRCh38
rs781463624 Chr.10: 116856625-116856625 on Build GRCh38
rs992746610 Chr.10: 116856590-116856590 on Build GRCh38
rs1024561765 Chr.10: 116856914-116856914 on Build GRCh38
rs938060122 Chr.10: 116857002-116857002 on Build GRCh38
rs942093828 Chr.10: 116856563-116856563 on Build GRCh38
rs547678562 Chr.10: 116856793-116856793 on Build GRCh38
rs910468625 Chr.10: 116856615-116856615 on Build GRCh38
rs1050059021 Chr.10: 116856612-116856612 on Build GRCh38
rs898007360 Chr.10: 116856585-116856585 on Build GRCh38
rs796506632 Chr.10: 116856990-116856991 on Build GRCh38
rs554303745 Chr.10: 116856846-116856846 on Build GRCh38
rs182056213 Chr.10: 116856724-116856724 on Build GRCh38
rs973437518 Chr.10: 116856976-116856976 on Build GRCh38

View Full Product Details