Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 131967613-131968115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNIP3
Entrez Gene ID
664
Gene Name
BCL2 interacting protein 3
Gene Aliases
NIP3
UniGene
Hs.144873
Gene Chromosome Location
Chr.10: 131967683-131982013 on Build GRCh38
Transcripts
NM_004052.3 XM_006717914.3 XM_017016407.1

Gene Symbol
PPP2R2D
Entrez Gene ID
55844
Gene Name
protein phosphatase 2 regulatory subunit Bdelta
Gene Aliases
MDS026
UniGene
Hs.380372
Gene Chromosome Location
Chr.10: 131900644-131971535 on Build GRCh38
Transcripts
NM_004052.3 XM_006717914.3 XM_017016407.1

SNP Detail

SNP ID Chromosome Location
rs1801945 Chr.10: 131968079-131968079 on Build GRCh38
rs1801943 Chr.10: 131967932-131967932 on Build GRCh38
rs576046309 Chr.10: 131967652-131967652 on Build GRCh38
rs781824927 Chr.10: 131967686-131967686 on Build GRCh38
rs59944351 Chr.10: 131967807-131967807 on Build GRCh38
rs61162388 Chr.10: 131967955-131967955 on Build GRCh38
rs557365163 Chr.10: 131967878-131967878 on Build GRCh38
rs374419972 Chr.10: 131968064-131968064 on Build GRCh38
rs554580169 Chr.10: 131968000-131968000 on Build GRCh38
rs1801944 Chr.10: 131967818-131967818 on Build GRCh38
rs59821801 Chr.10: 131967849-131967849 on Build GRCh38
rs185418198 Chr.10: 131967692-131967692 on Build GRCh38
rs45501697 Chr.10: 131967641-131967641 on Build GRCh38
rs141794142 Chr.10: 131967651-131967651 on Build GRCh38
rs6557 Chr.10: 131967945-131967945 on Build GRCh38
rs546626944 Chr.10: 131967899-131967899 on Build GRCh38
rs57368514 Chr.10: 131968013-131968013 on Build GRCh38
rs531210020 Chr.10: 131967793-131967793 on Build GRCh38
rs142348720 Chr.10: 131967856-131967856 on Build GRCh38
rs567620463 Chr.10: 131968067-131968067 on Build GRCh38
rs764029842 Chr.10: 131968081-131968082 on Build GRCh38
rs544375050 Chr.10: 131967646-131967647 on Build GRCh38
rs58784863 Chr.10: 131967864-131967864 on Build GRCh38
rs566454030 Chr.10: 131967900-131967900 on Build GRCh38
rs533535926 Chr.10: 131968012-131968012 on Build GRCh38
rs61697383 Chr.10: 131968062-131968062 on Build GRCh38
rs545609706 Chr.10: 131967679-131967679 on Build GRCh38
rs536262005 Chr.10: 131968075-131968075 on Build GRCh38
rs782673860 Chr.10: 131968043-131968043 on Build GRCh38
rs537426265 Chr.10: 131967844-131967844 on Build GRCh38
rs537730982 Chr.10: 131967993-131967993 on Build GRCh38
rs749659096 Chr.10: 131967770-131967770 on Build GRCh38
rs551949474 Chr.10: 131967950-131967950 on Build GRCh38
rs146088589 Chr.10: 131967656-131967656 on Build GRCh38
rs61074059 Chr.10: 131967834-131967834 on Build GRCh38

View Full Product Details