Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 4998905-4999412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR51L1
Entrez Gene ID
119682
Gene Name
olfactory receptor family 51 subfamily L member 1
Gene Aliases
OR11-31
UniGene
Hs.553559
Gene Chromosome Location
Chr.11: 4998983-4999930 on Build GRCh38
Transcripts
NM_001004755.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
689173 c.59C>T p.P20L Chr.11: 4999041-4999041 on Build GRCh38
4982939 c.173A>T p.H58L Chr.11: 4999155-4999155 on Build GRCh38
3448402 c.392C>T p.P131L Chr.11: 4999374-4999374 on Build GRCh38
1260361 c.46C>A p.L16M Chr.11: 4999028-4999028 on Build GRCh38
689170 c.225G>T p.M75I Chr.11: 4999207-4999207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749547529 Chr.11: 4999103-4999103 on Build GRCh38
rs138440814 Chr.11: 4999313-4999313 on Build GRCh38
rs770096924 Chr.11: 4998964-4998964 on Build GRCh38
rs371043717 Chr.11: 4999199-4999199 on Build GRCh38
rs757522991 Chr.11: 4999236-4999236 on Build GRCh38
rs374068048 Chr.11: 4999341-4999341 on Build GRCh38
rs764970849 Chr.11: 4999053-4999053 on Build GRCh38
rs141806275 Chr.11: 4998948-4998948 on Build GRCh38
rs987569301 Chr.11: 4998932-4998932 on Build GRCh38
rs781391024 Chr.11: 4999071-4999071 on Build GRCh38
rs746259216 Chr.11: 4999241-4999241 on Build GRCh38
rs759507472 Chr.11: 4999201-4999201 on Build GRCh38
rs569380711 Chr.11: 4999370-4999370 on Build GRCh38
rs138636727 Chr.11: 4999122-4999122 on Build GRCh38
rs770739453 Chr.11: 4999286-4999286 on Build GRCh38
rs199642881 Chr.11: 4999364-4999364 on Build GRCh38
rs747539456 Chr.11: 4999275-4999275 on Build GRCh38
rs146166863 Chr.11: 4999353-4999353 on Build GRCh38
rs775959370 Chr.11: 4999151-4999151 on Build GRCh38
rs913238396 Chr.11: 4999337-4999337 on Build GRCh38
rs780621507 Chr.11: 4999258-4999258 on Build GRCh38
rs199934313 Chr.11: 4999246-4999246 on Build GRCh38
rs752853746 Chr.11: 4999084-4999084 on Build GRCh38
rs747530446 Chr.11: 4999342-4999342 on Build GRCh38
rs759419787 Chr.11: 4999297-4999297 on Build GRCh38
rs201269119 Chr.11: 4999304-4999304 on Build GRCh38
rs11035064 Chr.11: 4998959-4998959 on Build GRCh38
rs142290760 Chr.11: 4999070-4999070 on Build GRCh38
rs146234178 Chr.11: 4999114-4999114 on Build GRCh38
rs373038829 Chr.11: 4998955-4998955 on Build GRCh38
rs188505113 Chr.11: 4998990-4998990 on Build GRCh38
rs141178578 Chr.11: 4999136-4999136 on Build GRCh38
rs771369267 Chr.11: 4999005-4999005 on Build GRCh38
rs764915130 Chr.11: 4998934-4998934 on Build GRCh38
rs149931922 Chr.11: 4999025-4999025 on Build GRCh38
rs777959813 Chr.11: 4998979-4998979 on Build GRCh38
rs761406267 Chr.11: 4999163-4999163 on Build GRCh38
rs776638517 Chr.11: 4999387-4999387 on Build GRCh38
rs535510477 Chr.11: 4999085-4999085 on Build GRCh38
rs749686928 Chr.11: 4998980-4998980 on Build GRCh38
rs772604721 Chr.11: 4999127-4999127 on Build GRCh38
rs373919370 Chr.11: 4998942-4998942 on Build GRCh38
rs762754122 Chr.11: 4999175-4999175 on Build GRCh38
rs147341630 Chr.11: 4999362-4999362 on Build GRCh38
rs144205047 Chr.11: 4999298-4999298 on Build GRCh38
rs769268885 Chr.11: 4999167-4999167 on Build GRCh38
rs377138037 Chr.11: 4998943-4998943 on Build GRCh38
rs781483028 Chr.11: 4998958-4998958 on Build GRCh38
rs772790375 Chr.11: 4999202-4999202 on Build GRCh38
rs778071155 Chr.11: 4999090-4999090 on Build GRCh38
rs11035066 Chr.11: 4999279-4999279 on Build GRCh38
rs927399811 Chr.11: 4999019-4999019 on Build GRCh38
rs113317534 Chr.11: 4999011-4999011 on Build GRCh38
rs201522839 Chr.11: 4999058-4999059 on Build GRCh38
rs750726834 Chr.11: 4999316-4999316 on Build GRCh38
rs752663337 Chr.11: 4999215-4999215 on Build GRCh38
rs115625304 Chr.11: 4999187-4999187 on Build GRCh38
rs755482024 Chr.11: 4999343-4999343 on Build GRCh38
rs774240039 Chr.11: 4999289-4999289 on Build GRCh38
rs765462952 Chr.11: 4999315-4999315 on Build GRCh38
rs184336225 Chr.11: 4999281-4999281 on Build GRCh38
rs775492437 Chr.11: 4999299-4999299 on Build GRCh38
rs756222153 Chr.11: 4999223-4999223 on Build GRCh38
rs750269408 Chr.11: 4998938-4998938 on Build GRCh38
rs766346263 Chr.11: 4999069-4999069 on Build GRCh38
rs370703740 Chr.11: 4999325-4999325 on Build GRCh38
rs767541099 Chr.11: 4998974-4998975 on Build GRCh38
rs760629509 Chr.11: 4999376-4999376 on Build GRCh38
rs762377268 Chr.11: 4999330-4999330 on Build GRCh38
rs768504439 Chr.11: 4999379-4999379 on Build GRCh38
rs374542301 Chr.11: 4998931-4998931 on Build GRCh38
rs1028726427 Chr.11: 4999165-4999165 on Build GRCh38
rs761337285 Chr.11: 4999039-4999039 on Build GRCh38
rs750132904 Chr.11: 4999068-4999068 on Build GRCh38
rs996957996 Chr.11: 4998944-4998944 on Build GRCh38
rs772916192 Chr.11: 4999173-4999173 on Build GRCh38
rs768140844 Chr.11: 4999033-4999033 on Build GRCh38
rs143037587 Chr.11: 4999352-4999352 on Build GRCh38
rs2445290 Chr.11: 4999186-4999186 on Build GRCh38
rs777300624 Chr.11: 4999346-4999346 on Build GRCh38
rs140344556 Chr.11: 4999035-4999035 on Build GRCh38
rs374482511 Chr.11: 4999228-4999228 on Build GRCh38
rs368384016 Chr.11: 4999312-4999312 on Build GRCh38
rs1003335436 Chr.11: 4999030-4999030 on Build GRCh38
rs772650638 Chr.11: 4999280-4999280 on Build GRCh38
rs61910724 Chr.11: 4999001-4999001 on Build GRCh38
rs771279270 Chr.11: 4999111-4999111 on Build GRCh38
rs750340820 Chr.11: 4999382-4999382 on Build GRCh38
rs748831457 Chr.11: 4999350-4999350 on Build GRCh38
rs868479858 Chr.11: 4999227-4999227 on Build GRCh38

View Full Product Details