Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6218832-6219353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
-
UniGene
Hs.278221
Gene Chromosome Location
Chr.11: 6202997-6226783 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Gene Symbol
FAM160A2
Entrez Gene ID
84067
Gene Name
family with sequence similarity 160 member A2
Gene Aliases
C11orf56
UniGene
Hs.501793
Gene Chromosome Location
Chr.11: 6211334-6234711 on Build GRCh38
Transcripts
XM_011519926.2 NM_001098794.1 NM_032127.3 XM_006718343.2 XM_011520397.2 XM_011520398.2 XM_011520399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5826993 c.1192-3_1192-2insC p.? Chr.11: 6219052-6219053 on Build GRCh38
6588107 c.1236C>T p.L412L Chr.11: 6219006-6219006 on Build GRCh38
6585625 c.1271+5_1271+6insAGGTATC p.? Chr.11: 6218965-6218966 on Build GRCh38
340035 c.1252C>G p.L418V Chr.11: 6218990-6218990 on Build GRCh38
6133708 c.1252C>T p.L418L Chr.11: 6218990-6218990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372584975 Chr.11: 6218941-6218941 on Build GRCh38
rs1000438109 Chr.11: 6219228-6219228 on Build GRCh38
rs996635541 Chr.11: 6218893-6218893 on Build GRCh38
rs926983957 Chr.11: 6219306-6219306 on Build GRCh38
rs113192632 Chr.11: 6219057-6219057 on Build GRCh38
rs779011170 Chr.11: 6219018-6219018 on Build GRCh38
rs763994886 Chr.11: 6218963-6218963 on Build GRCh38
rs538868950 Chr.11: 6219110-6219110 on Build GRCh38
rs767429737 Chr.11: 6218944-6218944 on Build GRCh38
rs756132827 Chr.11: 6218943-6218943 on Build GRCh38
rs937102326 Chr.11: 6219316-6219316 on Build GRCh38
rs745993384 Chr.11: 6218913-6218913 on Build GRCh38
rs566994676 Chr.11: 6218898-6218898 on Build GRCh38
rs576140330 Chr.11: 6219233-6219233 on Build GRCh38
rs1018997577 Chr.11: 6218855-6218855 on Build GRCh38
rs73392995 Chr.11: 6219128-6219128 on Build GRCh38
rs758549581 Chr.11: 6219034-6219034 on Build GRCh38
rs764000906 Chr.11: 6219074-6219074 on Build GRCh38
rs769045456 Chr.11: 6219048-6219048 on Build GRCh38
rs368281359 Chr.11: 6218951-6218951 on Build GRCh38
rs755286408 Chr.11: 6219118-6219118 on Build GRCh38
rs373260956 Chr.11: 6219220-6219220 on Build GRCh38
rs16910305 Chr.11: 6219287-6219287 on Build GRCh38
rs748263771 Chr.11: 6219189-6219189 on Build GRCh38
rs551678500 Chr.11: 6218896-6218896 on Build GRCh38
rs747434070 Chr.11: 6219046-6219046 on Build GRCh38
rs750570094 Chr.11: 6219084-6219084 on Build GRCh38
rs1018273919 Chr.11: 6219134-6219134 on Build GRCh38
rs556068801 Chr.11: 6219072-6219072 on Build GRCh38
rs1024000406 Chr.11: 6219242-6219242 on Build GRCh38
rs770448371 Chr.11: 6218921-6218921 on Build GRCh38
rs201559383 Chr.11: 6219065-6219065 on Build GRCh38
rs1047937356 Chr.11: 6219109-6219109 on Build GRCh38
rs1008893218 Chr.11: 6219124-6219124 on Build GRCh38
rs115108477 Chr.11: 6219096-6219096 on Build GRCh38
rs969724787 Chr.11: 6219273-6219273 on Build GRCh38
rs1022347132 Chr.11: 6219193-6219193 on Build GRCh38
rs757320813 Chr.11: 6219004-6219004 on Build GRCh38
rs750657567 Chr.11: 6219033-6219033 on Build GRCh38
rs138487643 Chr.11: 6219232-6219232 on Build GRCh38
rs201798905 Chr.11: 6218956-6218956 on Build GRCh38
rs183010034 Chr.11: 6219058-6219058 on Build GRCh38
rs983055654 Chr.11: 6218900-6218900 on Build GRCh38
rs868078243 Chr.11: 6219073-6219073 on Build GRCh38
rs758461507 Chr.11: 6219087-6219087 on Build GRCh38
rs996243065 Chr.11: 6219140-6219140 on Build GRCh38
rs964800541 Chr.11: 6218876-6218876 on Build GRCh38
rs781593048 Chr.11: 6219053-6219053 on Build GRCh38
rs760167191 Chr.11: 6219052-6219053 on Build GRCh38
rs900579240 Chr.11: 6219129-6219129 on Build GRCh38
rs941692324 Chr.11: 6219012-6219012 on Build GRCh38
rs998717170 Chr.11: 6219177-6219177 on Build GRCh38
rs899906572 Chr.11: 6219158-6219158 on Build GRCh38
rs376693585 Chr.11: 6219054-6219054 on Build GRCh38
rs777809731 Chr.11: 6219205-6219205 on Build GRCh38
rs527738340 Chr.11: 6218939-6218939 on Build GRCh38
rs765940206 Chr.11: 6219053-6219053 on Build GRCh38
rs17238727 Chr.11: 6219202-6219202 on Build GRCh38
rs375542465 Chr.11: 6219035-6219035 on Build GRCh38
rs771844879 Chr.11: 6219052-6219052 on Build GRCh38
rs760540227 Chr.11: 6218957-6218957 on Build GRCh38
rs753790891 Chr.11: 6218991-6218991 on Build GRCh38
rs963001227 Chr.11: 6219286-6219286 on Build GRCh38
rs772902652 Chr.11: 6218924-6218925 on Build GRCh38
rs780252040 Chr.11: 6219045-6219045 on Build GRCh38
rs73392996 Chr.11: 6219323-6219323 on Build GRCh38
rs930817706 Chr.11: 6219098-6219098 on Build GRCh38
rs992504244 Chr.11: 6219239-6219239 on Build GRCh38
rs751752273 Chr.11: 6219101-6219101 on Build GRCh38
rs753569407 Chr.11: 6218894-6218894 on Build GRCh38
rs12363184 Chr.11: 6218924-6218924 on Build GRCh38
rs765238167 Chr.11: 6219071-6219071 on Build GRCh38
rs572108100 Chr.11: 6219186-6219186 on Build GRCh38
rs922797096 Chr.11: 6218860-6218860 on Build GRCh38
rs773850615 Chr.11: 6219055-6219055 on Build GRCh38

View Full Product Details