Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34464027-34464543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAT
Entrez Gene ID
847
Gene Name
catalase
Gene Aliases
-
UniGene
Hs.502302
Gene Chromosome Location
Chr.11: 34438925-34472060 on Build GRCh38
Transcripts
NM_001752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4032532 c.1196-1G>T p.? Chr.11: 34464104-34464104 on Build GRCh38
5561051 c.1298A>G p.N433S Chr.11: 34464207-34464207 on Build GRCh38
3446970 c.1321A>T p.T441S Chr.11: 34464230-34464230 on Build GRCh38
1199692 c.1196-2A>G p.? Chr.11: 34464103-34464103 on Build GRCh38
7090246 c.1234C>A p.P412T Chr.11: 34464143-34464143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912087670 Chr.11: 34464362-34464362 on Build GRCh38
rs984012369 Chr.11: 34464346-34464346 on Build GRCh38
rs763932631 Chr.11: 34464174-34464174 on Build GRCh38
rs765139377 Chr.11: 34464063-34464063 on Build GRCh38
rs760302370 Chr.11: 34464263-34464263 on Build GRCh38
rs200213411 Chr.11: 34464207-34464207 on Build GRCh38
rs139875365 Chr.11: 34464197-34464197 on Build GRCh38
rs938348359 Chr.11: 34464324-34464324 on Build GRCh38
rs761019680 Chr.11: 34464246-34464246 on Build GRCh38
rs927059301 Chr.11: 34464261-34464261 on Build GRCh38
rs770216105 Chr.11: 34464144-34464144 on Build GRCh38
rs545807969 Chr.11: 34464329-34464329 on Build GRCh38
rs781427755 Chr.11: 34464198-34464198 on Build GRCh38
rs973049240 Chr.11: 34464313-34464313 on Build GRCh38
rs201105363 Chr.11: 34464127-34464127 on Build GRCh38
rs775035646 Chr.11: 34464232-34464232 on Build GRCh38
rs969565876 Chr.11: 34464487-34464487 on Build GRCh38
rs763698194 Chr.11: 34464272-34464272 on Build GRCh38
rs766785418 Chr.11: 34464342-34464342 on Build GRCh38
rs553130175 Chr.11: 34464455-34464455 on Build GRCh38
rs1054497847 Chr.11: 34464398-34464398 on Build GRCh38
rs704724 Chr.11: 34464164-34464164 on Build GRCh38
rs771347772 Chr.11: 34464219-34464219 on Build GRCh38
rs7126640 Chr.11: 34464470-34464470 on Build GRCh38
rs369392879 Chr.11: 34464176-34464176 on Build GRCh38
rs935949685 Chr.11: 34464396-34464396 on Build GRCh38
rs369039368 Chr.11: 34464090-34464090 on Build GRCh38
rs974996635 Chr.11: 34464376-34464376 on Build GRCh38
rs576962467 Chr.11: 34464327-34464327 on Build GRCh38
rs904371669 Chr.11: 34464377-34464377 on Build GRCh38
rs749574305 Chr.11: 34464215-34464215 on Build GRCh38
rs767118405 Chr.11: 34464067-34464067 on Build GRCh38
rs1015174215 Chr.11: 34464191-34464191 on Build GRCh38
rs140903091 Chr.11: 34464173-34464173 on Build GRCh38
rs201049382 Chr.11: 34464227-34464227 on Build GRCh38
rs890577449 Chr.11: 34464441-34464441 on Build GRCh38
rs770199564 Chr.11: 34464209-34464209 on Build GRCh38
rs900483030 Chr.11: 34464130-34464130 on Build GRCh38
rs752349991 Chr.11: 34464216-34464218 on Build GRCh38
rs763494494 Chr.11: 34464158-34464158 on Build GRCh38
rs200089024 Chr.11: 34464160-34464160 on Build GRCh38
rs753628651 Chr.11: 34464180-34464180 on Build GRCh38
rs1003485592 Chr.11: 34464230-34464230 on Build GRCh38
rs574257179 Chr.11: 34464395-34464395 on Build GRCh38
rs764364915 Chr.11: 34464289-34464289 on Build GRCh38
rs757190770 Chr.11: 34464193-34464193 on Build GRCh38
rs754311309 Chr.11: 34464372-34464372 on Build GRCh38
rs143563471 Chr.11: 34464132-34464132 on Build GRCh38
rs1014935201 Chr.11: 34464449-34464449 on Build GRCh38
rs752172460 Chr.11: 34464489-34464489 on Build GRCh38
rs752277640 Chr.11: 34464171-34464171 on Build GRCh38
rs1008866443 Chr.11: 34464448-34464448 on Build GRCh38
rs534846690 Chr.11: 34464099-34464099 on Build GRCh38
rs995112544 Chr.11: 34464481-34464481 on Build GRCh38
rs757195247 Chr.11: 34464108-34464108 on Build GRCh38
rs376840225 Chr.11: 34464161-34464161 on Build GRCh38
rs1042595689 Chr.11: 34464484-34464484 on Build GRCh38
rs950505199 Chr.11: 34464330-34464330 on Build GRCh38
rs562099712 Chr.11: 34464483-34464483 on Build GRCh38
rs945006829 Chr.11: 34464103-34464103 on Build GRCh38
rs867665733 Chr.11: 34464353-34464353 on Build GRCh38
rs145759100 Chr.11: 34464226-34464226 on Build GRCh38
rs768886685 Chr.11: 34464124-34464124 on Build GRCh38
rs774660725 Chr.11: 34464220-34464220 on Build GRCh38
rs780155754 Chr.11: 34464110-34464110 on Build GRCh38
rs753813511 Chr.11: 34464105-34464105 on Build GRCh38
rs945971158 Chr.11: 34464339-34464339 on Build GRCh38
rs959792435 Chr.11: 34464504-34464504 on Build GRCh38
rs554441051 Chr.11: 34464145-34464145 on Build GRCh38
rs1003798660 Chr.11: 34464494-34464494 on Build GRCh38
rs542422915 Chr.11: 34464479-34464479 on Build GRCh38
rs906342495 Chr.11: 34464492-34464492 on Build GRCh38
rs373692369 Chr.11: 34464195-34464195 on Build GRCh38
rs752371782 Chr.11: 34464076-34464076 on Build GRCh38

View Full Product Details