Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35619203-35619723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FJX1
Entrez Gene ID
24147
Gene Name
four jointed box 1
Gene Aliases
-
UniGene
Hs.39384
Gene Chromosome Location
Chr.11: 35618187-35620873 on Build GRCh38
Transcripts
NM_014344.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412676 c.668C>T p.A223V Chr.11: 35619304-35619304 on Build GRCh38
4808406 c.645G>A p.V215V Chr.11: 35619281-35619281 on Build GRCh38
4773485 c.606C>A p.G202G Chr.11: 35619242-35619242 on Build GRCh38
4777134 c.871G>C p.D291H Chr.11: 35619507-35619507 on Build GRCh38
6393049 c.701G>A p.W234* Chr.11: 35619337-35619337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776157019 Chr.11: 35619225-35619225 on Build GRCh38
rs773851065 Chr.11: 35619252-35619285 on Build GRCh38
rs371225997 Chr.11: 35619346-35619346 on Build GRCh38
rs1007452087 Chr.11: 35619341-35619341 on Build GRCh38
rs552298809 Chr.11: 35619335-35619335 on Build GRCh38
rs767460825 Chr.11: 35619386-35619386 on Build GRCh38
rs536959359 Chr.11: 35619626-35619626 on Build GRCh38
rs771598953 Chr.11: 35619605-35619605 on Build GRCh38
rs762629370 Chr.11: 35619366-35619366 on Build GRCh38
rs771587869 Chr.11: 35619434-35619434 on Build GRCh38
rs769184088 Chr.11: 35619326-35619326 on Build GRCh38
rs758563648 Chr.11: 35619401-35619401 on Build GRCh38
rs747342329 Chr.11: 35619406-35619406 on Build GRCh38
rs761248703 Chr.11: 35619272-35619283 on Build GRCh38
rs765190883 Chr.11: 35619629-35619629 on Build GRCh38
rs760487555 Chr.11: 35619448-35619448 on Build GRCh38
rs775788367 Chr.11: 35619305-35619305 on Build GRCh38
rs528278578 Chr.11: 35619451-35619451 on Build GRCh38
rs373983437 Chr.11: 35619674-35619674 on Build GRCh38
rs754874092 Chr.11: 35619375-35619375 on Build GRCh38
rs752983944 Chr.11: 35619574-35619574 on Build GRCh38
rs746774832 Chr.11: 35619618-35619618 on Build GRCh38
rs867504194 Chr.11: 35619237-35619237 on Build GRCh38
rs944722255 Chr.11: 35619571-35619571 on Build GRCh38
rs76580557 Chr.11: 35619520-35619520 on Build GRCh38
rs756545201 Chr.11: 35619578-35619578 on Build GRCh38
rs751664817 Chr.11: 35619563-35619563 on Build GRCh38
rs781345839 Chr.11: 35619492-35619492 on Build GRCh38
rs747959914 Chr.11: 35619690-35619690 on Build GRCh38
rs764499451 Chr.11: 35619652-35619652 on Build GRCh38
rs754281772 Chr.11: 35619656-35619656 on Build GRCh38
rs935819715 Chr.11: 35619287-35619287 on Build GRCh38
rs745974596 Chr.11: 35619400-35619400 on Build GRCh38
rs769699799 Chr.11: 35619697-35619697 on Build GRCh38
rs771785260 Chr.11: 35619538-35619538 on Build GRCh38
rs748578909 Chr.11: 35619494-35619494 on Build GRCh38
rs372820808 Chr.11: 35619365-35619365 on Build GRCh38
rs770151615 Chr.11: 35619497-35619497 on Build GRCh38
rs774192421 Chr.11: 35619437-35619437 on Build GRCh38
rs946222933 Chr.11: 35619678-35619678 on Build GRCh38
rs774137238 Chr.11: 35619533-35619533 on Build GRCh38
rs569715705 Chr.11: 35619703-35619703 on Build GRCh38
rs375472997 Chr.11: 35619461-35619461 on Build GRCh38
rs559530198 Chr.11: 35619390-35619390 on Build GRCh38
rs763987360 Chr.11: 35619546-35619546 on Build GRCh38
rs1018654723 Chr.11: 35619344-35619344 on Build GRCh38
rs1054248103 Chr.11: 35619290-35619290 on Build GRCh38
rs754926596 Chr.11: 35619277-35619277 on Build GRCh38
rs752538855 Chr.11: 35619447-35619447 on Build GRCh38
rs760901534 Chr.11: 35619449-35619449 on Build GRCh38
rs768302122 Chr.11: 35619676-35619676 on Build GRCh38
rs750108363 Chr.11: 35619242-35619242 on Build GRCh38
rs371151775 Chr.11: 35619558-35619558 on Build GRCh38
rs377652519 Chr.11: 35619542-35619542 on Build GRCh38
rs370548778 Chr.11: 35619600-35619600 on Build GRCh38
rs757992331 Chr.11: 35619666-35619666 on Build GRCh38
rs148087489 Chr.11: 35619583-35619583 on Build GRCh38
rs369246173 Chr.11: 35619356-35619356 on Build GRCh38
rs200130401 Chr.11: 35619645-35619645 on Build GRCh38
rs747393136 Chr.11: 35619306-35619306 on Build GRCh38
rs758255303 Chr.11: 35619243-35619243 on Build GRCh38
rs762575886 Chr.11: 35619426-35619426 on Build GRCh38
rs541864604 Chr.11: 35619278-35619278 on Build GRCh38
rs770688292 Chr.11: 35619434-35619434 on Build GRCh38
rs753839435 Chr.11: 35619551-35619551 on Build GRCh38
rs776978610 Chr.11: 35619489-35619489 on Build GRCh38
rs777044069 Chr.11: 35619330-35619330 on Build GRCh38
rs759712434 Chr.11: 35619637-35619637 on Build GRCh38
rs755117887 Chr.11: 35619566-35619566 on Build GRCh38
rs781499867 Chr.11: 35619567-35619567 on Build GRCh38
rs866361271 Chr.11: 35619598-35619598 on Build GRCh38
rs779520516 Chr.11: 35619659-35619659 on Build GRCh38
rs751835841 Chr.11: 35619487-35619487 on Build GRCh38
rs751352263 Chr.11: 35619374-35619374 on Build GRCh38
rs772863576 Chr.11: 35619358-35619358 on Build GRCh38
rs779557678 Chr.11: 35619672-35619672 on Build GRCh38
rs755239850 Chr.11: 35619488-35619488 on Build GRCh38
rs776424047 Chr.11: 35619624-35619624 on Build GRCh38
rs750476220 Chr.11: 35619467-35619467 on Build GRCh38
rs775244907 Chr.11: 35619608-35619608 on Build GRCh38
rs190229494 Chr.11: 35619255-35619255 on Build GRCh38
rs377240576 Chr.11: 35619368-35619368 on Build GRCh38
rs1034265952 Chr.11: 35619311-35619311 on Build GRCh38
rs201811393 Chr.11: 35619588-35619588 on Build GRCh38
rs779194365 Chr.11: 35619292-35619292 on Build GRCh38
rs764830840 Chr.11: 35619647-35619647 on Build GRCh38
rs77030854 Chr.11: 35619521-35619521 on Build GRCh38
rs778474284 Chr.11: 35619499-35619499 on Build GRCh38
rs773243175 Chr.11: 35619701-35619701 on Build GRCh38
rs772808472 Chr.11: 35619423-35619423 on Build GRCh38
rs761278273 Chr.11: 35619231-35619231 on Build GRCh38
rs748780489 Chr.11: 35619516-35619516 on Build GRCh38
rs774246802 Chr.11: 35619636-35619636 on Build GRCh38
rs780250139 Chr.11: 35619481-35619481 on Build GRCh38
rs767277196 Chr.11: 35619443-35619443 on Build GRCh38
rs935767201 Chr.11: 35619452-35619452 on Build GRCh38
rs370789301 Chr.11: 35619404-35619404 on Build GRCh38
rs781017522 Chr.11: 35619283-35619283 on Build GRCh38
rs776372343 Chr.11: 35619679-35619679 on Build GRCh38
rs200057673 Chr.11: 35619450-35619450 on Build GRCh38
rs768496322 Chr.11: 35619621-35619621 on Build GRCh38
rs745335153 Chr.11: 35619660-35619660 on Build GRCh38
rs950778818 Chr.11: 35619421-35619421 on Build GRCh38
rs757786737 Chr.11: 35619658-35619658 on Build GRCh38
rs768966225 Chr.11: 35619415-35619415 on Build GRCh38
rs749369349 Chr.11: 35619288-35619288 on Build GRCh38
rs767604436 Chr.11: 35619639-35619639 on Build GRCh38
rs548112170 Chr.11: 35619475-35619475 on Build GRCh38
rs887166378 Chr.11: 35619611-35619611 on Build GRCh38
rs917524425 Chr.11: 35619433-35619433 on Build GRCh38
rs781713757 Chr.11: 35619416-35619416 on Build GRCh38
rs757323111 Chr.11: 35619289-35619289 on Build GRCh38
rs770340270 Chr.11: 35619422-35619422 on Build GRCh38
rs752646867 Chr.11: 35619389-35619389 on Build GRCh38
rs998630452 Chr.11: 35619361-35619361 on Build GRCh38
rs772283158 Chr.11: 35619300-35619300 on Build GRCh38
rs778789924 Chr.11: 35619398-35619398 on Build GRCh38
rs375635974 Chr.11: 35619293-35619293 on Build GRCh38

View Full Product Details