Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57409034-57409529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC43A3
Entrez Gene ID
29015
Gene Name
solute carrier family 43 member 3
Gene Aliases
EEG1, FOAP-13, PRO1659, SEEEG-1
UniGene
Hs.99962
Gene Chromosome Location
Chr.11: 57406954-57427580 on Build GRCh38
Transcripts
NM_001278201.1 NM_001278206.1 NM_014096.3 NM_017611.2 NM_199329.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463693 c.1301C>T p.A434V Chr.11: 57409245-57409245 on Build GRCh38
1354769 c.1299G>A p.S433S Chr.11: 57409247-57409247 on Build GRCh38
6329241 c.1298C>T p.S433L Chr.11: 57409248-57409248 on Build GRCh38
4574486 c.1351C>T p.L451F Chr.11: 57409195-57409195 on Build GRCh38
4564536 c.1252_1253CC>TG p.P418C Chr.11: 57409293-57409294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748321685 Chr.11: 57409286-57409286 on Build GRCh38
rs760314789 Chr.11: 57409332-57409332 on Build GRCh38
rs897267938 Chr.11: 57409262-57409262 on Build GRCh38
rs968857804 Chr.11: 57409109-57409109 on Build GRCh38
rs184955878 Chr.11: 57409501-57409501 on Build GRCh38
rs768860779 Chr.11: 57409211-57409211 on Build GRCh38
rs202193567 Chr.11: 57409275-57409275 on Build GRCh38
rs921595835 Chr.11: 57409144-57409144 on Build GRCh38
rs755242132 Chr.11: 57409282-57409282 on Build GRCh38
rs758870554 Chr.11: 57409276-57409276 on Build GRCh38
rs781022603 Chr.11: 57409216-57409216 on Build GRCh38
rs781602865 Chr.11: 57409275-57409276 on Build GRCh38
rs772247461 Chr.11: 57409344-57409344 on Build GRCh38
rs762402264 Chr.11: 57409141-57409141 on Build GRCh38
rs763429441 Chr.11: 57409350-57409350 on Build GRCh38
rs374225715 Chr.11: 57409341-57409341 on Build GRCh38
rs540423539 Chr.11: 57409467-57409467 on Build GRCh38
rs746216412 Chr.11: 57409281-57409282 on Build GRCh38
rs760383396 Chr.11: 57409175-57409175 on Build GRCh38
rs367939252 Chr.11: 57409312-57409312 on Build GRCh38
rs951270660 Chr.11: 57409242-57409242 on Build GRCh38
rs557692341 Chr.11: 57409137-57409137 on Build GRCh38
rs138765536 Chr.11: 57409231-57409231 on Build GRCh38
rs1036587497 Chr.11: 57409240-57409240 on Build GRCh38
rs187399098 Chr.11: 57409261-57409261 on Build GRCh38
rs200096700 Chr.11: 57409197-57409197 on Build GRCh38
rs564534137 Chr.11: 57409382-57409382 on Build GRCh38
rs113836199 Chr.11: 57409383-57409383 on Build GRCh38
rs1035880706 Chr.11: 57409370-57409370 on Build GRCh38
rs981760280 Chr.11: 57409065-57409065 on Build GRCh38
rs768842280 Chr.11: 57409117-57409117 on Build GRCh38
rs774057594 Chr.11: 57409302-57409302 on Build GRCh38
rs952996969 Chr.11: 57409496-57409496 on Build GRCh38
rs773996402 Chr.11: 57409254-57409254 on Build GRCh38
rs183113015 Chr.11: 57409174-57409174 on Build GRCh38
rs576854610 Chr.11: 57409165-57409165 on Build GRCh38
rs749254089 Chr.11: 57409294-57409294 on Build GRCh38
rs754244875 Chr.11: 57409159-57409159 on Build GRCh38
rs761181516 Chr.11: 57409460-57409460 on Build GRCh38
rs769491369 Chr.11: 57409290-57409290 on Build GRCh38
rs373652452 Chr.11: 57409247-57409247 on Build GRCh38
rs780120974 Chr.11: 57409186-57409186 on Build GRCh38
rs781424633 Chr.11: 57409283-57409283 on Build GRCh38
rs1010431165 Chr.11: 57409388-57409388 on Build GRCh38
rs751186459 Chr.11: 57409147-57409147 on Build GRCh38
rs777559188 Chr.11: 57409292-57409292 on Build GRCh38
rs374023369 Chr.11: 57409306-57409306 on Build GRCh38
rs928858381 Chr.11: 57409104-57409104 on Build GRCh38
rs758427634 Chr.11: 57409185-57409185 on Build GRCh38
rs770891887 Chr.11: 57409295-57409295 on Build GRCh38
rs754612836 Chr.11: 57409151-57409151 on Build GRCh38
rs780063036 Chr.11: 57409280-57409280 on Build GRCh38
rs1027337305 Chr.11: 57409500-57409500 on Build GRCh38
rs773046546 Chr.11: 57409239-57409239 on Build GRCh38
rs193012070 Chr.11: 57409499-57409499 on Build GRCh38
rs759323526 Chr.11: 57409257-57409257 on Build GRCh38
rs747916405 Chr.11: 57409223-57409223 on Build GRCh38
rs762112096 Chr.11: 57409264-57409264 on Build GRCh38
rs1028671613 Chr.11: 57409480-57409480 on Build GRCh38
rs573981566 Chr.11: 57409250-57409250 on Build GRCh38
rs968536047 Chr.11: 57409408-57409408 on Build GRCh38
rs765584516 Chr.11: 57409273-57409273 on Build GRCh38
rs775301349 Chr.11: 57409328-57409328 on Build GRCh38
rs2241899 Chr.11: 57409155-57409155 on Build GRCh38
rs541887281 Chr.11: 57409259-57409259 on Build GRCh38
rs911743960 Chr.11: 57409157-57409157 on Build GRCh38
rs772819040 Chr.11: 57409140-57409140 on Build GRCh38
rs1020861751 Chr.11: 57409407-57409407 on Build GRCh38
rs761804568 Chr.11: 57409124-57409124 on Build GRCh38
rs765864990 Chr.11: 57409142-57409142 on Build GRCh38

View Full Product Details