Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64317366-64317873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT112
Entrez Gene ID
51504
Gene Name
tRNA methyltransferase 11-2 homolog (S. cerevisiae)
Gene Aliases
HSPC152, HSPC170, TRM112, TRMT11-2, hTrm112
UniGene
Hs.333579
Gene Chromosome Location
Chr.11: 64316492-64318084 on Build GRCh38
Transcripts
NM_001286082.1 NM_001286084.1 NM_016404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5990999 c.17A>G p.H6R Chr.11: 64317510-64317510 on Build GRCh38
3719457 c.42G>A p.R14R Chr.11: 64317485-64317485 on Build GRCh38
5033564 c.78+3A>C p.? Chr.11: 64317446-64317446 on Build GRCh38
1628146 c.20A>G p.N7S Chr.11: 64317507-64317507 on Build GRCh38
2163447 c.46delG p.V16fs*30 Chr.11: 64317481-64317481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375491426 Chr.11: 64317413-64317413 on Build GRCh38
rs559661851 Chr.11: 64317576-64317576 on Build GRCh38
rs929290395 Chr.11: 64317679-64317679 on Build GRCh38
rs1035093927 Chr.11: 64317817-64317817 on Build GRCh38
rs929546115 Chr.11: 64317839-64317839 on Build GRCh38
rs777383647 Chr.11: 64317440-64317440 on Build GRCh38
rs1000039151 Chr.11: 64317749-64317749 on Build GRCh38
rs1010796884 Chr.11: 64317788-64317788 on Build GRCh38
rs367914811 Chr.11: 64317575-64317575 on Build GRCh38
rs1044184981 Chr.11: 64317511-64317511 on Build GRCh38
rs1037936318 Chr.11: 64317660-64317660 on Build GRCh38
rs377669870 Chr.11: 64317506-64317506 on Build GRCh38
rs763156167 Chr.11: 64317403-64317403 on Build GRCh38
rs35573459 Chr.11: 64317561-64317561 on Build GRCh38
rs1020829484 Chr.11: 64317792-64317792 on Build GRCh38
rs748922321 Chr.11: 64317556-64317556 on Build GRCh38
rs780565402 Chr.11: 64317421-64317421 on Build GRCh38
rs942274975 Chr.11: 64317655-64317655 on Build GRCh38
rs28364831 Chr.11: 64317676-64317676 on Build GRCh38
rs747870648 Chr.11: 64317436-64317436 on Build GRCh38
rs968285613 Chr.11: 64317430-64317430 on Build GRCh38
rs745863361 Chr.11: 64317499-64317499 on Build GRCh38
rs994009920 Chr.11: 64317532-64317532 on Build GRCh38
rs749899733 Chr.11: 64317549-64317549 on Build GRCh38
rs200904021 Chr.11: 64317487-64317487 on Build GRCh38
rs577224688 Chr.11: 64317538-64317538 on Build GRCh38
rs751566570 Chr.11: 64317409-64317409 on Build GRCh38
rs960896964 Chr.11: 64317752-64317752 on Build GRCh38
rs979763593 Chr.11: 64317799-64317799 on Build GRCh38
rs916070833 Chr.11: 64317653-64317653 on Build GRCh38
rs1036801217 Chr.11: 64317660-64317661 on Build GRCh38
rs750834611 Chr.11: 64317416-64317416 on Build GRCh38
rs779914397 Chr.11: 64317555-64317555 on Build GRCh38
rs535716108 Chr.11: 64317486-64317486 on Build GRCh38
rs756072304 Chr.11: 64317554-64317554 on Build GRCh38
rs889705025 Chr.11: 64317711-64317711 on Build GRCh38
rs916101976 Chr.11: 64317822-64317822 on Build GRCh38
rs770924814 Chr.11: 64317462-64317462 on Build GRCh38
rs762738556 Chr.11: 64317480-64317481 on Build GRCh38
rs368657247 Chr.11: 64317595-64317595 on Build GRCh38
rs758101903 Chr.11: 64317439-64317439 on Build GRCh38
rs200065720 Chr.11: 64317808-64317808 on Build GRCh38
rs756469761 Chr.11: 64317515-64317515 on Build GRCh38
rs970051022 Chr.11: 64317797-64317797 on Build GRCh38
rs756653486 Chr.11: 64317491-64317491 on Build GRCh38
rs574861213 Chr.11: 64317497-64317497 on Build GRCh38
rs41294430 Chr.11: 64317534-64317534 on Build GRCh38
rs9787810 Chr.11: 64317826-64317826 on Build GRCh38
rs200361095 Chr.11: 64317526-64317526 on Build GRCh38
rs960927851 Chr.11: 64317592-64317592 on Build GRCh38
rs909556802 Chr.11: 64317807-64317807 on Build GRCh38
rs759763606 Chr.11: 64317522-64317522 on Build GRCh38
rs371811984 Chr.11: 64317559-64317559 on Build GRCh38
rs991693491 Chr.11: 64317639-64317639 on Build GRCh38
rs1007581684 Chr.11: 64317732-64317732 on Build GRCh38
rs938822025 Chr.11: 64317461-64317461 on Build GRCh38
rs10128595 Chr.11: 64317584-64317584 on Build GRCh38
rs773457282 Chr.11: 64317562-64317562 on Build GRCh38
rs769642779 Chr.11: 64317480-64317480 on Build GRCh38
rs573216664 Chr.11: 64317579-64317579 on Build GRCh38
rs981855947 Chr.11: 64317411-64317411 on Build GRCh38
rs775252346 Chr.11: 64317482-64317482 on Build GRCh38
rs749039296 Chr.11: 64317507-64317507 on Build GRCh38
rs539630674 Chr.11: 64317387-64317387 on Build GRCh38
rs775436432 Chr.11: 64317460-64317460 on Build GRCh38
rs942632712 Chr.11: 64317835-64317835 on Build GRCh38
rs183123201 Chr.11: 64317820-64317820 on Build GRCh38
rs760432232 Chr.11: 64317766-64317766 on Build GRCh38
rs1056063179 Chr.11: 64317475-64317475 on Build GRCh38
rs4930698 Chr.11: 64317591-64317591 on Build GRCh38
rs776522864 Chr.11: 64317398-64317398 on Build GRCh38
rs768404245 Chr.11: 64317557-64317557 on Build GRCh38
rs918448456 Chr.11: 64317669-64317669 on Build GRCh38
rs552847770 Chr.11: 64317401-64317401 on Build GRCh38
rs774517266 Chr.11: 64317483-64317483 on Build GRCh38
rs28699214 Chr.11: 64317412-64317412 on Build GRCh38
rs370461838 Chr.11: 64317512-64317512 on Build GRCh38
rs768943059 Chr.11: 64317508-64317508 on Build GRCh38
rs41294434 Chr.11: 64317638-64317638 on Build GRCh38
rs757820595 Chr.11: 64317410-64317410 on Build GRCh38
rs941163341 Chr.11: 64317514-64317514 on Build GRCh38
rs759280010 Chr.11: 64317400-64317400 on Build GRCh38
rs764289286 Chr.11: 64317405-64317405 on Build GRCh38
rs41294432 Chr.11: 64317609-64317609 on Build GRCh38
rs41294436 Chr.11: 64317724-64317724 on Build GRCh38
rs367624863 Chr.11: 64317558-64317558 on Build GRCh38
rs562975998 Chr.11: 64317530-64317530 on Build GRCh38
rs746464116 Chr.11: 64317445-64317445 on Build GRCh38
rs754279340 Chr.11: 64317521-64317521 on Build GRCh38
rs138907479 Chr.11: 64317460-64317460 on Build GRCh38
rs745665266 Chr.11: 64317479-64317479 on Build GRCh38
rs45462802 Chr.11: 64317766-64317767 on Build GRCh38
rs761305189 Chr.11: 64317517-64317517 on Build GRCh38
rs766953448 Chr.11: 64317518-64317518 on Build GRCh38
rs963502122 Chr.11: 64317823-64317823 on Build GRCh38
rs574303235 Chr.11: 64317699-64317699 on Build GRCh38
rs770744162 Chr.11: 64317392-64317392 on Build GRCh38
rs959131801 Chr.11: 64317819-64317819 on Build GRCh38
rs1013840741 Chr.11: 64317758-64317759 on Build GRCh38
rs546791547 Chr.11: 64317745-64317745 on Build GRCh38
rs1030379288 Chr.11: 64317533-64317533 on Build GRCh38
rs566781863 Chr.11: 64317419-64317419 on Build GRCh38
rs776373517 Chr.11: 64317471-64317471 on Build GRCh38
rs780435157 Chr.11: 64317510-64317510 on Build GRCh38
rs981832997 Chr.11: 64317813-64317813 on Build GRCh38
rs550819 Chr.11: 64317602-64317602 on Build GRCh38
rs751041580 Chr.11: 64317484-64317484 on Build GRCh38
rs910462138 Chr.11: 64317446-64317446 on Build GRCh38
rs763716858 Chr.11: 64317481-64317481 on Build GRCh38
rs893611045 Chr.11: 64317750-64317750 on Build GRCh38
rs913551366 Chr.11: 64317394-64317394 on Build GRCh38
rs375653671 Chr.11: 64317485-64317485 on Build GRCh38
rs919553398 Chr.11: 64317834-64317834 on Build GRCh38
rs992437050 Chr.11: 64317594-64317594 on Build GRCh38
rs1037280445 Chr.11: 64317515-64317516 on Build GRCh38
rs941120530 Chr.11: 64317647-64317647 on Build GRCh38

View Full Product Details