Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185292326-185292813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K13
Entrez Gene ID
9175
Gene Name
mitogen-activated protein kinase kinase kinase 13
Gene Aliases
LZK, MEKK13, MLK
UniGene
Hs.591306
Gene Chromosome Location
Chr.3: 185282938-185489094 on Build GRCh38
Transcripts
NM_001242314.1 NM_001242317.1 NR_038322.1 XM_017007456.1 XM_017007458.1

SNP Detail

SNP ID Chromosome Location
rs1132499 Chr.3: 185292492-185292492 on Build GRCh38
rs932364168 Chr.3: 185292379-185292379 on Build GRCh38
rs891822808 Chr.3: 185292488-185292488 on Build GRCh38
rs773006664 Chr.3: 185292677-185292677 on Build GRCh38
rs552999785 Chr.3: 185292718-185292718 on Build GRCh38
rs891728260 Chr.3: 185292779-185292779 on Build GRCh38
rs879217210 Chr.3: 185292617-185292617 on Build GRCh38
rs73887858 Chr.3: 185292508-185292508 on Build GRCh38
rs1015165667 Chr.3: 185292573-185292573 on Build GRCh38
rs1001988251 Chr.3: 185292522-185292522 on Build GRCh38
rs947260109 Chr.3: 185292782-185292782 on Build GRCh38
rs1035901372 Chr.3: 185292535-185292536 on Build GRCh38
rs547888353 Chr.3: 185292632-185292632 on Build GRCh38
rs570741575 Chr.3: 185292699-185292699 on Build GRCh38
rs534559146 Chr.3: 185292704-185292704 on Build GRCh38
rs963938729 Chr.3: 185292582-185292582 on Build GRCh38
rs191586560 Chr.3: 185292453-185292453 on Build GRCh38
rs1022015935 Chr.3: 185292495-185292495 on Build GRCh38
rs530419692 Chr.3: 185292475-185292475 on Build GRCh38
rs139544887 Chr.3: 185292354-185292354 on Build GRCh38
rs570217915 Chr.3: 185292538-185292538 on Build GRCh38
rs943857062 Chr.3: 185292743-185292743 on Build GRCh38
rs778461288 Chr.3: 185292515-185292515 on Build GRCh38
rs943996155 Chr.3: 185292476-185292476 on Build GRCh38
rs746868222 Chr.3: 185292724-185292724 on Build GRCh38
rs537264280 Chr.3: 185292541-185292541 on Build GRCh38
rs547395268 Chr.3: 185292396-185292396 on Build GRCh38
rs983731660 Chr.3: 185292571-185292571 on Build GRCh38
rs868238825 Chr.3: 185292411-185292411 on Build GRCh38
rs919550808 Chr.3: 185292609-185292609 on Build GRCh38
rs781607086 Chr.3: 185292413-185292413 on Build GRCh38
rs890918995 Chr.3: 185292458-185292458 on Build GRCh38
rs1052953852 Chr.3: 185292477-185292477 on Build GRCh38
rs974250346 Chr.3: 185292352-185292352 on Build GRCh38
rs558658638 Chr.3: 185292613-185292613 on Build GRCh38
rs973776910 Chr.3: 185292599-185292599 on Build GRCh38
rs183492013 Chr.3: 185292747-185292747 on Build GRCh38
rs919703158 Chr.3: 185292361-185292361 on Build GRCh38
rs746381564 Chr.3: 185292507-185292507 on Build GRCh38
rs747818322 Chr.3: 185292531-185292531 on Build GRCh38

View Full Product Details