Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75503722-75504239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRR1
Entrez Gene ID
11103
Gene Name
KRR1, small subunit processome component homolog
Gene Aliases
HRB2, RIP-1
UniGene
Hs.645517
Gene Chromosome Location
Chr.12: 75497639-75511638 on Build GRCh38
Transcripts
NM_007043.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1256070 c.793_794GA>TT p.E265L Chr.12: 75503941-75503942 on Build GRCh38
2102509 c.712A>T p.S238C Chr.12: 75504023-75504023 on Build GRCh38
2102506 c.818A>G p.Q273R Chr.12: 75503917-75503917 on Build GRCh38
1242446 c.736T>G p.F246V Chr.12: 75503999-75503999 on Build GRCh38
4696749 c.781delA p.T261fs*29 Chr.12: 75503954-75503954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371114324 Chr.12: 75503877-75503877 on Build GRCh38
rs17853240 Chr.12: 75503912-75503912 on Build GRCh38
rs551077175 Chr.12: 75503935-75503935 on Build GRCh38
rs747031287 Chr.12: 75504015-75504016 on Build GRCh38
rs892211632 Chr.12: 75504101-75504101 on Build GRCh38
rs556891574 Chr.12: 75504207-75504207 on Build GRCh38
rs957063484 Chr.12: 75503746-75503746 on Build GRCh38
rs901056089 Chr.12: 75504032-75504032 on Build GRCh38
rs571236018 Chr.12: 75504176-75504176 on Build GRCh38
rs183939405 Chr.12: 75503964-75503964 on Build GRCh38
rs1009305113 Chr.12: 75504120-75504120 on Build GRCh38
rs543269519 Chr.12: 75503780-75503780 on Build GRCh38
rs112004570 Chr.12: 75503934-75503934 on Build GRCh38
rs773976000 Chr.12: 75503955-75503955 on Build GRCh38
rs573629583 Chr.12: 75503796-75503796 on Build GRCh38
rs188885620 Chr.12: 75504031-75504031 on Build GRCh38
rs761971509 Chr.12: 75503980-75503980 on Build GRCh38
rs773580860 Chr.12: 75504076-75504077 on Build GRCh38
rs931346692 Chr.12: 75503835-75503835 on Build GRCh38
rs376593401 Chr.12: 75504055-75504055 on Build GRCh38
rs777235058 Chr.12: 75503978-75503978 on Build GRCh38
rs1023303797 Chr.12: 75504086-75504086 on Build GRCh38
rs551345271 Chr.12: 75504143-75504143 on Build GRCh38
rs778747459 Chr.12: 75503925-75503926 on Build GRCh38
rs757460554 Chr.12: 75503878-75503878 on Build GRCh38
rs748720677 Chr.12: 75503938-75503938 on Build GRCh38
rs777000919 Chr.12: 75504096-75504096 on Build GRCh38
rs372122256 Chr.12: 75504079-75504079 on Build GRCh38
rs559967256 Chr.12: 75503795-75503795 on Build GRCh38
rs536612778 Chr.12: 75504190-75504190 on Build GRCh38
rs375763606 Chr.12: 75504089-75504089 on Build GRCh38
rs779999846 Chr.12: 75504012-75504013 on Build GRCh38
rs781141506 Chr.12: 75503925-75503925 on Build GRCh38
rs752199746 Chr.12: 75503940-75503943 on Build GRCh38
rs75910030 Chr.12: 75504126-75504126 on Build GRCh38
rs1050532182 Chr.12: 75503941-75503941 on Build GRCh38
rs1036196546 Chr.12: 75503919-75503919 on Build GRCh38
rs374731315 Chr.12: 75503931-75503931 on Build GRCh38
rs765481167 Chr.12: 75503992-75503992 on Build GRCh38
rs1697751 Chr.12: 75503802-75503802 on Build GRCh38
rs12578286 Chr.12: 75503882-75503882 on Build GRCh38
rs988437031 Chr.12: 75504204-75504204 on Build GRCh38
rs940122183 Chr.12: 75503914-75503914 on Build GRCh38
rs367586880 Chr.12: 75504078-75504078 on Build GRCh38
rs762416932 Chr.12: 75504097-75504097 on Build GRCh38
rs547455750 Chr.12: 75503782-75503783 on Build GRCh38
rs749302630 Chr.12: 75504075-75504075 on Build GRCh38
rs772354362 Chr.12: 75503966-75503966 on Build GRCh38
rs758604785 Chr.12: 75503954-75503956 on Build GRCh38
rs997029796 Chr.12: 75504156-75504156 on Build GRCh38
rs200628608 Chr.12: 75503783-75503784 on Build GRCh38
rs781710988 Chr.12: 75504073-75504074 on Build GRCh38
rs763693574 Chr.12: 75504080-75504081 on Build GRCh38
rs139792543 Chr.12: 75503797-75503798 on Build GRCh38
rs767374508 Chr.12: 75504048-75504048 on Build GRCh38
rs372563816 Chr.12: 75504042-75504042 on Build GRCh38
rs368830982 Chr.12: 75503975-75503975 on Build GRCh38
rs17853239 Chr.12: 75503995-75503995 on Build GRCh38
rs565535318 Chr.12: 75504084-75504084 on Build GRCh38
rs761089940 Chr.12: 75503859-75503859 on Build GRCh38
rs745463656 Chr.12: 75504080-75504080 on Build GRCh38
rs775728423 Chr.12: 75503971-75503971 on Build GRCh38
rs750732688 Chr.12: 75503916-75503916 on Build GRCh38
rs765352026 Chr.12: 75504099-75504099 on Build GRCh38
rs988314612 Chr.12: 75503766-75503766 on Build GRCh38
rs921224283 Chr.12: 75503810-75503810 on Build GRCh38
rs200185870 Chr.12: 75503872-75503872 on Build GRCh38
rs761193901 Chr.12: 75503974-75503974 on Build GRCh38
rs1053012073 Chr.12: 75503850-75503851 on Build GRCh38
rs768705624 Chr.12: 75504044-75504044 on Build GRCh38
rs983864632 Chr.12: 75503775-75503775 on Build GRCh38
rs770108836 Chr.12: 75504075-75504076 on Build GRCh38
rs200548830 Chr.12: 75503755-75503755 on Build GRCh38
rs770472297 Chr.12: 75503952-75503952 on Build GRCh38
rs201035934 Chr.12: 75504068-75504068 on Build GRCh38
rs759146099 Chr.12: 75504077-75504078 on Build GRCh38
rs1005257012 Chr.12: 75504082-75504082 on Build GRCh38
rs771733195 Chr.12: 75504081-75504081 on Build GRCh38
rs748618258 Chr.12: 75504074-75504075 on Build GRCh38
rs147480080 Chr.12: 75504008-75504008 on Build GRCh38
rs570456869 Chr.12: 75503796-75503797 on Build GRCh38

View Full Product Details