Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120356793-120357336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
-
UniGene
Hs.158311
Gene Chromosome Location
Chr.12: 120339655-120369180 on Build GRCh38
Transcripts
NM_002442.3 XM_006719403.3 XM_006719404.3 XM_011538361.2 XM_011538362.2 XM_011538363.2 XM_011538364.2 XM_011538365.2 XM_011538366.2 XM_011538368.2 XM_011538370.2 XM_011538371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5729077 c.564G>A p.E188E Chr.12: 120356990-120356990 on Build GRCh38
6454315 c.644G>A p.G215D Chr.12: 120356910-120356910 on Build GRCh38
3457040 c.652+1G>A p.? Chr.12: 120356901-120356901 on Build GRCh38
2175176 c.578C>T p.T193M Chr.12: 120356976-120356976 on Build GRCh38
3368679 c.549A>C p.K183N Chr.12: 120357005-120357005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181246917 Chr.12: 120356886-120356886 on Build GRCh38
rs377467873 Chr.12: 120356885-120356885 on Build GRCh38
rs375135447 Chr.12: 120356982-120356982 on Build GRCh38
rs577553259 Chr.12: 120357202-120357202 on Build GRCh38
rs774866690 Chr.12: 120356907-120356907 on Build GRCh38
rs750439002 Chr.12: 120356965-120356965 on Build GRCh38
rs1010584887 Chr.12: 120357187-120357187 on Build GRCh38
rs756401860 Chr.12: 120356865-120356865 on Build GRCh38
rs779869447 Chr.12: 120356968-120356968 on Build GRCh38
rs142499554 Chr.12: 120357269-120357269 on Build GRCh38
rs757182790 Chr.12: 120356874-120356874 on Build GRCh38
rs746446964 Chr.12: 120357004-120357004 on Build GRCh38
rs1042102617 Chr.12: 120357286-120357286 on Build GRCh38
rs772595393 Chr.12: 120356984-120356984 on Build GRCh38
rs907869051 Chr.12: 120357089-120357089 on Build GRCh38
rs1179441 Chr.12: 120356850-120356850 on Build GRCh38
rs964834283 Chr.12: 120357236-120357236 on Build GRCh38
rs112963469 Chr.12: 120357215-120357215 on Build GRCh38
rs1009853474 Chr.12: 120356959-120356960 on Build GRCh38
rs768755542 Chr.12: 120356833-120356833 on Build GRCh38
rs1020607614 Chr.12: 120356915-120356915 on Build GRCh38
rs145402971 Chr.12: 120356894-120356894 on Build GRCh38
rs776037606 Chr.12: 120357012-120357012 on Build GRCh38
rs753553581 Chr.12: 120357245-120357245 on Build GRCh38
rs555207756 Chr.12: 120356964-120356964 on Build GRCh38
rs200525271 Chr.12: 120356930-120356930 on Build GRCh38
rs374684966 Chr.12: 120357037-120357037 on Build GRCh38
rs908466021 Chr.12: 120356997-120356997 on Build GRCh38
rs983391735 Chr.12: 120356819-120356819 on Build GRCh38
rs201101688 Chr.12: 120356879-120356879 on Build GRCh38
rs75437054 Chr.12: 120357203-120357203 on Build GRCh38
rs963529156 Chr.12: 120357127-120357127 on Build GRCh38
rs368814174 Chr.12: 120357041-120357041 on Build GRCh38
rs76006633 Chr.12: 120357153-120357153 on Build GRCh38
rs565598252 Chr.12: 120357155-120357155 on Build GRCh38
rs973893935 Chr.12: 120357146-120357146 on Build GRCh38
rs745663274 Chr.12: 120356878-120356878 on Build GRCh38
rs1021225035 Chr.12: 120356840-120356840 on Build GRCh38
rs780777611 Chr.12: 120356977-120356977 on Build GRCh38
rs145944432 Chr.12: 120356939-120356939 on Build GRCh38
rs566082242 Chr.12: 120356912-120356912 on Build GRCh38
rs575741004 Chr.12: 120357182-120357183 on Build GRCh38
rs751213592 Chr.12: 120356877-120356885 on Build GRCh38
rs751006130 Chr.12: 120357291-120357291 on Build GRCh38
rs541844330 Chr.12: 120357257-120357257 on Build GRCh38
rs768324684 Chr.12: 120356976-120356976 on Build GRCh38
rs753100021 Chr.12: 120356927-120356927 on Build GRCh38
rs561745022 Chr.12: 120357220-120357228 on Build GRCh38
rs752902077 Chr.12: 120356851-120356851 on Build GRCh38
rs573240902 Chr.12: 120357242-120357242 on Build GRCh38
rs769508876 Chr.12: 120356933-120356933 on Build GRCh38
rs932149701 Chr.12: 120357186-120357186 on Build GRCh38
rs545373141 Chr.12: 120356981-120356981 on Build GRCh38
rs759826798 Chr.12: 120356911-120356911 on Build GRCh38
rs768063621 Chr.12: 120356920-120356920 on Build GRCh38
rs1000499842 Chr.12: 120357297-120357297 on Build GRCh38
rs183878451 Chr.12: 120357056-120357056 on Build GRCh38
rs377613546 Chr.12: 120356895-120356895 on Build GRCh38
rs776844870 Chr.12: 120357038-120357038 on Build GRCh38
rs370653775 Chr.12: 120356896-120356896 on Build GRCh38
rs757303115 Chr.12: 120356960-120356960 on Build GRCh38
rs201781664 Chr.12: 120356870-120356870 on Build GRCh38
rs771367942 Chr.12: 120356898-120356898 on Build GRCh38
rs199780425 Chr.12: 120357036-120357036 on Build GRCh38
rs778059151 Chr.12: 120356866-120356866 on Build GRCh38
rs1019103698 Chr.12: 120357234-120357234 on Build GRCh38
rs367765145 Chr.12: 120357008-120357008 on Build GRCh38
rs765414173 Chr.12: 120357049-120357049 on Build GRCh38
rs1037897150 Chr.12: 120356877-120356877 on Build GRCh38
rs959472351 Chr.12: 120357029-120357029 on Build GRCh38
rs142127614 Chr.12: 120356975-120356975 on Build GRCh38

View Full Product Details