Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19216628-19217171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC1
Entrez Gene ID
23065
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
CAVIPMR, KIAA0090
UniGene
Hs.439200
Gene Chromosome Location
Chr.1: 19215664-19251559 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 NR_135114.1 NR_135115.1

Gene Symbol
LOC101927895
Entrez Gene ID
101927895
Gene Name
uncharacterized LOC101927895
Gene Aliases
-
UniGene
Hs.660534
Gene Chromosome Location
Chr.1: 19210386-19240704 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 NR_135114.1 NR_135115.1

SNP Detail

SNP ID Chromosome Location
rs374728161 Chr.1: 19217132-19217133 on Build GRCh38
rs920866350 Chr.1: 19217131-19217132 on Build GRCh38
rs534037230 Chr.1: 19217110-19217110 on Build GRCh38
rs1002993549 Chr.1: 19216968-19216968 on Build GRCh38
rs907294339 Chr.1: 19216960-19216960 on Build GRCh38
rs900711805 Chr.1: 19216761-19216761 on Build GRCh38
rs139019717 Chr.1: 19216700-19216700 on Build GRCh38
rs746215988 Chr.1: 19216981-19216981 on Build GRCh38
rs185957311 Chr.1: 19216974-19216974 on Build GRCh38
rs929537727 Chr.1: 19217139-19217139 on Build GRCh38
rs573450358 Chr.1: 19217116-19217116 on Build GRCh38
rs938305863 Chr.1: 19216717-19216717 on Build GRCh38
rs12756656 Chr.1: 19217117-19217117 on Build GRCh38
rs970485232 Chr.1: 19217076-19217078 on Build GRCh38
rs890549716 Chr.1: 19216817-19216817 on Build GRCh38
rs536708233 Chr.1: 19216927-19216927 on Build GRCh38
rs893374928 Chr.1: 19216867-19216867 on Build GRCh38
rs540608114 Chr.1: 19217131-19217131 on Build GRCh38
rs1008949798 Chr.1: 19216823-19216823 on Build GRCh38
rs981824326 Chr.1: 19217081-19217081 on Build GRCh38
rs938169755 Chr.1: 19217124-19217124 on Build GRCh38
rs183106225 Chr.1: 19216898-19216898 on Build GRCh38
rs764618178 Chr.1: 19216689-19216694 on Build GRCh38
rs373592561 Chr.1: 19216982-19216982 on Build GRCh38
rs1034433005 Chr.1: 19216972-19216972 on Build GRCh38
rs557027457 Chr.1: 19216852-19216852 on Build GRCh38
rs951309365 Chr.1: 19217002-19217002 on Build GRCh38
rs770604036 Chr.1: 19216904-19216904 on Build GRCh38
rs899195364 Chr.1: 19216758-19216758 on Build GRCh38
rs943748794 Chr.1: 19216664-19216664 on Build GRCh38
rs1040525199 Chr.1: 19216671-19216671 on Build GRCh38
rs190344789 Chr.1: 19216802-19216802 on Build GRCh38
rs779604597 Chr.1: 19217001-19217001 on Build GRCh38
rs1038341810 Chr.1: 19216754-19216754 on Build GRCh38
rs1046557245 Chr.1: 19216663-19216663 on Build GRCh38
rs963869296 Chr.1: 19216865-19216865 on Build GRCh38
rs751589363 Chr.1: 19216724-19216724 on Build GRCh38
rs878875326 Chr.1: 19216688-19216688 on Build GRCh38
rs993920106 Chr.1: 19216760-19216760 on Build GRCh38
rs571593641 Chr.1: 19216789-19216789 on Build GRCh38
rs80338328 Chr.1: 19216737-19216737 on Build GRCh38
rs777198992 Chr.1: 19216878-19216878 on Build GRCh38
rs982855723 Chr.1: 19217073-19217073 on Build GRCh38

View Full Product Details