Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 21812186-21812685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAD2
Entrez Gene ID
401944
Gene Name
low density lipoprotein receptor class A domain containing 2
Gene Aliases
-
UniGene
Hs.745158
Gene Chromosome Location
Chr.1: 21812209-21825221 on Build GRCh38
Transcripts
NM_001013693.2 XM_005245873.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5845554 c.53C>T p.A18V Chr.1: 21812504-21812504 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377661703 Chr.1: 21812578-21812578 on Build GRCh38
rs745710108 Chr.1: 21812519-21812519 on Build GRCh38
rs1014686176 Chr.1: 21812479-21812479 on Build GRCh38
rs775850284 Chr.1: 21812541-21812541 on Build GRCh38
rs750851320 Chr.1: 21812483-21812483 on Build GRCh38
rs765635357 Chr.1: 21812482-21812482 on Build GRCh38
rs747046790 Chr.1: 21812545-21812545 on Build GRCh38
rs935427000 Chr.1: 21812522-21812522 on Build GRCh38
rs34297508 Chr.1: 21812648-21812648 on Build GRCh38
rs371543117 Chr.1: 21812448-21812448 on Build GRCh38
rs945850016 Chr.1: 21812232-21812232 on Build GRCh38
rs563548932 Chr.1: 21812463-21812463 on Build GRCh38
rs552816278 Chr.1: 21812508-21812508 on Build GRCh38
rs569444922 Chr.1: 21812475-21812475 on Build GRCh38
rs544857294 Chr.1: 21812452-21812452 on Build GRCh38
rs777334981 Chr.1: 21812464-21812464 on Build GRCh38
rs1014489083 Chr.1: 21812383-21812383 on Build GRCh38
rs375740651 Chr.1: 21812449-21812449 on Build GRCh38
rs936010771 Chr.1: 21812362-21812362 on Build GRCh38
rs572836401 Chr.1: 21812562-21812562 on Build GRCh38
rs562017205 Chr.1: 21812561-21812561 on Build GRCh38
rs774982274 Chr.1: 21812584-21812585 on Build GRCh38
rs370685390 Chr.1: 21812388-21812388 on Build GRCh38
rs763554712 Chr.1: 21812586-21812586 on Build GRCh38
rs1006424106 Chr.1: 21812225-21812225 on Build GRCh38
rs547835372 Chr.1: 21812640-21812640 on Build GRCh38
rs1004763292 Chr.1: 21812460-21812462 on Build GRCh38
rs772154839 Chr.1: 21812523-21812523 on Build GRCh38
rs1030418294 Chr.1: 21812346-21812346 on Build GRCh38
rs760849649 Chr.1: 21812459-21812459 on Build GRCh38
rs772359611 Chr.1: 21812439-21812439 on Build GRCh38
rs559196119 Chr.1: 21812265-21812265 on Build GRCh38
rs755725252 Chr.1: 21812585-21812585 on Build GRCh38
rs967249989 Chr.1: 21812609-21812609 on Build GRCh38
rs767060404 Chr.1: 21812572-21812572 on Build GRCh38
rs767236018 Chr.1: 21812491-21812491 on Build GRCh38
rs953281779 Chr.1: 21812382-21812382 on Build GRCh38
rs963421689 Chr.1: 21812505-21812505 on Build GRCh38
rs752141916 Chr.1: 21812506-21812506 on Build GRCh38
rs762427414 Chr.1: 21812473-21812473 on Build GRCh38
rs894474112 Chr.1: 21812377-21812377 on Build GRCh38
rs1045711657 Chr.1: 21812607-21812607 on Build GRCh38
rs749156607 Chr.1: 21812509-21812509 on Build GRCh38
rs1014664403 Chr.1: 21812239-21812239 on Build GRCh38
rs1055681865 Chr.1: 21812374-21812374 on Build GRCh38
rs911772201 Chr.1: 21812601-21812601 on Build GRCh38
rs369050129 Chr.1: 21812588-21812588 on Build GRCh38
rs754540779 Chr.1: 21812486-21812486 on Build GRCh38
rs757216331 Chr.1: 21812510-21812510 on Build GRCh38
rs986450671 Chr.1: 21812434-21812434 on Build GRCh38
rs977623019 Chr.1: 21812611-21812611 on Build GRCh38
rs904576761 Chr.1: 21812353-21812353 on Build GRCh38
rs202151173 Chr.1: 21812518-21812518 on Build GRCh38
rs74060179 Chr.1: 21812270-21812270 on Build GRCh38
rs369934300 Chr.1: 21812458-21812458 on Build GRCh38
rs888054601 Chr.1: 21812637-21812637 on Build GRCh38
rs1036545312 Chr.1: 21812662-21812662 on Build GRCh38
rs759713366 Chr.1: 21812418-21812418 on Build GRCh38
rs767234683 Chr.1: 21812584-21812584 on Build GRCh38
rs374372684 Chr.1: 21812556-21812556 on Build GRCh38
rs887937087 Chr.1: 21812216-21812216 on Build GRCh38

View Full Product Details