Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23321462-23322009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPR
Entrez Gene ID
10236
Gene Name
heterogeneous nuclear ribonucleoprotein R
Gene Aliases
HNRPR, hnRNP-R
UniGene
Hs.373763
Gene Chromosome Location
Chr.1: 23304688-23344650 on Build GRCh38
Transcripts
NM_001102397.2 NM_001102398.2 NM_001102399.2 NM_001297620.1 NM_001297621.1 NM_001297622.1 NM_005826.4 XM_005245711.4 XM_011540471.2 XM_011540472.2 XM_011540474.2 XM_011540475.2 XM_011540476.2 XM_011540477.2 XM_017000008.1 XM_017000009.1 XM_017000010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1473640 c.676-2A>G p.? Chr.1: 23321665-23321665 on Build GRCh38
6301816 c.798C>T p.F266F Chr.1: 23321541-23321541 on Build GRCh38
3485604 c.712G>A p.G238R Chr.1: 23321627-23321627 on Build GRCh38
6125255 c.748G>C p.V250L Chr.1: 23321591-23321591 on Build GRCh38
1687198 c.755C>T p.S252F Chr.1: 23321584-23321584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200798900 Chr.1: 23321534-23321534 on Build GRCh38
rs764215177 Chr.1: 23321644-23321644 on Build GRCh38
rs754560878 Chr.1: 23321525-23321525 on Build GRCh38
rs530696343 Chr.1: 23321747-23321747 on Build GRCh38
rs1004434548 Chr.1: 23321778-23321778 on Build GRCh38
rs967705375 Chr.1: 23321919-23321919 on Build GRCh38
rs375010625 Chr.1: 23321652-23321652 on Build GRCh38
rs1018754305 Chr.1: 23321908-23321908 on Build GRCh38
rs746692388 Chr.1: 23321494-23321494 on Build GRCh38
rs774124964 Chr.1: 23321516-23321516 on Build GRCh38
rs747651413 Chr.1: 23321541-23321541 on Build GRCh38
rs1008756713 Chr.1: 23321901-23321901 on Build GRCh38
rs371701269 Chr.1: 23321583-23321583 on Build GRCh38
rs376874398 Chr.1: 23321488-23321488 on Build GRCh38
rs11803953 Chr.1: 23321852-23321852 on Build GRCh38
rs528396894 Chr.1: 23321825-23321825 on Build GRCh38
rs545465612 Chr.1: 23321613-23321613 on Build GRCh38
rs770792006 Chr.1: 23321602-23321602 on Build GRCh38
rs553134553 Chr.1: 23321839-23321840 on Build GRCh38
rs775783663 Chr.1: 23321618-23321618 on Build GRCh38
rs965997983 Chr.1: 23321985-23321985 on Build GRCh38
rs953218271 Chr.1: 23321890-23321890 on Build GRCh38
rs370805246 Chr.1: 23321493-23321493 on Build GRCh38
rs568636558 Chr.1: 23321964-23321964 on Build GRCh38
rs894807792 Chr.1: 23321811-23321811 on Build GRCh38
rs376283755 Chr.1: 23321887-23321887 on Build GRCh38
rs754725819 Chr.1: 23321707-23321707 on Build GRCh38
rs141121420 Chr.1: 23321577-23321577 on Build GRCh38
rs745306777 Chr.1: 23321873-23321873 on Build GRCh38
rs546626282 Chr.1: 23321826-23321826 on Build GRCh38
rs936404661 Chr.1: 23321774-23321774 on Build GRCh38
rs773968974 Chr.1: 23321607-23321607 on Build GRCh38
rs550629408 Chr.1: 23321959-23321959 on Build GRCh38
rs12038885 Chr.1: 23321507-23321507 on Build GRCh38
rs757652779 Chr.1: 23321648-23321648 on Build GRCh38
rs780808223 Chr.1: 23321710-23321710 on Build GRCh38
rs759851126 Chr.1: 23321612-23321612 on Build GRCh38
rs977323982 Chr.1: 23321926-23321926 on Build GRCh38
rs944320258 Chr.1: 23321742-23321742 on Build GRCh38
rs77029355 Chr.1: 23321579-23321579 on Build GRCh38
rs79255301 Chr.1: 23321716-23321716 on Build GRCh38
rs1046069156 Chr.1: 23321822-23321825 on Build GRCh38
rs887375651 Chr.1: 23321761-23321761 on Build GRCh38
rs770602334 Chr.1: 23321490-23321490 on Build GRCh38
rs760826644 Chr.1: 23321642-23321642 on Build GRCh38
rs1033464922 Chr.1: 23321936-23321936 on Build GRCh38
rs924160339 Chr.1: 23321751-23321751 on Build GRCh38
rs572960203 Chr.1: 23321849-23321849 on Build GRCh38
rs185458376 Chr.1: 23321808-23321808 on Build GRCh38
rs572391527 Chr.1: 23321839-23321843 on Build GRCh38
rs912767632 Chr.1: 23321742-23321743 on Build GRCh38
rs906421399 Chr.1: 23321823-23321824 on Build GRCh38
rs200114895 Chr.1: 23321547-23321547 on Build GRCh38
rs370494002 Chr.1: 23321824-23321824 on Build GRCh38
rs759133817 Chr.1: 23321546-23321546 on Build GRCh38
rs190161975 Chr.1: 23321820-23321820 on Build GRCh38
rs754424759 Chr.1: 23321645-23321645 on Build GRCh38
rs1019624801 Chr.1: 23321946-23321946 on Build GRCh38
rs769779258 Chr.1: 23321568-23321568 on Build GRCh38
rs750717980 Chr.1: 23321686-23321686 on Build GRCh38
rs760453452 Chr.1: 23321756-23321756 on Build GRCh38
rs527763565 Chr.1: 23321839-23321842 on Build GRCh38

View Full Product Details