Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74830052-74830563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPLX3
Entrez Gene ID
594855
Gene Name
complexin 3
Gene Aliases
CPX-III, CPXIII, Nbla11589
UniGene
Hs.187694
Gene Chromosome Location
Chr.15: 74826610-74831795 on Build GRCh38
Transcripts
NM_001030005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5594443 c.334G>A p.D112N Chr.15: 74830211-74830211 on Build GRCh38
1374573 c.294C>T p.D98D Chr.15: 74830171-74830171 on Build GRCh38
5896238 c.415G>A p.D139N Chr.15: 74830292-74830292 on Build GRCh38
1493509 c.276C>A p.I92I Chr.15: 74830153-74830153 on Build GRCh38
5383834 c.414G>A p.K138K Chr.15: 74830291-74830291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745700690 Chr.15: 74830215-74830215 on Build GRCh38
rs932236103 Chr.15: 74830523-74830523 on Build GRCh38
rs141104376 Chr.15: 74830466-74830466 on Build GRCh38
rs779982482 Chr.15: 74830345-74830345 on Build GRCh38
rs1049433493 Chr.15: 74830362-74830362 on Build GRCh38
rs751061419 Chr.15: 74830310-74830310 on Build GRCh38
rs896973887 Chr.15: 74830412-74830412 on Build GRCh38
rs756612706 Chr.15: 74830124-74830124 on Build GRCh38
rs1011274019 Chr.15: 74830102-74830102 on Build GRCh38
rs567043629 Chr.15: 74830435-74830435 on Build GRCh38
rs755073779 Chr.15: 74830367-74830367 on Build GRCh38
rs749978130 Chr.15: 74830333-74830333 on Build GRCh38
rs551795671 Chr.15: 74830419-74830419 on Build GRCh38
rs550173144 Chr.15: 74830533-74830533 on Build GRCh38
rs774540746 Chr.15: 74830261-74830261 on Build GRCh38
rs745837076 Chr.15: 74830153-74830153 on Build GRCh38
rs772406720 Chr.15: 74830217-74830219 on Build GRCh38
rs751854584 Chr.15: 74830323-74830323 on Build GRCh38
rs947000094 Chr.15: 74830361-74830361 on Build GRCh38
rs936181688 Chr.15: 74830144-74830144 on Build GRCh38
rs761172588 Chr.15: 74830349-74830350 on Build GRCh38
rs144213956 Chr.15: 74830185-74830185 on Build GRCh38
rs753894200 Chr.15: 74830346-74830346 on Build GRCh38
rs760087747 Chr.15: 74830380-74830380 on Build GRCh38
rs766880854 Chr.15: 74830392-74830392 on Build GRCh38
rs775522144 Chr.15: 74830169-74830169 on Build GRCh38
rs147366687 Chr.15: 74830288-74830288 on Build GRCh38
rs781040541 Chr.15: 74830150-74830150 on Build GRCh38
rs749179855 Chr.15: 74830234-74830234 on Build GRCh38
rs746083924 Chr.15: 74830216-74830218 on Build GRCh38
rs147232026 Chr.15: 74830181-74830181 on Build GRCh38
rs756133730 Chr.15: 74830538-74830538 on Build GRCh38
rs374967692 Chr.15: 74830364-74830364 on Build GRCh38
rs763638070 Chr.15: 74830308-74830308 on Build GRCh38
rs756906140 Chr.15: 74830207-74830207 on Build GRCh38
rs369575878 Chr.15: 74830126-74830126 on Build GRCh38
rs754273793 Chr.15: 74830195-74830195 on Build GRCh38
rs780901157 Chr.15: 74830210-74830210 on Build GRCh38
rs889512938 Chr.15: 74830366-74830366 on Build GRCh38
rs997320576 Chr.15: 74830528-74830528 on Build GRCh38
rs761392286 Chr.15: 74830389-74830389 on Build GRCh38
rs774431843 Chr.15: 74830117-74830118 on Build GRCh38
rs767054980 Chr.15: 74830111-74830111 on Build GRCh38
rs867613978 Chr.15: 74830423-74830423 on Build GRCh38
rs982198182 Chr.15: 74830451-74830451 on Build GRCh38
rs375784018 Chr.15: 74830100-74830100 on Build GRCh38
rs757054611 Chr.15: 74830149-74830149 on Build GRCh38
rs924826574 Chr.15: 74830406-74830406 on Build GRCh38
rs778358268 Chr.15: 74830520-74830520 on Build GRCh38
rs982214885 Chr.15: 74830235-74830235 on Build GRCh38
rs760401104 Chr.15: 74830395-74830395 on Build GRCh38
rs747181678 Chr.15: 74830369-74830369 on Build GRCh38
rs750959297 Chr.15: 74830108-74830108 on Build GRCh38
rs538360936 Chr.15: 74830521-74830521 on Build GRCh38
rs557888970 Chr.15: 74830133-74830133 on Build GRCh38
rs199540758 Chr.15: 74830303-74830303 on Build GRCh38
rs750148517 Chr.15: 74830393-74830393 on Build GRCh38
rs369385409 Chr.15: 74830105-74830105 on Build GRCh38
rs771019880 Chr.15: 74830106-74830113 on Build GRCh38
rs140289965 Chr.15: 74830184-74830184 on Build GRCh38
rs970761151 Chr.15: 74830449-74830449 on Build GRCh38
rs367757715 Chr.15: 74830204-74830204 on Build GRCh38
rs755827310 Chr.15: 74830216-74830216 on Build GRCh38
rs371177006 Chr.15: 74830116-74830116 on Build GRCh38
rs780146194 Chr.15: 74830228-74830228 on Build GRCh38
rs377287963 Chr.15: 74830109-74830109 on Build GRCh38
rs3743487 Chr.15: 74830515-74830515 on Build GRCh38
rs1017849101 Chr.15: 74830516-74830516 on Build GRCh38
rs138053302 Chr.15: 74830137-74830137 on Build GRCh38
rs1026900616 Chr.15: 74830428-74830428 on Build GRCh38
rs751283611 Chr.15: 74830205-74830205 on Build GRCh38
rs1045510272 Chr.15: 74830429-74830429 on Build GRCh38
rs118068328 Chr.15: 74830178-74830178 on Build GRCh38
rs772359742 Chr.15: 74830286-74830286 on Build GRCh38
rs372883520 Chr.15: 74830131-74830131 on Build GRCh38
rs775564497 Chr.15: 74830312-74830312 on Build GRCh38
rs993357721 Chr.15: 74830208-74830208 on Build GRCh38
rs372733021 Chr.15: 74830298-74830298 on Build GRCh38
rs775822421 Chr.15: 74830388-74830388 on Build GRCh38
rs771347048 Chr.15: 74830371-74830371 on Build GRCh38
rs1023603316 Chr.15: 74830440-74830440 on Build GRCh38
rs952633307 Chr.15: 74830433-74830433 on Build GRCh38
rs368430383 Chr.15: 74830365-74830365 on Build GRCh38
rs768915432 Chr.15: 74830247-74830247 on Build GRCh38
rs748367134 Chr.15: 74830276-74830276 on Build GRCh38
rs532691124 Chr.15: 74830501-74830501 on Build GRCh38
rs143709534 Chr.15: 74830171-74830171 on Build GRCh38
rs147966681 Chr.15: 74830132-74830132 on Build GRCh38
rs748035827 Chr.15: 74830105-74830106 on Build GRCh38
rs533626135 Chr.15: 74830376-74830376 on Build GRCh38
rs769751571 Chr.15: 74830165-74830165 on Build GRCh38
rs770310639 Chr.15: 74830387-74830387 on Build GRCh38
rs562166313 Chr.15: 74830368-74830368 on Build GRCh38
rs887659217 Chr.15: 74830500-74830500 on Build GRCh38
rs767865910 Chr.15: 74830106-74830106 on Build GRCh38
rs199684945 Chr.15: 74830172-74830172 on Build GRCh38
rs1024355808 Chr.15: 74830190-74830190 on Build GRCh38
rs755952079 Chr.15: 74830339-74830339 on Build GRCh38

View Full Product Details