Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 16263270-16263775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOMO3
Entrez Gene ID
408050
Gene Name
NODAL modulator 3
Gene Aliases
Nomo
UniGene
Hs.460141
Gene Chromosome Location
Chr.16: 16232532-16294811 on Build GRCh38
Transcripts
NM_001004067.3 XM_005255318.1

SNP Detail

SNP ID Chromosome Location
rs754337163 Chr.16: 16263599-16263599 on Build GRCh38
rs1046979817 Chr.16: 16263601-16263601 on Build GRCh38
rs180828331 Chr.16: 16263380-16263380 on Build GRCh38
rs4992353 Chr.16: 16263363-16263363 on Build GRCh38
rs984343963 Chr.16: 16263496-16263496 on Build GRCh38
rs368579985 Chr.16: 16263555-16263555 on Build GRCh38
rs375238777 Chr.16: 16263529-16263529 on Build GRCh38
rs894156208 Chr.16: 16263684-16263684 on Build GRCh38
rs1011218798 Chr.16: 16263750-16263750 on Build GRCh38
rs796070515 Chr.16: 16263359-16263359 on Build GRCh38
rs552344411 Chr.16: 16263425-16263425 on Build GRCh38
rs755821330 Chr.16: 16263384-16263384 on Build GRCh38
rs568931338 Chr.16: 16263664-16263664 on Build GRCh38
rs766274746 Chr.16: 16263544-16263544 on Build GRCh38
rs965832241 Chr.16: 16263398-16263398 on Build GRCh38
rs931273761 Chr.16: 16263483-16263483 on Build GRCh38
rs550636357 Chr.16: 16263654-16263654 on Build GRCh38
rs912671951 Chr.16: 16263361-16263361 on Build GRCh38
rs1021511212 Chr.16: 16263313-16263313 on Build GRCh38
rs797000108 Chr.16: 16263367-16263367 on Build GRCh38
rs540558617 Chr.16: 16263293-16263293 on Build GRCh38
rs796955671 Chr.16: 16263352-16263352 on Build GRCh38
rs564152937 Chr.16: 16263453-16263453 on Build GRCh38
rs546940057 Chr.16: 16263613-16263613 on Build GRCh38
rs778309193 Chr.16: 16263598-16263598 on Build GRCh38
rs757889417 Chr.16: 16263599-16263599 on Build GRCh38
rs756626085 Chr.16: 16263587-16263587 on Build GRCh38
rs533302242 Chr.16: 16263404-16263404 on Build GRCh38
rs752185268 Chr.16: 16263569-16263569 on Build GRCh38
rs751212849 Chr.16: 16263554-16263554 on Build GRCh38
rs185232185 Chr.16: 16263391-16263391 on Build GRCh38
rs568016285 Chr.16: 16263619-16263619 on Build GRCh38
rs190997567 Chr.16: 16263326-16263326 on Build GRCh38
rs200067201 Chr.16: 16263645-16263645 on Build GRCh38
rs769791856 Chr.16: 16263475-16263475 on Build GRCh38
rs779310945 Chr.16: 16263636-16263636 on Build GRCh38
rs528355662 Chr.16: 16263542-16263542 on Build GRCh38

View Full Product Details