Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71284108-71284628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR2
Entrez Gene ID
55783
Gene Name
cap methyltransferase 2
Gene Aliases
AFT, FTSJD1, HMTr2, MTr2
UniGene
Hs.72782
Gene Chromosome Location
Chr.16: 71282300-71290178 on Build GRCh38
Transcripts
NM_001099642.1 NM_001324374.1 NM_001324377.1 NM_001324378.1 NM_001324379.1 NM_018348.5 XM_011523228.1 XM_011523229.1 XM_017023444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667664 c.1765C>T p.R589C Chr.16: 71284156-71284156 on Build GRCh38
1600373 c.1330_1331insA p.Y444fs*1 Chr.16: 71284590-71284591 on Build GRCh38
4466305 c.1427C>T p.P476L Chr.16: 71284494-71284494 on Build GRCh38
973259 c.1659A>G p.E553E Chr.16: 71284262-71284262 on Build GRCh38
5747757 c.1777_1778insA p.M593fs*14 Chr.16: 71284143-71284144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138858702 Chr.16: 71284413-71284413 on Build GRCh38
rs769373314 Chr.16: 71284379-71284379 on Build GRCh38
rs760668163 Chr.16: 71284506-71284506 on Build GRCh38
rs774794348 Chr.16: 71284504-71284505 on Build GRCh38
rs771388385 Chr.16: 71284361-71284361 on Build GRCh38
rs752766298 Chr.16: 71284427-71284427 on Build GRCh38
rs757067089 Chr.16: 71284275-71284275 on Build GRCh38
rs146301240 Chr.16: 71284537-71284537 on Build GRCh38
rs757252813 Chr.16: 71284227-71284227 on Build GRCh38
rs774739666 Chr.16: 71284196-71284196 on Build GRCh38
rs768395076 Chr.16: 71284389-71284389 on Build GRCh38
rs774067087 Chr.16: 71284397-71284397 on Build GRCh38
rs763851216 Chr.16: 71284155-71284155 on Build GRCh38
rs752854395 Chr.16: 71284350-71284350 on Build GRCh38
rs764950988 Chr.16: 71284242-71284242 on Build GRCh38
rs149004042 Chr.16: 71284163-71284163 on Build GRCh38
rs749668675 Chr.16: 71284132-71284132 on Build GRCh38
rs369132059 Chr.16: 71284309-71284309 on Build GRCh38
rs373298047 Chr.16: 71284408-71284408 on Build GRCh38
rs373425212 Chr.16: 71284364-71284364 on Build GRCh38
rs940157417 Chr.16: 71284202-71284202 on Build GRCh38
rs866016844 Chr.16: 71284523-71284523 on Build GRCh38
rs766084517 Chr.16: 71284149-71284149 on Build GRCh38
rs189091912 Chr.16: 71284134-71284134 on Build GRCh38
rs543435476 Chr.16: 71284133-71284133 on Build GRCh38
rs748256920 Chr.16: 71284165-71284165 on Build GRCh38
rs753889553 Chr.16: 71284338-71284338 on Build GRCh38
rs770286365 Chr.16: 71284229-71284229 on Build GRCh38
rs780337305 Chr.16: 71284181-71284181 on Build GRCh38
rs768922763 Chr.16: 71284192-71284192 on Build GRCh38
rs749535167 Chr.16: 71284187-71284187 on Build GRCh38
rs993816314 Chr.16: 71284529-71284529 on Build GRCh38
rs141956899 Chr.16: 71284332-71284332 on Build GRCh38
rs373666425 Chr.16: 71284573-71284573 on Build GRCh38
rs759979920 Chr.16: 71284578-71284582 on Build GRCh38
rs1021905565 Chr.16: 71284367-71284367 on Build GRCh38
rs767994557 Chr.16: 71284574-71284574 on Build GRCh38
rs139490864 Chr.16: 71284250-71284250 on Build GRCh38
rs372970522 Chr.16: 71284575-71284575 on Build GRCh38
rs771464317 Chr.16: 71284288-71284288 on Build GRCh38
rs747896676 Chr.16: 71284500-71284500 on Build GRCh38
rs748493382 Chr.16: 71284201-71284201 on Build GRCh38
rs747367339 Chr.16: 71284359-71284359 on Build GRCh38
rs577001969 Chr.16: 71284590-71284590 on Build GRCh38
rs767470714 Chr.16: 71284325-71284325 on Build GRCh38
rs760052126 Chr.16: 71284146-71284146 on Build GRCh38
rs781564563 Chr.16: 71284460-71284460 on Build GRCh38
rs757600163 Chr.16: 71284451-71284451 on Build GRCh38
rs868756180 Chr.16: 71284159-71284159 on Build GRCh38
rs766271722 Chr.16: 71284513-71284513 on Build GRCh38
rs116415286 Chr.16: 71284299-71284299 on Build GRCh38
rs974124295 Chr.16: 71284586-71284586 on Build GRCh38
rs776044913 Chr.16: 71284292-71284292 on Build GRCh38
rs375814094 Chr.16: 71284580-71284580 on Build GRCh38
rs376674471 Chr.16: 71284237-71284237 on Build GRCh38
rs778262150 Chr.16: 71284287-71284287 on Build GRCh38
rs771408358 Chr.16: 71284333-71284335 on Build GRCh38
rs533359535 Chr.16: 71284281-71284281 on Build GRCh38
rs753360412 Chr.16: 71284150-71284150 on Build GRCh38
rs548970341 Chr.16: 71284411-71284411 on Build GRCh38
rs144187091 Chr.16: 71284354-71284354 on Build GRCh38
rs760233068 Chr.16: 71284222-71284222 on Build GRCh38
rs745375782 Chr.16: 71284376-71284376 on Build GRCh38
rs370043476 Chr.16: 71284557-71284557 on Build GRCh38
rs745977271 Chr.16: 71284165-71284165 on Build GRCh38
rs755244383 Chr.16: 71284279-71284279 on Build GRCh38
rs772354347 Chr.16: 71284284-71284284 on Build GRCh38
rs772760499 Chr.16: 71284143-71284143 on Build GRCh38
rs779110256 Chr.16: 71284341-71284341 on Build GRCh38
rs148127346 Chr.16: 71284314-71284314 on Build GRCh38
rs898159773 Chr.16: 71284522-71284522 on Build GRCh38
rs754811555 Chr.16: 71284154-71284154 on Build GRCh38
rs756685693 Chr.16: 71284475-71284475 on Build GRCh38
rs775596139 Chr.16: 71284531-71284531 on Build GRCh38
rs200731984 Chr.16: 71284249-71284249 on Build GRCh38
rs201117712 Chr.16: 71284504-71284504 on Build GRCh38
rs749831737 Chr.16: 71284193-71284193 on Build GRCh38
rs376479919 Chr.16: 71284447-71284447 on Build GRCh38
rs1049980698 Chr.16: 71284276-71284276 on Build GRCh38
rs748788837 Chr.16: 71284486-71284486 on Build GRCh38
rs756082420 Chr.16: 71284169-71284169 on Build GRCh38
rs772586675 Chr.16: 71284204-71284204 on Build GRCh38
rs146282486 Chr.16: 71284156-71284156 on Build GRCh38
rs764354938 Chr.16: 71284429-71284429 on Build GRCh38
rs149735283 Chr.16: 71284605-71284605 on Build GRCh38
rs146080989 Chr.16: 71284386-71284386 on Build GRCh38
rs757206938 Chr.16: 71284270-71284270 on Build GRCh38
rs775217324 Chr.16: 71284300-71284300 on Build GRCh38
rs764039394 Chr.16: 71284313-71284313 on Build GRCh38
rs780661359 Chr.16: 71284581-71284581 on Build GRCh38
rs773667247 Chr.16: 71284210-71284210 on Build GRCh38
rs768217999 Chr.16: 71284492-71284492 on Build GRCh38
rs139275656 Chr.16: 71284589-71284589 on Build GRCh38
rs142068931 Chr.16: 71284468-71284468 on Build GRCh38
rs754448049 Chr.16: 71284604-71284604 on Build GRCh38
rs759623445 Chr.16: 71284416-71284416 on Build GRCh38
rs755007276 Chr.16: 71284340-71284340 on Build GRCh38
rs779061347 Chr.16: 71284280-71284280 on Build GRCh38
rs759062095 Chr.16: 71284296-71284296 on Build GRCh38
rs769970271 Chr.16: 71284185-71284188 on Build GRCh38
rs369141257 Chr.16: 71284352-71284352 on Build GRCh38
rs774017517 Chr.16: 71284495-71284495 on Build GRCh38
rs779751456 Chr.16: 71284480-71284480 on Build GRCh38
rs758658803 Chr.16: 71284428-71284428 on Build GRCh38
rs762377510 Chr.16: 71284137-71284137 on Build GRCh38
rs559476134 Chr.16: 71284265-71284265 on Build GRCh38
rs369814717 Chr.16: 71284383-71284383 on Build GRCh38
rs753927764 Chr.16: 71284277-71284277 on Build GRCh38
rs764133703 Chr.16: 71284262-71284262 on Build GRCh38
rs1038360049 Chr.16: 71284216-71284216 on Build GRCh38
rs771840302 Chr.16: 71284502-71284502 on Build GRCh38
rs777823715 Chr.16: 71284186-71284187 on Build GRCh38
rs757343059 Chr.16: 71284422-71284422 on Build GRCh38
rs761701086 Chr.16: 71284317-71284317 on Build GRCh38

View Full Product Details