Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34626014-34626508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132E
Entrez Gene ID
124842
Gene Name
transmembrane protein 132E
Gene Aliases
DFNB99
UniGene
Hs.310482
Gene Chromosome Location
Chr.17: 34580749-34639318 on Build GRCh38
Transcripts
NM_001304438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
472574 c.413C>T p.S138L Chr.17: 34626472-34626472 on Build GRCh38
3755392 c.180G>A p.E60E Chr.17: 34626239-34626239 on Build GRCh38
3691482 c.199G>C p.A67P Chr.17: 34626258-34626258 on Build GRCh38
5032544 c.299G>A p.S100N Chr.17: 34626358-34626358 on Build GRCh38
1382363 c.409G>A p.A137T Chr.17: 34626468-34626468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745393011 Chr.17: 34626113-34626113 on Build GRCh38
rs754603575 Chr.17: 34626289-34626289 on Build GRCh38
rs570070256 Chr.17: 34626281-34626281 on Build GRCh38
rs868256899 Chr.17: 34626154-34626154 on Build GRCh38
rs772541435 Chr.17: 34626231-34626231 on Build GRCh38
rs763854856 Chr.17: 34626465-34626465 on Build GRCh38
rs918510902 Chr.17: 34626134-34626134 on Build GRCh38
rs767556248 Chr.17: 34626259-34626259 on Build GRCh38
rs1032479778 Chr.17: 34626174-34626174 on Build GRCh38
rs747420328 Chr.17: 34626411-34626411 on Build GRCh38
rs754068568 Chr.17: 34626162-34626162 on Build GRCh38
rs555049321 Chr.17: 34626181-34626181 on Build GRCh38
rs200982924 Chr.17: 34626297-34626297 on Build GRCh38
rs141502241 Chr.17: 34626415-34626415 on Build GRCh38
rs990546172 Chr.17: 34626057-34626057 on Build GRCh38
rs776774685 Chr.17: 34626240-34626240 on Build GRCh38
rs539445182 Chr.17: 34626250-34626250 on Build GRCh38
rs774649666 Chr.17: 34626217-34626217 on Build GRCh38
rs752214418 Chr.17: 34626146-34626172 on Build GRCh38
rs748827129 Chr.17: 34626207-34626207 on Build GRCh38
rs780806388 Chr.17: 34626110-34626110 on Build GRCh38
rs751836818 Chr.17: 34626467-34626467 on Build GRCh38
rs770107846 Chr.17: 34626327-34626327 on Build GRCh38
rs763720630 Chr.17: 34626394-34626395 on Build GRCh38
rs755611078 Chr.17: 34626363-34626363 on Build GRCh38
rs760483390 Chr.17: 34626373-34626373 on Build GRCh38
rs372634022 Chr.17: 34626169-34626169 on Build GRCh38
rs775918902 Chr.17: 34626332-34626332 on Build GRCh38
rs918380807 Chr.17: 34626043-34626043 on Build GRCh38
rs371248579 Chr.17: 34626320-34626320 on Build GRCh38
rs867759275 Chr.17: 34626145-34626145 on Build GRCh38
rs924907023 Chr.17: 34626051-34626051 on Build GRCh38
rs369122850 Chr.17: 34626397-34626397 on Build GRCh38
rs1011967633 Chr.17: 34626346-34626346 on Build GRCh38
rs147874855 Chr.17: 34626316-34626316 on Build GRCh38
rs753259918 Chr.17: 34626389-34626389 on Build GRCh38
rs754552346 Chr.17: 34626395-34626395 on Build GRCh38
rs762229395 Chr.17: 34626246-34626246 on Build GRCh38
rs1048932398 Chr.17: 34626064-34626064 on Build GRCh38
rs549722356 Chr.17: 34626079-34626079 on Build GRCh38
rs766622532 Chr.17: 34626161-34626161 on Build GRCh38
rs779393561 Chr.17: 34626274-34626274 on Build GRCh38
rs764581147 Chr.17: 34626163-34626163 on Build GRCh38
rs772862890 Chr.17: 34626439-34626439 on Build GRCh38
rs894687958 Chr.17: 34626337-34626337 on Build GRCh38
rs1003222082 Chr.17: 34626380-34626380 on Build GRCh38
rs998795450 Chr.17: 34626449-34626449 on Build GRCh38
rs767355812 Chr.17: 34626339-34626339 on Build GRCh38
rs553628300 Chr.17: 34626201-34626201 on Build GRCh38
rs998345077 Chr.17: 34626199-34626199 on Build GRCh38
rs758047867 Chr.17: 34626410-34626410 on Build GRCh38
rs745351904 Chr.17: 34626188-34626188 on Build GRCh38
rs929827394 Chr.17: 34626054-34626054 on Build GRCh38
rs533992168 Chr.17: 34626160-34626160 on Build GRCh38
rs984483614 Chr.17: 34626447-34626447 on Build GRCh38
rs373712983 Chr.17: 34626365-34626365 on Build GRCh38
rs747404686 Chr.17: 34626318-34626318 on Build GRCh38
rs375386140 Chr.17: 34626170-34626170 on Build GRCh38
rs531471200 Chr.17: 34626109-34626109 on Build GRCh38
rs768034783 Chr.17: 34626146-34626146 on Build GRCh38
rs771316857 Chr.17: 34626414-34626414 on Build GRCh38
rs766389805 Chr.17: 34626376-34626376 on Build GRCh38
rs756083769 Chr.17: 34626272-34626272 on Build GRCh38
rs942784325 Chr.17: 34626271-34626271 on Build GRCh38
rs760560071 Chr.17: 34626440-34626440 on Build GRCh38
rs748511938 Chr.17: 34626306-34626306 on Build GRCh38
rs760798406 Chr.17: 34626237-34626237 on Build GRCh38
rs867741927 Chr.17: 34626248-34626248 on Build GRCh38
rs761740460 Chr.17: 34626257-34626257 on Build GRCh38
rs760107973 Chr.17: 34626262-34626263 on Build GRCh38
rs1046769345 Chr.17: 34626127-34626127 on Build GRCh38
rs768387006 Chr.17: 34626209-34626209 on Build GRCh38
rs564430970 Chr.17: 34626458-34626458 on Build GRCh38
rs775043144 Chr.17: 34626128-34626128 on Build GRCh38
rs144668878 Chr.17: 34626315-34626315 on Build GRCh38
rs761687275 Chr.17: 34626338-34626338 on Build GRCh38
rs757261559 Chr.17: 34626098-34626098 on Build GRCh38
rs750264831 Chr.17: 34626344-34626344 on Build GRCh38
rs1054611173 Chr.17: 34626324-34626324 on Build GRCh38
rs1023296725 Chr.17: 34626214-34626214 on Build GRCh38
rs4795954 Chr.17: 34626239-34626239 on Build GRCh38
rs971008822 Chr.17: 34626310-34626310 on Build GRCh38
rs868028004 Chr.17: 34626266-34626266 on Build GRCh38
rs572596534 Chr.17: 34626404-34626404 on Build GRCh38
rs541603154 Chr.17: 34626422-34626422 on Build GRCh38
rs749482561 Chr.17: 34626432-34626432 on Build GRCh38
rs781327570 Chr.17: 34626182-34626182 on Build GRCh38
rs983957108 Chr.17: 34626260-34626260 on Build GRCh38
rs749658557 Chr.17: 34626136-34626136 on Build GRCh38
rs768945189 Chr.17: 34626434-34626434 on Build GRCh38
rs757706114 Chr.17: 34626479-34626479 on Build GRCh38
rs764664450 Chr.17: 34626455-34626455 on Build GRCh38
rs755645329 Chr.17: 34626200-34626200 on Build GRCh38
rs774704736 Chr.17: 34626143-34626143 on Build GRCh38
rs949235660 Chr.17: 34626107-34626107 on Build GRCh38
rs776520313 Chr.17: 34626322-34626322 on Build GRCh38
rs776690930 Chr.17: 34626444-34626444 on Build GRCh38
rs769464532 Chr.17: 34626115-34626115 on Build GRCh38
rs773257960 Chr.17: 34626159-34626159 on Build GRCh38
rs766225189 Chr.17: 34626441-34626441 on Build GRCh38
rs568193524 Chr.17: 34626142-34626142 on Build GRCh38
rs759706514 Chr.17: 34626323-34626323 on Build GRCh38
rs547995253 Chr.17: 34626120-34626120 on Build GRCh38
rs150026307 Chr.17: 34626263-34626263 on Build GRCh38
rs372929322 Chr.17: 34626454-34626454 on Build GRCh38
rs909710873 Chr.17: 34626448-34626448 on Build GRCh38
rs773777668 Chr.17: 34626233-34626233 on Build GRCh38
rs144304954 Chr.17: 34626287-34626287 on Build GRCh38
rs757992923 Chr.17: 34626461-34626461 on Build GRCh38
rs199589445 Chr.17: 34626082-34626082 on Build GRCh38

View Full Product Details