Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74847760-74848256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2C
Entrez Gene ID
2905
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Gene Aliases
GluN2C, NMDAR2C, NR2C
UniGene
Hs.436980
Gene Chromosome Location
Chr.17: 74842021-74860843 on Build GRCh38
Transcripts
NM_000835.4 NM_001278553.1 NR_103735.1 XM_006721845.3 XM_006721846.3 XM_011524686.2 XM_011524687.2 XM_011524688.2 XM_011524689.2 XM_011524692.2 XM_017024544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5614505 c.1757T>A p.L586H Chr.17: 74847866-74847866 on Build GRCh38
4569382 c.1714T>C p.F572L Chr.17: 74847909-74847909 on Build GRCh38
1610738 c.1695T>G p.T565T Chr.17: 74847928-74847928 on Build GRCh38
5619539 c.1711G>T p.V571F Chr.17: 74847912-74847912 on Build GRCh38
984130 c.1722C>T p.F574F Chr.17: 74847901-74847901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549726395 Chr.17: 74848216-74848216 on Build GRCh38
rs182147976 Chr.17: 74848110-74848110 on Build GRCh38
rs752276567 Chr.17: 74848001-74848001 on Build GRCh38
rs748229825 Chr.17: 74847901-74847901 on Build GRCh38
rs748205127 Chr.17: 74847826-74847826 on Build GRCh38
rs551649225 Chr.17: 74848064-74848064 on Build GRCh38
rs752166394 Chr.17: 74847845-74847845 on Build GRCh38
rs751447098 Chr.17: 74848004-74848004 on Build GRCh38
rs772040978 Chr.17: 74847820-74847820 on Build GRCh38
rs1014581446 Chr.17: 74848143-74848143 on Build GRCh38
rs749558742 Chr.17: 74847912-74847912 on Build GRCh38
rs890219760 Chr.17: 74847785-74847785 on Build GRCh38
rs767237427 Chr.17: 74847952-74847952 on Build GRCh38
rs940345257 Chr.17: 74847834-74847834 on Build GRCh38
rs906447390 Chr.17: 74847942-74847942 on Build GRCh38
rs759997177 Chr.17: 74847837-74847837 on Build GRCh38
rs202213203 Chr.17: 74847862-74847862 on Build GRCh38
rs922184444 Chr.17: 74848116-74848116 on Build GRCh38
rs938663215 Chr.17: 74847963-74847963 on Build GRCh38
rs547591082 Chr.17: 74848147-74848147 on Build GRCh38
rs3744192 Chr.17: 74847784-74847784 on Build GRCh38
rs567723844 Chr.17: 74848159-74848159 on Build GRCh38
rs961746135 Chr.17: 74848162-74848162 on Build GRCh38
rs570741986 Chr.17: 74848114-74848114 on Build GRCh38
rs3744193 Chr.17: 74848026-74848026 on Build GRCh38
rs776102975 Chr.17: 74847931-74847931 on Build GRCh38
rs932157853 Chr.17: 74847783-74847783 on Build GRCh38
rs761626049 Chr.17: 74847965-74847965 on Build GRCh38
rs115547593 Chr.17: 74848059-74848059 on Build GRCh38
rs369978472 Chr.17: 74847989-74847989 on Build GRCh38
rs779982912 Chr.17: 74847900-74847900 on Build GRCh38
rs536255874 Chr.17: 74848189-74848189 on Build GRCh38
rs987136246 Chr.17: 74847792-74847792 on Build GRCh38
rs373944929 Chr.17: 74847926-74847926 on Build GRCh38
rs745672170 Chr.17: 74847879-74847879 on Build GRCh38
rs754160979 Chr.17: 74847954-74847954 on Build GRCh38
rs764929779 Chr.17: 74847970-74847970 on Build GRCh38
rs370282910 Chr.17: 74847832-74847832 on Build GRCh38
rs763873866 Chr.17: 74847856-74847856 on Build GRCh38
rs374561167 Chr.17: 74847889-74847889 on Build GRCh38
rs753369315 Chr.17: 74847956-74847956 on Build GRCh38
rs755621550 Chr.17: 74847852-74847852 on Build GRCh38
rs754716736 Chr.17: 74847947-74847948 on Build GRCh38
rs751152216 Chr.17: 74848108-74848108 on Build GRCh38
rs987674687 Chr.17: 74848210-74848210 on Build GRCh38
rs1020310639 Chr.17: 74848093-74848093 on Build GRCh38
rs772465103 Chr.17: 74847928-74847928 on Build GRCh38
rs779128118 Chr.17: 74848015-74848015 on Build GRCh38
rs904995077 Chr.17: 74847934-74847934 on Build GRCh38
rs539932832 Chr.17: 74848009-74848009 on Build GRCh38
rs766674879 Chr.17: 74847833-74847833 on Build GRCh38
rs750198223 Chr.17: 74847971-74847971 on Build GRCh38
rs773385102 Chr.17: 74847830-74847830 on Build GRCh38
rs770026107 Chr.17: 74847827-74847827 on Build GRCh38
rs190480328 Chr.17: 74847997-74847997 on Build GRCh38
rs545207905 Chr.17: 74847913-74847913 on Build GRCh38
rs766110294 Chr.17: 74848053-74848053 on Build GRCh38
rs757043507 Chr.17: 74847806-74847806 on Build GRCh38
rs759891692 Chr.17: 74847922-74847922 on Build GRCh38
rs778614339 Chr.17: 74847873-74847873 on Build GRCh38
rs536614481 Chr.17: 74847822-74847822 on Build GRCh38
rs758325247 Chr.17: 74847881-74847881 on Build GRCh38
rs562778755 Chr.17: 74848042-74848042 on Build GRCh38
rs565053824 Chr.17: 74848084-74848084 on Build GRCh38
rs778921448 Chr.17: 74847813-74847813 on Build GRCh38
rs756949493 Chr.17: 74847867-74847867 on Build GRCh38
rs947875817 Chr.17: 74848195-74848195 on Build GRCh38
rs372730559 Chr.17: 74848010-74848010 on Build GRCh38
rs969134944 Chr.17: 74848094-74848094 on Build GRCh38
rs201241904 Chr.17: 74847918-74847918 on Build GRCh38
rs745849557 Chr.17: 74847814-74847814 on Build GRCh38
rs769823971 Chr.17: 74847904-74847904 on Build GRCh38
rs768060104 Chr.17: 74847839-74847839 on Build GRCh38
rs542562116 Chr.17: 74848037-74848037 on Build GRCh38
rs760286848 Chr.17: 74847936-74847936 on Build GRCh38
rs920349561 Chr.17: 74848190-74848190 on Build GRCh38
rs1036068341 Chr.17: 74847841-74847841 on Build GRCh38

View Full Product Details