Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7741547-7742090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD209
Entrez Gene ID
30835
Gene Name
CD209 molecule
Gene Aliases
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1
UniGene
Hs.278694
Gene Chromosome Location
Chr.19: 7739993-7747609 on Build GRCh38
Transcripts
NM_001144893.1 NM_001144894.1 NM_001144895.1 NM_001144896.1 NM_001144897.1 NM_001144899.1 NM_021155.3 NR_026692.1 XM_005272472.4

SNP Detail

SNP ID Chromosome Location
rs542451078 Chr.19: 7741956-7741956 on Build GRCh38
rs573735640 Chr.19: 7741950-7741950 on Build GRCh38
rs190284057 Chr.19: 7741734-7741734 on Build GRCh38
rs535014704 Chr.19: 7741764-7741764 on Build GRCh38
rs770005068 Chr.19: 7742010-7742010 on Build GRCh38
rs182574144 Chr.19: 7741850-7741850 on Build GRCh38
rs528395931 Chr.19: 7742016-7742016 on Build GRCh38
rs572817510 Chr.19: 7741757-7741757 on Build GRCh38
rs899450073 Chr.19: 7741694-7741694 on Build GRCh38
rs11465404 Chr.19: 7741597-7741597 on Build GRCh38
rs578023555 Chr.19: 7741844-7741844 on Build GRCh38
rs111742529 Chr.19: 7741720-7741720 on Build GRCh38
rs959431587 Chr.19: 7741797-7741797 on Build GRCh38
rs1049442577 Chr.19: 7741641-7741641 on Build GRCh38
rs1026905857 Chr.19: 7741707-7741707 on Build GRCh38
rs186513902 Chr.19: 7741573-7741573 on Build GRCh38
rs941110407 Chr.19: 7742005-7742005 on Build GRCh38
rs11465403 Chr.19: 7741705-7741705 on Build GRCh38
rs571230911 Chr.19: 7741599-7741599 on Build GRCh38
rs970855817 Chr.19: 7741712-7741712 on Build GRCh38
rs563655029 Chr.19: 7741920-7741920 on Build GRCh38
rs758346559 Chr.19: 7741787-7741787 on Build GRCh38
rs1544767 Chr.19: 7741982-7741982 on Build GRCh38
rs933848415 Chr.19: 7741937-7741937 on Build GRCh38
rs550418394 Chr.19: 7741653-7741653 on Build GRCh38
rs1544766 Chr.19: 7741837-7741837 on Build GRCh38
rs889565892 Chr.19: 7741997-7741997 on Build GRCh38
rs781528766 Chr.19: 7741973-7741973 on Build GRCh38
rs550603471 Chr.19: 7742056-7742056 on Build GRCh38
rs1003743319 Chr.19: 7742057-7742057 on Build GRCh38
rs570203160 Chr.19: 7741665-7741665 on Build GRCh38
rs751707109 Chr.19: 7741859-7741859 on Build GRCh38
rs564862388 Chr.19: 7742037-7742037 on Build GRCh38
rs567258680 Chr.19: 7742059-7742059 on Build GRCh38
rs568910734 Chr.19: 7742018-7742018 on Build GRCh38
rs537182979 Chr.19: 7741638-7741638 on Build GRCh38
rs369709603 Chr.19: 7741577-7741577 on Build GRCh38
rs757318089 Chr.19: 7741919-7741919 on Build GRCh38
rs889512119 Chr.19: 7741652-7741652 on Build GRCh38
rs1005202871 Chr.19: 7741654-7741654 on Build GRCh38
rs1047953074 Chr.19: 7742047-7742047 on Build GRCh38
rs906765832 Chr.19: 7742049-7742049 on Build GRCh38
rs759414242 Chr.19: 7741647-7741647 on Build GRCh38
rs372268442 Chr.19: 7741630-7741630 on Build GRCh38
rs945291789 Chr.19: 7741895-7741895 on Build GRCh38
rs11465402 Chr.19: 7742041-7742041 on Build GRCh38

View Full Product Details