Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8521148-8521680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO1F
Entrez Gene ID
4542
Gene Name
myosin IF
Gene Aliases
-
UniGene
Hs.465818
Gene Chromosome Location
Chr.19: 8520790-8577447 on Build GRCh38
Transcripts
NM_012335.3 XM_011528024.1 XM_011528025.1 XM_011528026.1 XM_011528027.1 XM_017026821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4542950 c.3286G>A p.E1096K Chr.19: 8521539-8521539 on Build GRCh38
4954962 c.3254G>T p.G1085V Chr.19: 8521571-8521571 on Build GRCh38
1003548 c.3227C>T p.S1076L Chr.19: 8521598-8521598 on Build GRCh38
715613 c.3253G>A p.G1085S Chr.19: 8521572-8521572 on Build GRCh38
6320750 c.3250C>A p.H1084N Chr.19: 8521575-8521575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13346588 Chr.19: 8521342-8521342 on Build GRCh38
rs944922976 Chr.19: 8521503-8521503 on Build GRCh38
rs1028128399 Chr.19: 8521184-8521184 on Build GRCh38
rs763012452 Chr.19: 8521497-8521497 on Build GRCh38
rs961550191 Chr.19: 8521459-8521459 on Build GRCh38
rs367568601 Chr.19: 8521580-8521580 on Build GRCh38
rs1054622 Chr.19: 8521237-8521237 on Build GRCh38
rs762362221 Chr.19: 8521618-8521618 on Build GRCh38
rs187544741 Chr.19: 8521629-8521629 on Build GRCh38
rs916916833 Chr.19: 8521278-8521278 on Build GRCh38
rs774252995 Chr.19: 8521581-8521581 on Build GRCh38
rs900514266 Chr.19: 8521370-8521370 on Build GRCh38
rs779568004 Chr.19: 8521635-8521635 on Build GRCh38
rs897001491 Chr.19: 8521617-8521617 on Build GRCh38
rs533611108 Chr.19: 8521498-8521498 on Build GRCh38
rs930871056 Chr.19: 8521396-8521396 on Build GRCh38
rs373721466 Chr.19: 8521492-8521492 on Build GRCh38
rs1008693374 Chr.19: 8521507-8521507 on Build GRCh38
rs774189690 Chr.19: 8521579-8521579 on Build GRCh38
rs541384191 Chr.19: 8521657-8521657 on Build GRCh38
rs764689507 Chr.19: 8521522-8521522 on Build GRCh38
rs926774337 Chr.19: 8521427-8521427 on Build GRCh38
rs745692023 Chr.19: 8521478-8521478 on Build GRCh38
rs754375689 Chr.19: 8521616-8521616 on Build GRCh38
rs1054619 Chr.19: 8521303-8521303 on Build GRCh38
rs372580076 Chr.19: 8521433-8521433 on Build GRCh38
rs775985136 Chr.19: 8521597-8521597 on Build GRCh38
rs888270674 Chr.19: 8521434-8521434 on Build GRCh38
rs1054618 Chr.19: 8521333-8521333 on Build GRCh38
rs944979165 Chr.19: 8521351-8521351 on Build GRCh38
rs138505600 Chr.19: 8521258-8521258 on Build GRCh38
rs778479659 Chr.19: 8521539-8521539 on Build GRCh38
rs746766742 Chr.19: 8521556-8521556 on Build GRCh38
rs895532930 Chr.19: 8521468-8521468 on Build GRCh38
rs34276116 Chr.19: 8521176-8521177 on Build GRCh38
rs747216890 Chr.19: 8521179-8521179 on Build GRCh38
rs932000722 Chr.19: 8521542-8521542 on Build GRCh38
rs767959721 Chr.19: 8521508-8521508 on Build GRCh38
rs987952932 Chr.19: 8521347-8521347 on Build GRCh38
rs540599977 Chr.19: 8521218-8521218 on Build GRCh38
rs759811979 Chr.19: 8521582-8521582 on Build GRCh38
rs772130350 Chr.19: 8521594-8521594 on Build GRCh38
rs1049199927 Chr.19: 8521411-8521411 on Build GRCh38
rs190908621 Chr.19: 8521494-8521494 on Build GRCh38
rs575779374 Chr.19: 8521241-8521241 on Build GRCh38
rs989344208 Chr.19: 8521496-8521496 on Build GRCh38
rs146225868 Chr.19: 8521349-8521349 on Build GRCh38
rs1045265639 Chr.19: 8521408-8521408 on Build GRCh38
rs200910214 Chr.19: 8521493-8521493 on Build GRCh38
rs759625300 Chr.19: 8521505-8521505 on Build GRCh38
rs969746798 Chr.19: 8521289-8521289 on Build GRCh38
rs756797956 Chr.19: 8521533-8521533 on Build GRCh38
rs186103433 Chr.19: 8521177-8521177 on Build GRCh38
rs753465471 Chr.19: 8521526-8521526 on Build GRCh38
rs371750206 Chr.19: 8521633-8521633 on Build GRCh38
rs184207950 Chr.19: 8521626-8521626 on Build GRCh38
rs544506974 Chr.19: 8521253-8521253 on Build GRCh38
rs780912457 Chr.19: 8521642-8521642 on Build GRCh38
rs926100423 Chr.19: 8521327-8521327 on Build GRCh38
rs1005833984 Chr.19: 8521440-8521440 on Build GRCh38
rs779066519 Chr.19: 8521653-8521653 on Build GRCh38
rs768646479 Chr.19: 8521572-8521572 on Build GRCh38
rs771881166 Chr.19: 8521479-8521479 on Build GRCh38
rs764738737 Chr.19: 8521602-8521602 on Build GRCh38
rs913512662 Chr.19: 8521350-8521350 on Build GRCh38
rs897934975 Chr.19: 8521460-8521460 on Build GRCh38
rs1015510167 Chr.19: 8521447-8521447 on Build GRCh38
rs769553424 Chr.19: 8521650-8521650 on Build GRCh38
rs748113084 Chr.19: 8521647-8521647 on Build GRCh38
rs370292190 Chr.19: 8521598-8521598 on Build GRCh38
rs374597578 Chr.19: 8521519-8521519 on Build GRCh38
rs996268828 Chr.19: 8521183-8521183 on Build GRCh38
rs905528394 Chr.19: 8521658-8521658 on Build GRCh38
rs758259822 Chr.19: 8521200-8521200 on Build GRCh38
rs757920201 Chr.19: 8521547-8521547 on Build GRCh38
rs781073533 Chr.19: 8521573-8521573 on Build GRCh38
rs751391836 Chr.19: 8521638-8521638 on Build GRCh38
rs141749665 Chr.19: 8521489-8521489 on Build GRCh38
rs1014181318 Chr.19: 8521465-8521465 on Build GRCh38
rs559272215 Chr.19: 8521640-8521640 on Build GRCh38
rs575777950 Chr.19: 8521386-8521386 on Build GRCh38
rs148428772 Chr.19: 8521352-8521352 on Build GRCh38
rs1054623 Chr.19: 8521202-8521202 on Build GRCh38
rs953331775 Chr.19: 8521504-8521504 on Build GRCh38
rs769420539 Chr.19: 8521216-8521216 on Build GRCh38
rs1009865591 Chr.19: 8521177-8521177 on Build GRCh38
rs547516698 Chr.19: 8521543-8521543 on Build GRCh38
rs1054621 Chr.19: 8521263-8521263 on Build GRCh38
rs780509524 Chr.19: 8521336-8521337 on Build GRCh38
rs779911108 Chr.19: 8521552-8521552 on Build GRCh38
rs952539849 Chr.19: 8521191-8521191 on Build GRCh38
rs752941186 Chr.19: 8521517-8521517 on Build GRCh38
rs141649251 Chr.19: 8521299-8521299 on Build GRCh38

View Full Product Details