Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10353441-10353932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYK2
Entrez Gene ID
7297
Gene Name
tyrosine kinase 2
Gene Aliases
IMD35, JTK1
UniGene
Hs.75516
Gene Chromosome Location
Chr.19: 10350528-10380572 on Build GRCh38
Transcripts
NM_003331.4 XM_011528245.1 XM_011528246.2 XM_011528247.1 XM_011528249.2 XM_011528251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1390086 c.2996A>G p.Q999R Chr.19: 10353559-10353559 on Build GRCh38
1231474 c.2974C>T p.R992W Chr.19: 10353581-10353581 on Build GRCh38
4876359 c.2997G>A p.Q999Q Chr.19: 10353558-10353558 on Build GRCh38
4738521 c.2932A>G p.M978V Chr.19: 10353623-10353623 on Build GRCh38
6890415 c.2973C>T p.P991P Chr.19: 10353582-10353582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550165008 Chr.19: 10353659-10353659 on Build GRCh38
rs758470420 Chr.19: 10353582-10353582 on Build GRCh38
rs933108783 Chr.19: 10353818-10353818 on Build GRCh38
rs531721749 Chr.19: 10353620-10353620 on Build GRCh38
rs566259323 Chr.19: 10353767-10353767 on Build GRCh38
rs576276690 Chr.19: 10353654-10353654 on Build GRCh38
rs999381843 Chr.19: 10353875-10353875 on Build GRCh38
rs749667172 Chr.19: 10353534-10353534 on Build GRCh38
rs914581858 Chr.19: 10353512-10353512 on Build GRCh38
rs943627776 Chr.19: 10353732-10353732 on Build GRCh38
rs990154876 Chr.19: 10353494-10353494 on Build GRCh38
rs749613263 Chr.19: 10353617-10353617 on Build GRCh38
rs148484862 Chr.19: 10353576-10353576 on Build GRCh38
rs142823384 Chr.19: 10353822-10353822 on Build GRCh38
rs895060492 Chr.19: 10353857-10353857 on Build GRCh38
rs1047718390 Chr.19: 10353591-10353591 on Build GRCh38
rs974651361 Chr.19: 10353805-10353805 on Build GRCh38
rs577866503 Chr.19: 10353885-10353885 on Build GRCh38
rs147442318 Chr.19: 10353570-10353570 on Build GRCh38
rs1048301147 Chr.19: 10353816-10353816 on Build GRCh38
rs545844060 Chr.19: 10353464-10353464 on Build GRCh38
rs753617141 Chr.19: 10353574-10353574 on Build GRCh38
rs746369465 Chr.19: 10353621-10353621 on Build GRCh38
rs1039399074 Chr.19: 10353551-10353551 on Build GRCh38
rs762883998 Chr.19: 10353691-10353691 on Build GRCh38
rs771379400 Chr.19: 10353536-10353536 on Build GRCh38
rs886468039 Chr.19: 10353599-10353599 on Build GRCh38
rs2304253 Chr.19: 10353682-10353682 on Build GRCh38
rs537502768 Chr.19: 10353723-10353723 on Build GRCh38
rs901645819 Chr.19: 10353562-10353562 on Build GRCh38
rs990680591 Chr.19: 10353759-10353759 on Build GRCh38
rs1079301 Chr.19: 10353830-10353830 on Build GRCh38
rs754602912 Chr.19: 10353600-10353600 on Build GRCh38
rs72563148 Chr.19: 10353886-10353886 on Build GRCh38
rs866757379 Chr.19: 10353537-10353537 on Build GRCh38
rs1043553040 Chr.19: 10353872-10353872 on Build GRCh38
rs112915394 Chr.19: 10353874-10353874 on Build GRCh38
rs764232789 Chr.19: 10353572-10353572 on Build GRCh38
rs201397594 Chr.19: 10353578-10353578 on Build GRCh38
rs751038982 Chr.19: 10353592-10353592 on Build GRCh38
rs1030646152 Chr.19: 10353695-10353695 on Build GRCh38
rs533669041 Chr.19: 10353789-10353789 on Build GRCh38
rs865983705 Chr.19: 10353531-10353531 on Build GRCh38
rs768261169 Chr.19: 10353499-10353499 on Build GRCh38
rs72563147 Chr.19: 10353880-10353880 on Build GRCh38
rs567693879 Chr.19: 10353619-10353619 on Build GRCh38
rs765114505 Chr.19: 10353669-10353669 on Build GRCh38
rs1007811933 Chr.19: 10353898-10353898 on Build GRCh38
rs12720358 Chr.19: 10353864-10353864 on Build GRCh38
rs780416116 Chr.19: 10353655-10353656 on Build GRCh38
rs760882131 Chr.19: 10353571-10353571 on Build GRCh38
rs760083588 Chr.19: 10353546-10353546 on Build GRCh38
rs1015911210 Chr.19: 10353609-10353609 on Build GRCh38
rs12720359 Chr.19: 10353462-10353462 on Build GRCh38
rs1006410018 Chr.19: 10353584-10353584 on Build GRCh38
rs370401636 Chr.19: 10353581-10353581 on Build GRCh38
rs765912261 Chr.19: 10353589-10353589 on Build GRCh38
rs762179978 Chr.19: 10353663-10353663 on Build GRCh38
rs780731468 Chr.19: 10353616-10353616 on Build GRCh38
rs12720321 Chr.19: 10353798-10353798 on Build GRCh38
rs372740500 Chr.19: 10353849-10353849 on Build GRCh38
rs762898203 Chr.19: 10353680-10353680 on Build GRCh38
rs1017935804 Chr.19: 10353912-10353912 on Build GRCh38
rs747077516 Chr.19: 10353651-10353651 on Build GRCh38
rs894841890 Chr.19: 10353673-10353673 on Build GRCh38
rs560137311 Chr.19: 10353890-10353890 on Build GRCh38
rs1022176322 Chr.19: 10353678-10353678 on Build GRCh38
rs551288067 Chr.19: 10353474-10353474 on Build GRCh38
rs554110174 Chr.19: 10353731-10353731 on Build GRCh38
rs772460702 Chr.19: 10353624-10353624 on Build GRCh38
rs757596255 Chr.19: 10353618-10353618 on Build GRCh38
rs202110875 Chr.19: 10353544-10353544 on Build GRCh38
rs1023180749 Chr.19: 10353693-10353693 on Build GRCh38
rs907971702 Chr.19: 10353819-10353819 on Build GRCh38
rs934634930 Chr.19: 10353492-10353492 on Build GRCh38
rs12720322 Chr.19: 10353702-10353702 on Build GRCh38
rs924717510 Chr.19: 10353491-10353491 on Build GRCh38
rs375387600 Chr.19: 10353661-10353661 on Build GRCh38
rs981589698 Chr.19: 10353703-10353703 on Build GRCh38
rs34669146 Chr.19: 10353614-10353614 on Build GRCh38
rs1044597782 Chr.19: 10353902-10353902 on Build GRCh38
rs772177678 Chr.19: 10353569-10353569 on Build GRCh38
rs763073939 Chr.19: 10353869-10353870 on Build GRCh38
rs923199793 Chr.19: 10353813-10353813 on Build GRCh38
rs775578531 Chr.19: 10353648-10353648 on Build GRCh38
rs920797604 Chr.19: 10353790-10353790 on Build GRCh38
rs778134416 Chr.19: 10353516-10353516 on Build GRCh38
rs746663364 Chr.19: 10353498-10353498 on Build GRCh38
rs1056479937 Chr.19: 10353833-10353833 on Build GRCh38
rs766243440 Chr.19: 10353692-10353692 on Build GRCh38

View Full Product Details