Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39201200-39201697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCCRP1
Entrez Gene ID
342897
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
FBXO50, NCCRP-1
UniGene
Hs.726934
Gene Chromosome Location
Chr.19: 39196964-39201882 on Build GRCh38
Transcripts
NM_001001414.1

SNP Detail

SNP ID Chromosome Location
rs955921853 Chr.19: 39201661-39201661 on Build GRCh38
rs879630267 Chr.19: 39201674-39201674 on Build GRCh38
rs889094511 Chr.19: 39201511-39201511 on Build GRCh38
rs942880234 Chr.19: 39201517-39201517 on Build GRCh38
rs543231113 Chr.19: 39201283-39201283 on Build GRCh38
rs935587793 Chr.19: 39201584-39201584 on Build GRCh38
rs1016846428 Chr.19: 39201430-39201430 on Build GRCh38
rs113033973 Chr.19: 39201313-39201313 on Build GRCh38
rs545911526 Chr.19: 39201335-39201335 on Build GRCh38
rs60036622 Chr.19: 39201328-39201329 on Build GRCh38
rs572488283 Chr.19: 39201590-39201590 on Build GRCh38
rs750987345 Chr.19: 39201603-39201603 on Build GRCh38
rs190245809 Chr.19: 39201341-39201341 on Build GRCh38
rs750507660 Chr.19: 39201243-39201243 on Build GRCh38
rs1036236167 Chr.19: 39201558-39201558 on Build GRCh38
rs555659840 Chr.19: 39201289-39201289 on Build GRCh38
rs866095254 Chr.19: 39201432-39201432 on Build GRCh38
rs898058232 Chr.19: 39201553-39201553 on Build GRCh38
rs978631107 Chr.19: 39201343-39201343 on Build GRCh38
rs960020399 Chr.19: 39201236-39201236 on Build GRCh38
rs915555150 Chr.19: 39201267-39201267 on Build GRCh38
rs538464400 Chr.19: 39201467-39201467 on Build GRCh38
rs545178005 Chr.19: 39201500-39201500 on Build GRCh38
rs960270534 Chr.19: 39201576-39201576 on Build GRCh38
rs924282416 Chr.19: 39201618-39201618 on Build GRCh38
rs149443907 Chr.19: 39201490-39201490 on Build GRCh38
rs1001816369 Chr.19: 39201374-39201374 on Build GRCh38
rs529188462 Chr.19: 39201672-39201672 on Build GRCh38
rs962607649 Chr.19: 39201439-39201439 on Build GRCh38
rs556473594 Chr.19: 39201483-39201483 on Build GRCh38
rs781742641 Chr.19: 39201471-39201471 on Build GRCh38
rs1006253594 Chr.19: 39201521-39201521 on Build GRCh38
rs564811483 Chr.19: 39201312-39201313 on Build GRCh38
rs1044317818 Chr.19: 39201536-39201536 on Build GRCh38
rs969900322 Chr.19: 39201366-39201366 on Build GRCh38
rs564198570 Chr.19: 39201337-39201337 on Build GRCh38
rs1014019258 Chr.19: 39201331-39201331 on Build GRCh38
rs561992481 Chr.19: 39201616-39201616 on Build GRCh38
rs924560180 Chr.19: 39201436-39201436 on Build GRCh38
rs551409231 Chr.19: 39201226-39201226 on Build GRCh38
rs972667123 Chr.19: 39201461-39201461 on Build GRCh38
rs186824026 Chr.19: 39201606-39201606 on Build GRCh38
rs911319047 Chr.19: 39201510-39201510 on Build GRCh38
rs533508340 Chr.19: 39201285-39201285 on Build GRCh38
rs34988938 Chr.19: 39201394-39201395 on Build GRCh38
rs1035883076 Chr.19: 39201231-39201231 on Build GRCh38
rs1037720938 Chr.19: 39201531-39201531 on Build GRCh38
rs576525625 Chr.19: 39201508-39201508 on Build GRCh38
rs182179459 Chr.19: 39201557-39201557 on Build GRCh38
rs933391906 Chr.19: 39201507-39201507 on Build GRCh38
rs949775784 Chr.19: 39201660-39201660 on Build GRCh38

View Full Product Details