Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51350271-51350480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETFB
Entrez Gene ID
2109
Gene Name
electron transfer flavoprotein beta subunit
Gene Aliases
FP585, MADD
UniGene
Hs.348531
Gene Chromosome Location
Chr.19: 51345155-51366418 on Build GRCh38
Transcripts
NM_001014763.1 NM_001985.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4965015 c.704G>A p.W235* Chr.19: 51350336-51350336 on Build GRCh38
3414127 c.408G>T p.Q136H Chr.19: 51350359-51350359 on Build GRCh38
7105551 c.378C>T p.A126A Chr.19: 51350389-51350389 on Build GRCh38
6656626 c.649-2A>C p.? Chr.19: 51350393-51350393 on Build GRCh38
4080589 c.706C>T p.P236S Chr.19: 51350334-51350334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944134585 Chr.19: 51350338-51350338 on Build GRCh38
rs149238320 Chr.19: 51350386-51350386 on Build GRCh38
rs745985216 Chr.19: 51350375-51350375 on Build GRCh38
rs777987200 Chr.19: 51350348-51350348 on Build GRCh38
rs761435953 Chr.19: 51350420-51350420 on Build GRCh38
rs114985874 Chr.19: 51350309-51350309 on Build GRCh38
rs373523434 Chr.19: 51350416-51350416 on Build GRCh38
rs104894678 Chr.19: 51350385-51350385 on Build GRCh38
rs746757758 Chr.19: 51350403-51350403 on Build GRCh38
rs200448956 Chr.19: 51350304-51350304 on Build GRCh38
rs1000451099 Chr.19: 51350454-51350454 on Build GRCh38
rs754039782 Chr.19: 51350365-51350365 on Build GRCh38
rs754119769 Chr.19: 51350415-51350416 on Build GRCh38
rs757361405 Chr.19: 51350372-51350372 on Build GRCh38
rs749987457 Chr.19: 51350299-51350299 on Build GRCh38
rs1042832534 Chr.19: 51350450-51350450 on Build GRCh38
rs371564310 Chr.19: 51350374-51350374 on Build GRCh38
rs912384773 Chr.19: 51350326-51350326 on Build GRCh38
rs768083453 Chr.19: 51350307-51350307 on Build GRCh38
rs370979664 Chr.19: 51350314-51350314 on Build GRCh38
rs769480387 Chr.19: 51350434-51350434 on Build GRCh38
rs768338384 Chr.19: 51350412-51350412 on Build GRCh38

View Full Product Details