Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54220260-54220760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRB3
Entrez Gene ID
11025
Gene Name
leukocyte immunoglobulin like receptor B3
Gene Aliases
CD85A, HL9, ILT-5, ILT5, LILRA6, LIR-3, LIR3, PIR-B, PIRB
UniGene
Hs.631592
Gene Chromosome Location
Chr.19: 54216278-54223087 on Build GRCh38
Transcripts
NM_001081450.2 NM_001320960.1 NM_006864.3 NR_135493.1 NR_135494.1 NR_135495.1 NR_135496.1 XM_011526381.2 XM_011526382.2 XM_017026218.1 XM_017026219.1 XM_017026220.1 XM_017026221.1 XM_017026222.1 XM_017026223.1

SNP Detail

SNP ID Chromosome Location
rs778389068 Chr.19: 54220583-54220583 on Build GRCh38
rs1052985 Chr.19: 54220741-54220741 on Build GRCh38
rs767911387 Chr.19: 54220615-54220615 on Build GRCh38
rs761071657 Chr.19: 54220619-54220619 on Build GRCh38
rs367643312 Chr.19: 54220652-54220652 on Build GRCh38
rs201553860 Chr.19: 54220603-54220603 on Build GRCh38
rs765459269 Chr.19: 54220508-54220508 on Build GRCh38
rs760598744 Chr.19: 54220502-54220502 on Build GRCh38
rs561742830 Chr.19: 54220354-54220354 on Build GRCh38
rs570493959 Chr.19: 54220392-54220392 on Build GRCh38
rs756156246 Chr.19: 54220699-54220699 on Build GRCh38
rs765393514 Chr.19: 54220564-54220564 on Build GRCh38
rs529579591 Chr.19: 54220297-54220297 on Build GRCh38
rs752182138 Chr.19: 54220518-54220518 on Build GRCh38
rs201735825 Chr.19: 54220573-54220573 on Build GRCh38
rs529161494 Chr.19: 54220622-54220622 on Build GRCh38
rs751294272 Chr.19: 54220690-54220690 on Build GRCh38
rs751953523 Chr.19: 54220578-54220578 on Build GRCh38
rs772007174 Chr.19: 54220483-54220483 on Build GRCh38
rs748787411 Chr.19: 54220521-54220521 on Build GRCh38
rs539638589 Chr.19: 54220409-54220409 on Build GRCh38
rs774597233 Chr.19: 54220613-54220613 on Build GRCh38
rs771265921 Chr.19: 54220587-54220593 on Build GRCh38
rs778843337 Chr.19: 54220734-54220734 on Build GRCh38
rs111355853 Chr.19: 54220511-54220511 on Build GRCh38
rs145718796 Chr.19: 54220658-54220658 on Build GRCh38
rs371484692 Chr.19: 54220670-54220670 on Build GRCh38
rs16985497 Chr.19: 54220736-54220736 on Build GRCh38
rs1052993 Chr.19: 54220588-54220588 on Build GRCh38
rs747334922 Chr.19: 54220551-54220551 on Build GRCh38
rs757732370 Chr.19: 54220579-54220579 on Build GRCh38
rs764776278 Chr.19: 54220620-54220620 on Build GRCh38
rs767397347 Chr.19: 54220695-54220695 on Build GRCh38
rs778263053 Chr.19: 54220545-54220546 on Build GRCh38
rs768972757 Chr.19: 54220610-54220610 on Build GRCh38
rs200786676 Chr.19: 54220546-54220546 on Build GRCh38
rs375174631 Chr.19: 54220724-54220724 on Build GRCh38
rs754441781 Chr.19: 54220581-54220581 on Build GRCh38
rs774159228 Chr.19: 54220480-54220480 on Build GRCh38
rs535771586 Chr.19: 54220462-54220462 on Build GRCh38
rs796363001 Chr.19: 54220363-54220363 on Build GRCh38
rs773065803 Chr.19: 54220497-54220497 on Build GRCh38
rs201534767 Chr.19: 54220517-54220517 on Build GRCh38
rs1132608 Chr.19: 54220537-54220537 on Build GRCh38
rs756849617 Chr.19: 54220691-54220691 on Build GRCh38
rs780074442 Chr.19: 54220705-54220705 on Build GRCh38
rs569949929 Chr.19: 54220443-54220443 on Build GRCh38
rs558231147 Chr.19: 54220510-54220510 on Build GRCh38
rs771919192 Chr.19: 54220549-54220549 on Build GRCh38
rs908129965 Chr.19: 54220468-54220468 on Build GRCh38
rs984948975 Chr.19: 54220466-54220466 on Build GRCh38
rs777356243 Chr.19: 54220627-54220627 on Build GRCh38
rs770797369 Chr.19: 54220592-54220592 on Build GRCh38
rs376658580 Chr.19: 54220673-54220673 on Build GRCh38
rs780847886 Chr.19: 54220653-54220653 on Build GRCh38
rs753298938 Chr.19: 54220580-54220580 on Build GRCh38
rs1052995 Chr.19: 54220587-54220587 on Build GRCh38
rs368403726 Chr.19: 54220482-54220482 on Build GRCh38
rs750157279 Chr.19: 54220698-54220698 on Build GRCh38
rs201291137 Chr.19: 54220520-54220520 on Build GRCh38
rs369888262 Chr.19: 54220686-54220686 on Build GRCh38
rs367877729 Chr.19: 54220522-54220522 on Build GRCh38
rs368800344 Chr.19: 54220381-54220381 on Build GRCh38
rs372212249 Chr.19: 54220377-54220377 on Build GRCh38
rs1132605 Chr.19: 54220561-54220561 on Build GRCh38
rs769836804 Chr.19: 54220657-54220657 on Build GRCh38
rs763319782 Chr.19: 54220605-54220605 on Build GRCh38
rs762176596 Chr.19: 54220614-54220614 on Build GRCh38
rs745941530 Chr.19: 54220595-54220595 on Build GRCh38
rs112557327 Chr.19: 54220529-54220529 on Build GRCh38
rs775701907 Chr.19: 54220604-54220604 on Build GRCh38
rs563620415 Chr.19: 54220284-54220284 on Build GRCh38
rs555429239 Chr.19: 54220473-54220473 on Build GRCh38
rs745878582 Chr.19: 54220655-54220655 on Build GRCh38
rs751004807 Chr.19: 54220616-54220616 on Build GRCh38
rs763265915 Chr.19: 54220575-54220575 on Build GRCh38
rs202100495 Chr.19: 54220550-54220550 on Build GRCh38
rs772977943 Chr.19: 54220559-54220559 on Build GRCh38
rs199865602 Chr.19: 54220507-54220507 on Build GRCh38
rs776749953 Chr.19: 54220506-54220506 on Build GRCh38
rs764615783 Chr.19: 54220577-54220577 on Build GRCh38
rs368027150 Chr.19: 54220309-54220309 on Build GRCh38
rs768319822 Chr.19: 54220479-54220479 on Build GRCh38
rs140601439 Chr.19: 54220662-54220662 on Build GRCh38
rs200018123 Chr.19: 54220519-54220519 on Build GRCh38
rs138012018 Chr.19: 54220687-54220687 on Build GRCh38
rs200516269 Chr.19: 54220679-54220679 on Build GRCh38
rs761209714 Chr.19: 54220674-54220674 on Build GRCh38
rs758078761 Chr.19: 54220625-54220625 on Build GRCh38
rs550028683 Chr.19: 54220442-54220442 on Build GRCh38
rs1052987 Chr.19: 54220738-54220738 on Build GRCh38
rs371802808 Chr.19: 54220723-54220723 on Build GRCh38
rs752402706 Chr.19: 54220584-54220584 on Build GRCh38
rs751327297 Chr.19: 54220636-54220636 on Build GRCh38
rs1132606 Chr.19: 54220560-54220560 on Build GRCh38
rs754606243 Chr.19: 54220524-54220524 on Build GRCh38
rs921662537 Chr.19: 54220415-54220415 on Build GRCh38
rs150467020 Chr.19: 54220628-54220628 on Build GRCh38
rs200497859 Chr.19: 54220551-54220551 on Build GRCh38
rs879448849 Chr.19: 54220587-54220588 on Build GRCh38
rs1132607 Chr.19: 54220541-54220541 on Build GRCh38
rs781128474 Chr.19: 54220594-54220594 on Build GRCh38
rs145725393 Chr.19: 54220669-54220669 on Build GRCh38
rs757012079 Chr.19: 54220641-54220641 on Build GRCh38

View Full Product Details