Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54253935-54254442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LILRB5
Entrez Gene ID
10990
Gene Name
leukocyte immunoglobulin like receptor B5
Gene Aliases
CD85C, LIR-8, LIR8
UniGene
Hs.655338
Gene Chromosome Location
Chr.19: 54250405-54257308 on Build GRCh38
Transcripts
NM_001081442.2 NM_001081443.2 NM_001304457.1 NM_006840.4 XM_011526359.2 XM_011526360.2 XM_011526361.2 XM_011526362.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5685642 c.1262C>A p.S421Y Chr.19: 54254409-54254409 on Build GRCh38
5678181 c.1350G>T p.Q450H Chr.19: 54254022-54254022 on Build GRCh38
4639070 c.1331C>T p.P444L Chr.19: 54254041-54254041 on Build GRCh38
4755538 c.1312G>A p.E438K Chr.19: 54254060-54254060 on Build GRCh38
4525230 c.1315G>A p.D439N Chr.19: 54254057-54254057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745443183 Chr.19: 54253986-54253986 on Build GRCh38
rs774461065 Chr.19: 54254319-54254319 on Build GRCh38
rs756703766 Chr.19: 54253981-54253981 on Build GRCh38
rs769642003 Chr.19: 54253987-54253987 on Build GRCh38
rs1037723754 Chr.19: 54254136-54254136 on Build GRCh38
rs531672779 Chr.19: 54254165-54254165 on Build GRCh38
rs779725229 Chr.19: 54254112-54254112 on Build GRCh38
rs758542287 Chr.19: 54254412-54254412 on Build GRCh38
rs940221672 Chr.19: 54254286-54254286 on Build GRCh38
rs202131847 Chr.19: 54254064-54254064 on Build GRCh38
rs773226206 Chr.19: 54254327-54254327 on Build GRCh38
rs766614279 Chr.19: 54254040-54254040 on Build GRCh38
rs532096157 Chr.19: 54254320-54254320 on Build GRCh38
rs780850726 Chr.19: 54253983-54253983 on Build GRCh38
rs751333155 Chr.19: 54254040-54254040 on Build GRCh38
rs779392850 Chr.19: 54253977-54253977 on Build GRCh38
rs935639485 Chr.19: 54254292-54254292 on Build GRCh38
rs367868825 Chr.19: 54254408-54254408 on Build GRCh38
rs932390177 Chr.19: 54254244-54254244 on Build GRCh38
rs561939835 Chr.19: 54254212-54254212 on Build GRCh38
rs911612500 Chr.19: 54254009-54254009 on Build GRCh38
rs774274476 Chr.19: 54254003-54254003 on Build GRCh38
rs201808266 Chr.19: 54254080-54254080 on Build GRCh38
rs7258131 Chr.19: 54254253-54254253 on Build GRCh38
rs920906246 Chr.19: 54254216-54254216 on Build GRCh38
rs768523068 Chr.19: 54254318-54254318 on Build GRCh38
rs375473834 Chr.19: 54254007-54254007 on Build GRCh38
rs765423190 Chr.19: 54253997-54254000 on Build GRCh38
rs1024560209 Chr.19: 54254142-54254142 on Build GRCh38
rs370888692 Chr.19: 54254093-54254093 on Build GRCh38
rs143092695 Chr.19: 54254028-54254028 on Build GRCh38
rs370294576 Chr.19: 54254004-54254004 on Build GRCh38
rs756762328 Chr.19: 54254081-54254081 on Build GRCh38
rs189712762 Chr.19: 54254160-54254160 on Build GRCh38
rs375671699 Chr.19: 54254373-54254373 on Build GRCh38
rs1038822131 Chr.19: 54254050-54254050 on Build GRCh38
rs765514520 Chr.19: 54254360-54254360 on Build GRCh38
rs774407775 Chr.19: 54254008-54254008 on Build GRCh38
rs942916615 Chr.19: 54254219-54254219 on Build GRCh38
rs755498160 Chr.19: 54253968-54253968 on Build GRCh38
rs755786612 Chr.19: 54254108-54254108 on Build GRCh38
rs930107622 Chr.19: 54253996-54253996 on Build GRCh38
rs757412574 Chr.19: 54254167-54254167 on Build GRCh38
rs865801780 Chr.19: 54254406-54254406 on Build GRCh38
rs376977892 Chr.19: 54253973-54253973 on Build GRCh38
rs907071426 Chr.19: 54254157-54254157 on Build GRCh38
rs891726776 Chr.19: 54254110-54254110 on Build GRCh38
rs896065974 Chr.19: 54254137-54254137 on Build GRCh38
rs112804184 Chr.19: 54254266-54254266 on Build GRCh38
rs775204610 Chr.19: 54253994-54253994 on Build GRCh38
rs907015226 Chr.19: 54254396-54254396 on Build GRCh38
rs767664795 Chr.19: 54254016-54254016 on Build GRCh38
rs527892378 Chr.19: 54254213-54254213 on Build GRCh38
rs754227621 Chr.19: 54254044-54254044 on Build GRCh38
rs1054057474 Chr.19: 54254298-54254298 on Build GRCh38
rs762008141 Chr.19: 54254015-54254015 on Build GRCh38
rs545894661 Chr.19: 54254123-54254123 on Build GRCh38
rs12611057 Chr.19: 54253957-54253957 on Build GRCh38
rs568897572 Chr.19: 54254382-54254382 on Build GRCh38
rs753213992 Chr.19: 54254366-54254366 on Build GRCh38
rs539865080 Chr.19: 54254092-54254092 on Build GRCh38
rs760806615 Chr.19: 54254330-54254330 on Build GRCh38
rs1037183587 Chr.19: 54254287-54254287 on Build GRCh38
rs945963048 Chr.19: 54254376-54254376 on Build GRCh38
rs571081370 Chr.19: 54254254-54254254 on Build GRCh38
rs541967333 Chr.19: 54254193-54254193 on Build GRCh38
rs951618290 Chr.19: 54254183-54254183 on Build GRCh38
rs746565876 Chr.19: 54253978-54253978 on Build GRCh38
rs1025874071 Chr.19: 54254182-54254182 on Build GRCh38
rs943722058 Chr.19: 54254037-54254037 on Build GRCh38
rs757144931 Chr.19: 54254372-54254372 on Build GRCh38
rs988426259 Chr.19: 54253990-54253990 on Build GRCh38
rs900153459 Chr.19: 54254072-54254072 on Build GRCh38
rs974310207 Chr.19: 54253966-54253966 on Build GRCh38
rs974089875 Chr.19: 54254179-54254179 on Build GRCh38
rs974880094 Chr.19: 54254241-54254241 on Build GRCh38
rs537937144 Chr.19: 54254388-54254388 on Build GRCh38
rs962688154 Chr.19: 54254176-54254176 on Build GRCh38
rs909624518 Chr.19: 54254285-54254285 on Build GRCh38
rs987986307 Chr.19: 54254185-54254185 on Build GRCh38
rs551870057 Chr.19: 54254340-54254340 on Build GRCh38
rs771102337 Chr.19: 54254339-54254339 on Build GRCh38
rs772292829 Chr.19: 54254322-54254322 on Build GRCh38
rs1022908858 Chr.19: 54254151-54254151 on Build GRCh38
rs1033546756 Chr.19: 54254350-54254350 on Build GRCh38
rs531344109 Chr.19: 54254038-54254038 on Build GRCh38
rs765168898 Chr.19: 54254175-54254175 on Build GRCh38
rs760752339 Chr.19: 54254030-54254030 on Build GRCh38

View Full Product Details