Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52354374-52354879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1B
Entrez Gene ID
200014
Gene Name
coiled-coil and C2 domain containing 1B
Gene Aliases
-
UniGene
Hs.591451
Gene Chromosome Location
Chr.1: 52348437-52366291 on Build GRCh38
Transcripts
NM_032449.2 XM_005270590.2 XM_005270591.2 XM_005270593.2 XM_006710423.2 XM_017000559.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3400872 c.524T>C p.L175P Chr.1: 52354657-52354657 on Build GRCh38
5521569 c.1699G>T p.G567W Chr.1: 52354840-52354840 on Build GRCh38
5811152 c.2344A>G p.I782V Chr.1: 52354853-52354853 on Build GRCh38
6621103 c.573G>T p.E191D Chr.1: 52354608-52354608 on Build GRCh38
3419306 c.461A>C p.K154T Chr.1: 52354861-52354861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759140758 Chr.1: 52354456-52354456 on Build GRCh38
rs1024987884 Chr.1: 52354540-52354540 on Build GRCh38
rs201834369 Chr.1: 52354631-52354631 on Build GRCh38
rs773042929 Chr.1: 52354705-52354705 on Build GRCh38
rs144497192 Chr.1: 52354647-52354647 on Build GRCh38
rs758047992 Chr.1: 52354714-52354716 on Build GRCh38
rs747485105 Chr.1: 52354601-52354601 on Build GRCh38
rs770089143 Chr.1: 52354772-52354772 on Build GRCh38
rs765341858 Chr.1: 52354732-52354732 on Build GRCh38
rs748941720 Chr.1: 52354615-52354615 on Build GRCh38
rs754975155 Chr.1: 52354644-52354644 on Build GRCh38
rs780819082 Chr.1: 52354795-52354795 on Build GRCh38
rs779464501 Chr.1: 52354569-52354569 on Build GRCh38
rs780110772 Chr.1: 52354557-52354557 on Build GRCh38
rs771686013 Chr.1: 52354802-52354802 on Build GRCh38
rs372793189 Chr.1: 52354717-52354717 on Build GRCh38
rs773817214 Chr.1: 52354450-52354450 on Build GRCh38
rs752911837 Chr.1: 52354646-52354646 on Build GRCh38
rs375103060 Chr.1: 52354789-52354789 on Build GRCh38
rs368295888 Chr.1: 52354799-52354799 on Build GRCh38
rs201162397 Chr.1: 52354804-52354804 on Build GRCh38
rs748858934 Chr.1: 52354673-52354673 on Build GRCh38
rs760652283 Chr.1: 52354635-52354635 on Build GRCh38
rs776385917 Chr.1: 52354720-52354720 on Build GRCh38
rs574096429 Chr.1: 52354825-52354825 on Build GRCh38
rs909768022 Chr.1: 52354704-52354704 on Build GRCh38
rs751195632 Chr.1: 52354727-52354728 on Build GRCh38
rs983990550 Chr.1: 52354428-52354428 on Build GRCh38
rs752239017 Chr.1: 52354843-52354843 on Build GRCh38
rs748717360 Chr.1: 52354427-52354427 on Build GRCh38
rs146416404 Chr.1: 52354645-52354645 on Build GRCh38
rs753058125 Chr.1: 52354437-52354437 on Build GRCh38
rs750309077 Chr.1: 52354493-52354493 on Build GRCh38
rs202002125 Chr.1: 52354542-52354543 on Build GRCh38
rs9660764 Chr.1: 52354668-52354668 on Build GRCh38
rs754070552 Chr.1: 52354568-52354568 on Build GRCh38
rs749855699 Chr.1: 52354564-52354565 on Build GRCh38
rs767238948 Chr.1: 52354632-52354632 on Build GRCh38
rs536513021 Chr.1: 52354663-52354663 on Build GRCh38
rs766423165 Chr.1: 52354636-52354636 on Build GRCh38
rs775690054 Chr.1: 52354811-52354811 on Build GRCh38
rs1041477395 Chr.1: 52354767-52354767 on Build GRCh38
rs777868711 Chr.1: 52354630-52354630 on Build GRCh38
rs990457620 Chr.1: 52354556-52354556 on Build GRCh38
rs1054430599 Chr.1: 52354594-52354594 on Build GRCh38
rs546741428 Chr.1: 52354521-52354521 on Build GRCh38
rs764837536 Chr.1: 52354418-52354419 on Build GRCh38
rs761777888 Chr.1: 52354491-52354491 on Build GRCh38
rs41312652 Chr.1: 52354788-52354788 on Build GRCh38
rs972773586 Chr.1: 52354582-52354582 on Build GRCh38
rs749465385 Chr.1: 52354413-52354413 on Build GRCh38
rs764408084 Chr.1: 52354819-52354819 on Build GRCh38
rs1039101713 Chr.1: 52354520-52354520 on Build GRCh38
rs754486502 Chr.1: 52354797-52354797 on Build GRCh38
rs757521860 Chr.1: 52354750-52354750 on Build GRCh38
rs1040719444 Chr.1: 52354442-52354442 on Build GRCh38
rs766084997 Chr.1: 52354725-52354726 on Build GRCh38
rs768902745 Chr.1: 52354414-52354414 on Build GRCh38
rs770576894 Chr.1: 52354805-52354805 on Build GRCh38
rs758705154 Chr.1: 52354443-52354443 on Build GRCh38
rs756595465 Chr.1: 52354837-52354837 on Build GRCh38
rs778439710 Chr.1: 52354587-52354587 on Build GRCh38
rs370812466 Chr.1: 52354730-52354730 on Build GRCh38
rs769290817 Chr.1: 52354461-52354461 on Build GRCh38
rs375797081 Chr.1: 52354809-52354809 on Build GRCh38
rs534008580 Chr.1: 52354777-52354777 on Build GRCh38
rs780159851 Chr.1: 52354409-52354409 on Build GRCh38
rs763307100 Chr.1: 52354812-52354812 on Build GRCh38
rs368215179 Chr.1: 52354682-52354682 on Build GRCh38
rs747529075 Chr.1: 52354660-52354660 on Build GRCh38
rs945831456 Chr.1: 52354453-52354453 on Build GRCh38
rs775010394 Chr.1: 52354464-52354464 on Build GRCh38
rs763852227 Chr.1: 52354478-52354478 on Build GRCh38
rs751332540 Chr.1: 52354490-52354490 on Build GRCh38
rs757858268 Chr.1: 52354846-52354846 on Build GRCh38
rs369565318 Chr.1: 52354790-52354790 on Build GRCh38
rs1030895939 Chr.1: 52354528-52354528 on Build GRCh38
rs772518347 Chr.1: 52354579-52354579 on Build GRCh38
rs200270526 Chr.1: 52354810-52354810 on Build GRCh38
rs528703743 Chr.1: 52354435-52354435 on Build GRCh38
rs532128379 Chr.1: 52354440-52354440 on Build GRCh38
rs751920219 Chr.1: 52354748-52354748 on Build GRCh38
rs543621141 Chr.1: 52354530-52354532 on Build GRCh38
rs762662895 Chr.1: 52354469-52354469 on Build GRCh38
rs911176063 Chr.1: 52354433-52354433 on Build GRCh38
rs1012627066 Chr.1: 52354533-52354533 on Build GRCh38
rs1023408150 Chr.1: 52354564-52354564 on Build GRCh38
rs753036811 Chr.1: 52354745-52354745 on Build GRCh38
rs768323645 Chr.1: 52354625-52354625 on Build GRCh38
rs768104151 Chr.1: 52354680-52354680 on Build GRCh38
rs572925926 Chr.1: 52354747-52354747 on Build GRCh38
rs374849367 Chr.1: 52354826-52354826 on Build GRCh38
rs753926100 Chr.1: 52354639-52354639 on Build GRCh38
rs933395361 Chr.1: 52354784-52354784 on Build GRCh38
rs1053964898 Chr.1: 52354779-52354779 on Build GRCh38
rs751002979 Chr.1: 52354835-52354835 on Build GRCh38
rs778278216 Chr.1: 52354656-52354656 on Build GRCh38
rs747730332 Chr.1: 52354683-52354683 on Build GRCh38
rs756159261 Chr.1: 52354504-52354504 on Build GRCh38
rs769256266 Chr.1: 52354603-52354603 on Build GRCh38
rs767704421 Chr.1: 52354833-52354833 on Build GRCh38
rs773135163 Chr.1: 52354633-52354633 on Build GRCh38
rs180997825 Chr.1: 52354532-52354532 on Build GRCh38
rs553799683 Chr.1: 52354851-52354851 on Build GRCh38
rs752076347 Chr.1: 52354550-52354550 on Build GRCh38
rs779412267 Chr.1: 52354421-52354421 on Build GRCh38
rs1032557433 Chr.1: 52354539-52354539 on Build GRCh38
rs887775562 Chr.1: 52354476-52354476 on Build GRCh38
rs866517513 Chr.1: 52354806-52354806 on Build GRCh38
rs2820334 Chr.1: 52354554-52354554 on Build GRCh38
rs760573414 Chr.1: 52354712-52354712 on Build GRCh38
rs775052653 Chr.1: 52354606-52354606 on Build GRCh38
rs748557856 Chr.1: 52354570-52354570 on Build GRCh38
rs904311126 Chr.1: 52354769-52354769 on Build GRCh38
rs941995134 Chr.1: 52354488-52354488 on Build GRCh38
rs201575152 Chr.1: 52354522-52354522 on Build GRCh38
rs745971774 Chr.1: 52354703-52354703 on Build GRCh38
rs754815981 Chr.1: 52354738-52354739 on Build GRCh38
rs906520911 Chr.1: 52354796-52354796 on Build GRCh38
rs373811663 Chr.1: 52354422-52354422 on Build GRCh38

View Full Product Details