Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54174213-54174739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5RL
Entrez Gene ID
606495
Gene Name
cytochrome b5 reductase like
Gene Aliases
-
UniGene
Hs.591426
Gene Chromosome Location
Chr.1: 54172336-54200073 on Build GRCh38
Transcripts
NM_001031672.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919815 c.848A>G p.K283R Chr.1: 54174719-54174719 on Build GRCh38
4885258 c.925G>T p.E309* Chr.1: 54174642-54174642 on Build GRCh38
5086532 c.931T>G p.S311A Chr.1: 54174636-54174636 on Build GRCh38
3985143 c.845G>A p.R282K Chr.1: 54174722-54174722 on Build GRCh38
6888745 c.932C>T p.S311F Chr.1: 54174635-54174635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998152422 Chr.1: 54174433-54174433 on Build GRCh38
rs918966021 Chr.1: 54174565-54174565 on Build GRCh38
rs777476366 Chr.1: 54174581-54174581 on Build GRCh38
rs61741270 Chr.1: 54174655-54174655 on Build GRCh38
rs1056731180 Chr.1: 54174252-54174252 on Build GRCh38
rs554995442 Chr.1: 54174486-54174486 on Build GRCh38
rs948127991 Chr.1: 54174283-54174283 on Build GRCh38
rs780876546 Chr.1: 54174711-54174711 on Build GRCh38
rs530303527 Chr.1: 54174712-54174712 on Build GRCh38
rs559056030 Chr.1: 54174392-54174392 on Build GRCh38
rs375961783 Chr.1: 54174613-54174613 on Build GRCh38
rs758325962 Chr.1: 54174694-54174694 on Build GRCh38
rs555009230 Chr.1: 54174335-54174335 on Build GRCh38
rs1035545029 Chr.1: 54174409-54174409 on Build GRCh38
rs753489709 Chr.1: 54174657-54174657 on Build GRCh38
rs755737482 Chr.1: 54174692-54174692 on Build GRCh38
rs181141391 Chr.1: 54174281-54174281 on Build GRCh38
rs759133773 Chr.1: 54174672-54174672 on Build GRCh38
rs752455211 Chr.1: 54174691-54174691 on Build GRCh38
rs186035697 Chr.1: 54174674-54174674 on Build GRCh38
rs974857450 Chr.1: 54174562-54174562 on Build GRCh38
rs771042809 Chr.1: 54174591-54174591 on Build GRCh38
rs1032211741 Chr.1: 54174421-54174421 on Build GRCh38
rs751330524 Chr.1: 54174695-54174695 on Build GRCh38
rs775730518 Chr.1: 54174638-54174638 on Build GRCh38
rs368711832 Chr.1: 54174681-54174681 on Build GRCh38
rs774845290 Chr.1: 54174652-54174652 on Build GRCh38
rs539168115 Chr.1: 54174346-54174346 on Build GRCh38
rs755886673 Chr.1: 54174564-54174564 on Build GRCh38
rs769874126 Chr.1: 54174623-54174623 on Build GRCh38
rs61741274 Chr.1: 54174645-54174645 on Build GRCh38
rs546818088 Chr.1: 54174259-54174259 on Build GRCh38
rs956151271 Chr.1: 54174424-54174424 on Build GRCh38
rs745637580 Chr.1: 54174537-54174537 on Build GRCh38
rs746870652 Chr.1: 54174586-54174586 on Build GRCh38
rs780279305 Chr.1: 54174715-54174715 on Build GRCh38
rs781522510 Chr.1: 54174607-54174607 on Build GRCh38
rs542849665 Chr.1: 54174466-54174466 on Build GRCh38
rs1044249148 Chr.1: 54174320-54174320 on Build GRCh38
rs1002755173 Chr.1: 54174360-54174360 on Build GRCh38
rs1016333854 Chr.1: 54174538-54174538 on Build GRCh38
rs757112805 Chr.1: 54174702-54174702 on Build GRCh38
rs762288981 Chr.1: 54174654-54174654 on Build GRCh38
rs769265229 Chr.1: 54174719-54174719 on Build GRCh38
rs141591932 Chr.1: 54174458-54174458 on Build GRCh38
rs929005596 Chr.1: 54174245-54174245 on Build GRCh38
rs763937870 Chr.1: 54174616-54174616 on Build GRCh38
rs763491587 Chr.1: 54174640-54174640 on Build GRCh38
rs199663086 Chr.1: 54174718-54174718 on Build GRCh38
rs954255078 Chr.1: 54174467-54174467 on Build GRCh38
rs1029682472 Chr.1: 54174451-54174451 on Build GRCh38
rs929068902 Chr.1: 54174595-54174595 on Build GRCh38
rs757206577 Chr.1: 54174277-54174277 on Build GRCh38
rs963197085 Chr.1: 54174541-54174541 on Build GRCh38
rs377135102 Chr.1: 54174628-54174628 on Build GRCh38
rs372959398 Chr.1: 54174588-54174588 on Build GRCh38
rs569280503 Chr.1: 54174397-54174397 on Build GRCh38
rs910040821 Chr.1: 54174518-54174518 on Build GRCh38

View Full Product Details