Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46761563-46762078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS5
Entrez Gene ID
9655
Gene Name
suppressor of cytokine signaling 5
Gene Aliases
CIS6, CISH6, Cish5, SOCS-5
UniGene
Hs.468426
Gene Chromosome Location
Chr.2: 46698960-46762788 on Build GRCh38
Transcripts
NM_014011.4 NM_144949.2

SNP Detail

SNP ID Chromosome Location
rs936525169 Chr.2: 46761647-46761647 on Build GRCh38
rs957774124 Chr.2: 46761876-46761876 on Build GRCh38
rs925023269 Chr.2: 46761636-46761636 on Build GRCh38
rs568397480 Chr.2: 46761979-46761979 on Build GRCh38
rs985708550 Chr.2: 46761893-46761893 on Build GRCh38
rs41418645 Chr.2: 46761681-46761681 on Build GRCh38
rs563575419 Chr.2: 46761645-46761645 on Build GRCh38
rs111672860 Chr.2: 46761691-46761691 on Build GRCh38
rs918495654 Chr.2: 46761951-46761951 on Build GRCh38
rs191708882 Chr.2: 46761717-46761717 on Build GRCh38
rs929986935 Chr.2: 46761966-46761966 on Build GRCh38
rs1034501571 Chr.2: 46761747-46761747 on Build GRCh38
rs762304268 Chr.2: 46762004-46762004 on Build GRCh38
rs374087214 Chr.2: 46761660-46761660 on Build GRCh38
rs531033768 Chr.2: 46761716-46761716 on Build GRCh38
rs976675104 Chr.2: 46761946-46761946 on Build GRCh38
rs1032103459 Chr.2: 46761867-46761867 on Build GRCh38
rs775081189 Chr.2: 46761819-46761819 on Build GRCh38
rs888497325 Chr.2: 46761732-46761732 on Build GRCh38
rs528698409 Chr.2: 46761961-46761961 on Build GRCh38
rs748657651 Chr.2: 46761699-46761699 on Build GRCh38
rs977747622 Chr.2: 46761856-46761856 on Build GRCh38
rs546816851 Chr.2: 46761972-46761972 on Build GRCh38
rs557452740 Chr.2: 46761932-46761932 on Build GRCh38
rs764745339 Chr.2: 46761766-46761766 on Build GRCh38
rs547778322 Chr.2: 46762023-46762023 on Build GRCh38
rs888547770 Chr.2: 46762035-46762035 on Build GRCh38
rs563715351 Chr.2: 46761713-46761713 on Build GRCh38
rs943939271 Chr.2: 46761666-46761666 on Build GRCh38
rs1002111059 Chr.2: 46761736-46761736 on Build GRCh38
rs879223186 Chr.2: 46762042-46762042 on Build GRCh38
rs535785023 Chr.2: 46762008-46762008 on Build GRCh38
rs911084813 Chr.2: 46761911-46761911 on Build GRCh38
rs1020530762 Chr.2: 46761790-46761795 on Build GRCh38
rs1014536554 Chr.2: 46761777-46761777 on Build GRCh38
rs62134767 Chr.2: 46761874-46761874 on Build GRCh38
rs184073268 Chr.2: 46761651-46761651 on Build GRCh38
rs564665156 Chr.2: 46761923-46761923 on Build GRCh38
rs112267674 Chr.2: 46761762-46761762 on Build GRCh38
rs943848404 Chr.2: 46761921-46761921 on Build GRCh38

View Full Product Details