Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111824803-111825314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
UniGene
Hs.436527
Gene Chromosome Location
Chr.2: 111766147-111884196 on Build GRCh38
Transcripts
NM_022662.3 XM_011511633.2 XM_011511634.2 XM_011511636.2 XM_011511637.2 XM_017004710.1 XM_017004711.1 XM_017004712.1 XM_017004713.1 XM_017004714.1 XM_017004715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3565264 c.2781T>G p.A927A Chr.2: 111824997-111824997 on Build GRCh38
6432693 c.2722G>T p.V908L Chr.2: 111825150-111825150 on Build GRCh38
1180120 c.2709C>A p.P903P Chr.2: 111825163-111825163 on Build GRCh38
1158907 c.2729A>G p.Q910R Chr.2: 111825143-111825143 on Build GRCh38
5755798 c.2795T>G p.V932G Chr.2: 111824983-111824983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759755444 Chr.2: 111825163-111825163 on Build GRCh38
rs186857819 Chr.2: 111825207-111825207 on Build GRCh38
rs765503968 Chr.2: 111825189-111825190 on Build GRCh38
rs751991270 Chr.2: 111824956-111824956 on Build GRCh38
rs780931696 Chr.2: 111825112-111825112 on Build GRCh38
rs758965429 Chr.2: 111825101-111825101 on Build GRCh38
rs947877974 Chr.2: 111824927-111824927 on Build GRCh38
rs553121206 Chr.2: 111824948-111824948 on Build GRCh38
rs190067229 Chr.2: 111825267-111825267 on Build GRCh38
rs761976439 Chr.2: 111824937-111824937 on Build GRCh38
rs773241652 Chr.2: 111825141-111825141 on Build GRCh38
rs781745176 Chr.2: 111824970-111824970 on Build GRCh38
rs370742851 Chr.2: 111825102-111825102 on Build GRCh38
rs760675254 Chr.2: 111824928-111824928 on Build GRCh38
rs576850615 Chr.2: 111824900-111824900 on Build GRCh38
rs751903394 Chr.2: 111825068-111825068 on Build GRCh38
rs528718391 Chr.2: 111825014-111825014 on Build GRCh38
rs374649331 Chr.2: 111825039-111825039 on Build GRCh38
rs778360759 Chr.2: 111825003-111825003 on Build GRCh38
rs1005662431 Chr.2: 111825243-111825243 on Build GRCh38
rs888081943 Chr.2: 111825221-111825221 on Build GRCh38
rs1039451687 Chr.2: 111825275-111825275 on Build GRCh38
rs906488871 Chr.2: 111824954-111824954 on Build GRCh38
rs776663609 Chr.2: 111825041-111825041 on Build GRCh38
rs74602614 Chr.2: 111824946-111824946 on Build GRCh38
rs775509798 Chr.2: 111824926-111824926 on Build GRCh38
rs772327654 Chr.2: 111825162-111825162 on Build GRCh38
rs373767122 Chr.2: 111825154-111825154 on Build GRCh38
rs748100620 Chr.2: 111825134-111825134 on Build GRCh38
rs755450793 Chr.2: 111824961-111824961 on Build GRCh38
rs544269253 Chr.2: 111825066-111825066 on Build GRCh38
rs374993162 Chr.2: 111824933-111824933 on Build GRCh38
rs769933324 Chr.2: 111825048-111825048 on Build GRCh38
rs372736708 Chr.2: 111825055-111825055 on Build GRCh38
rs771996120 Chr.2: 111824919-111824919 on Build GRCh38
rs758013682 Chr.2: 111825201-111825201 on Build GRCh38
rs141439178 Chr.2: 111825136-111825136 on Build GRCh38
rs200380766 Chr.2: 111825192-111825192 on Build GRCh38
rs189604622 Chr.2: 111824989-111824989 on Build GRCh38
rs763160348 Chr.2: 111825064-111825064 on Build GRCh38
rs372554847 Chr.2: 111824938-111824938 on Build GRCh38
rs746840821 Chr.2: 111825019-111825019 on Build GRCh38
rs148737841 Chr.2: 111825065-111825065 on Build GRCh38
rs750664237 Chr.2: 111824941-111824941 on Build GRCh38
rs201447717 Chr.2: 111825167-111825167 on Build GRCh38
rs60136848 Chr.2: 111825190-111825191 on Build GRCh38
rs958280582 Chr.2: 111825114-111825114 on Build GRCh38
rs750021413 Chr.2: 111825087-111825087 on Build GRCh38
rs774590349 Chr.2: 111825158-111825158 on Build GRCh38
rs778473928 Chr.2: 111825086-111825086 on Build GRCh38
rs779310506 Chr.2: 111825043-111825045 on Build GRCh38
rs373735901 Chr.2: 111825202-111825202 on Build GRCh38
rs376941023 Chr.2: 111825166-111825166 on Build GRCh38
rs116810520 Chr.2: 111825072-111825072 on Build GRCh38
rs753254626 Chr.2: 111825075-111825075 on Build GRCh38
rs557062023 Chr.2: 111824863-111824863 on Build GRCh38
rs182214701 Chr.2: 111825043-111825043 on Build GRCh38
rs533438048 Chr.2: 111825104-111825104 on Build GRCh38
rs183264328 Chr.2: 111824944-111824944 on Build GRCh38
rs779990434 Chr.2: 111825015-111825015 on Build GRCh38
rs368266743 Chr.2: 111824935-111824935 on Build GRCh38
rs745913736 Chr.2: 111825110-111825111 on Build GRCh38
rs528398939 Chr.2: 111825200-111825200 on Build GRCh38
rs781034296 Chr.2: 111825208-111825208 on Build GRCh38
rs936739452 Chr.2: 111824832-111824832 on Build GRCh38
rs370220917 Chr.2: 111824998-111824998 on Build GRCh38
rs761240970 Chr.2: 111825176-111825176 on Build GRCh38
rs759971512 Chr.2: 111825070-111825070 on Build GRCh38
rs775693507 Chr.2: 111824953-111824955 on Build GRCh38
rs756665952 Chr.2: 111825083-111825083 on Build GRCh38
rs549638086 Chr.2: 111824839-111824839 on Build GRCh38
rs551958003 Chr.2: 111825098-111825098 on Build GRCh38
rs747930519 Chr.2: 111825213-111825213 on Build GRCh38
rs373471129 Chr.2: 111825013-111825013 on Build GRCh38

View Full Product Details