Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119157767-119158291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QL2
Entrez Gene ID
165257
Gene Name
complement C1q like 2
Gene Aliases
C1QTNF10, CTRP10
UniGene
Hs.433493
Gene Chromosome Location
Chr.2: 119156243-119158895 on Build GRCh38
Transcripts
NM_182528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707737 c.302C>A p.P101H Chr.2: 119157968-119157968 on Build GRCh38
5687003 c.151C>A p.P51T Chr.2: 119158119-119158119 on Build GRCh38
84622 c.80G>C p.G27A Chr.2: 119158190-119158190 on Build GRCh38
7358338 c.134C>T p.P45L Chr.2: 119158136-119158136 on Build GRCh38
3749590 c.337C>A p.R113R Chr.2: 119157933-119157933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769474197 Chr.2: 119158183-119158183 on Build GRCh38
rs768914994 Chr.2: 119157796-119157796 on Build GRCh38
rs757745131 Chr.2: 119158062-119158062 on Build GRCh38
rs984209401 Chr.2: 119158097-119158097 on Build GRCh38
rs554293273 Chr.2: 119157983-119157983 on Build GRCh38
rs757642915 Chr.2: 119158236-119158236 on Build GRCh38
rs946660207 Chr.2: 119158071-119158071 on Build GRCh38
rs904193888 Chr.2: 119158153-119158153 on Build GRCh38
rs199528939 Chr.2: 119158131-119158131 on Build GRCh38
rs748574253 Chr.2: 119157896-119157896 on Build GRCh38
rs770629944 Chr.2: 119158207-119158207 on Build GRCh38
rs966222359 Chr.2: 119157950-119157950 on Build GRCh38
rs539572942 Chr.2: 119158068-119158068 on Build GRCh38
rs778492472 Chr.2: 119157902-119157902 on Build GRCh38
rs760809875 Chr.2: 119158226-119158226 on Build GRCh38
rs755186486 Chr.2: 119157894-119157894 on Build GRCh38
rs770773618 Chr.2: 119158261-119158261 on Build GRCh38
rs766398956 Chr.2: 119157994-119157994 on Build GRCh38
rs770271281 Chr.2: 119157901-119157901 on Build GRCh38
rs781569526 Chr.2: 119157804-119157804 on Build GRCh38
rs972798504 Chr.2: 119158078-119158078 on Build GRCh38
rs752251314 Chr.2: 119158251-119158251 on Build GRCh38
rs761571469 Chr.2: 119157865-119157865 on Build GRCh38
rs190305077 Chr.2: 119157912-119157912 on Build GRCh38
rs1007036535 Chr.2: 119158181-119158181 on Build GRCh38
rs889318406 Chr.2: 119157875-119157875 on Build GRCh38
rs764940568 Chr.2: 119157979-119157979 on Build GRCh38
rs765458110 Chr.2: 119158238-119158238 on Build GRCh38
rs895693195 Chr.2: 119157977-119157977 on Build GRCh38
rs748827179 Chr.2: 119158257-119158257 on Build GRCh38
rs752963277 Chr.2: 119158019-119158019 on Build GRCh38
rs772180390 Chr.2: 119158270-119158270 on Build GRCh38
rs766387504 Chr.2: 119158102-119158102 on Build GRCh38
rs768044358 Chr.2: 119158080-119158080 on Build GRCh38
rs775675676 Chr.2: 119158225-119158225 on Build GRCh38
rs749839428 Chr.2: 119158046-119158046 on Build GRCh38
rs927120716 Chr.2: 119157938-119157938 on Build GRCh38
rs79015645 Chr.2: 119157856-119157856 on Build GRCh38
rs1009931388 Chr.2: 119157978-119157978 on Build GRCh38
rs754297699 Chr.2: 119158147-119158147 on Build GRCh38
rs778229091 Chr.2: 119158044-119158044 on Build GRCh38
rs768401401 Chr.2: 119157928-119157928 on Build GRCh38
rs762827849 Chr.2: 119157993-119157993 on Build GRCh38
rs748694650 Chr.2: 119157811-119157811 on Build GRCh38
rs770465469 Chr.2: 119157823-119157823 on Build GRCh38
rs1043685255 Chr.2: 119158267-119158267 on Build GRCh38
rs754993161 Chr.2: 119158013-119158013 on Build GRCh38
rs772951375 Chr.2: 119158095-119158095 on Build GRCh38
rs866615224 Chr.2: 119158158-119158158 on Build GRCh38
rs775020011 Chr.2: 119157914-119157914 on Build GRCh38
rs751751172 Chr.2: 119157888-119157888 on Build GRCh38
rs374776152 Chr.2: 119158156-119158156 on Build GRCh38
rs1020198771 Chr.2: 119158206-119158206 on Build GRCh38
rs746290387 Chr.2: 119158166-119158166 on Build GRCh38
rs939211120 Chr.2: 119158026-119158026 on Build GRCh38
rs755607319 Chr.2: 119158252-119158252 on Build GRCh38
rs745852288 Chr.2: 119158264-119158264 on Build GRCh38
rs747736799 Chr.2: 119158085-119158085 on Build GRCh38
rs756358855 Chr.2: 119158143-119158143 on Build GRCh38
rs772827744 Chr.2: 119158188-119158188 on Build GRCh38
rs781238787 Chr.2: 119157895-119157895 on Build GRCh38
rs1009102754 Chr.2: 119157878-119157878 on Build GRCh38
rs764589165 Chr.2: 119158146-119158146 on Build GRCh38
rs753998926 Chr.2: 119158227-119158227 on Build GRCh38
rs760320943 Chr.2: 119157858-119157858 on Build GRCh38
rs937119741 Chr.2: 119157955-119157955 on Build GRCh38
rs750352100 Chr.2: 119157868-119157868 on Build GRCh38
rs758473715 Chr.2: 119157873-119157873 on Build GRCh38
rs564432703 Chr.2: 119158099-119158099 on Build GRCh38
rs774350154 Chr.2: 119158208-119158208 on Build GRCh38
rs867582720 Chr.2: 119158083-119158083 on Build GRCh38
rs749166655 Chr.2: 119158193-119158193 on Build GRCh38
rs769597893 Chr.2: 119158088-119158088 on Build GRCh38
rs895095336 Chr.2: 119158048-119158048 on Build GRCh38
rs945951429 Chr.2: 119158139-119158139 on Build GRCh38
rs754527078 Chr.2: 119158170-119158170 on Build GRCh38
rs1057087559 Chr.2: 119157968-119157968 on Build GRCh38
rs375510748 Chr.2: 119158255-119158255 on Build GRCh38
rs775548812 Chr.2: 119158273-119158273 on Build GRCh38
rs375216486 Chr.2: 119157832-119157832 on Build GRCh38
rs761659381 Chr.2: 119157976-119157976 on Build GRCh38
rs747828824 Chr.2: 119158182-119158182 on Build GRCh38
rs752981656 Chr.2: 119158140-119158140 on Build GRCh38
rs1050146765 Chr.2: 119158169-119158169 on Build GRCh38
rs1056970943 Chr.2: 119158110-119158110 on Build GRCh38
rs759175451 Chr.2: 119157849-119157849 on Build GRCh38
rs967751202 Chr.2: 119157991-119157991 on Build GRCh38
rs1317848 Chr.2: 119157933-119157933 on Build GRCh38
rs370204561 Chr.2: 119158018-119158018 on Build GRCh38
rs922210107 Chr.2: 119157965-119157965 on Build GRCh38
rs746583974 Chr.2: 119158079-119158079 on Build GRCh38
rs879501674 Chr.2: 119158218-119158218 on Build GRCh38
rs751699255 Chr.2: 119158003-119158003 on Build GRCh38
rs780507498 Chr.2: 119158249-119158249 on Build GRCh38
rs370760026 Chr.2: 119158222-119158222 on Build GRCh38
rs901917590 Chr.2: 119158144-119158144 on Build GRCh38
rs767638127 Chr.2: 119158133-119158133 on Build GRCh38
rs750958577 Chr.2: 119158241-119158241 on Build GRCh38
rs774144378 Chr.2: 119158114-119158114 on Build GRCh38
rs759334873 Chr.2: 119158209-119158209 on Build GRCh38
rs1044003406 Chr.2: 119158072-119158072 on Build GRCh38
rs1028665125 Chr.2: 119158001-119158001 on Build GRCh38
rs760279009 Chr.2: 119157926-119157926 on Build GRCh38
rs937168891 Chr.2: 119158101-119158101 on Build GRCh38
rs749745611 Chr.2: 119157905-119157905 on Build GRCh38
rs753631540 Chr.2: 119157862-119157862 on Build GRCh38
rs747188269 Chr.2: 119157793-119157793 on Build GRCh38
rs530953165 Chr.2: 119157850-119157850 on Build GRCh38
rs373009823 Chr.2: 119157809-119157809 on Build GRCh38
rs972360215 Chr.2: 119158215-119158215 on Build GRCh38
rs544057672 Chr.2: 119158150-119158150 on Build GRCh38
rs113019193 Chr.2: 119158174-119158174 on Build GRCh38
rs756552955 Chr.2: 119158027-119158027 on Build GRCh38
rs4643545 Chr.2: 119158228-119158228 on Build GRCh38
rs550498525 Chr.2: 119157877-119157877 on Build GRCh38
rs776265878 Chr.2: 119158084-119158084 on Build GRCh38
rs973131377 Chr.2: 119157874-119157874 on Build GRCh38
rs758639473 Chr.2: 119158054-119158056 on Build GRCh38

View Full Product Details