Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23070185-23070680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAZ
Entrez Gene ID
2781
Gene Name
G protein subunit alpha z
Gene Aliases
-
UniGene
Hs.584760
Gene Chromosome Location
Chr.22: 23070483-23125037 on Build GRCh38
Transcripts
NM_002073.2 NM_014433.2 XM_011530149.2 XM_011530150.1 XM_011530151.2 XM_011530152.2 XM_011530154.2 XM_017028774.1

Gene Symbol
RSPH14
Entrez Gene ID
27156
Gene Name
radial spoke head 14 homolog
Gene Aliases
RTDR1
UniGene
Hs.526920
Gene Chromosome Location
Chr.22: 23059408-23180048 on Build GRCh38
Transcripts
NM_002073.2 NM_014433.2 XM_011530149.2 XM_011530150.1 XM_011530151.2 XM_011530152.2 XM_011530154.2 XM_017028774.1

SNP Detail

SNP ID Chromosome Location
rs989011641 Chr.22: 23070558-23070558 on Build GRCh38
rs900490330 Chr.22: 23070260-23070261 on Build GRCh38
rs180957154 Chr.22: 23070267-23070267 on Build GRCh38
rs1055150185 Chr.22: 23070659-23070659 on Build GRCh38
rs948419139 Chr.22: 23070226-23070226 on Build GRCh38
rs554342528 Chr.22: 23070483-23070483 on Build GRCh38
rs538490171 Chr.22: 23070258-23070258 on Build GRCh38
rs559100518 Chr.22: 23070235-23070235 on Build GRCh38
rs959778748 Chr.22: 23070215-23070215 on Build GRCh38
rs888345641 Chr.22: 23070485-23070485 on Build GRCh38
rs926193259 Chr.22: 23070350-23070350 on Build GRCh38
rs959749075 Chr.22: 23070304-23070304 on Build GRCh38
rs992470700 Chr.22: 23070219-23070219 on Build GRCh38
rs1008803228 Chr.22: 23070223-23070236 on Build GRCh38
rs995283601 Chr.22: 23070262-23070262 on Build GRCh38
rs978870296 Chr.22: 23070324-23070324 on Build GRCh38
rs926238299 Chr.22: 23070236-23070236 on Build GRCh38
rs999827335 Chr.22: 23070531-23070531 on Build GRCh38
rs988916844 Chr.22: 23070367-23070367 on Build GRCh38
rs1033112259 Chr.22: 23070534-23070534 on Build GRCh38
rs900616401 Chr.22: 23070423-23070423 on Build GRCh38
rs930736859 Chr.22: 23070614-23070614 on Build GRCh38
rs549891937 Chr.22: 23070513-23070514 on Build GRCh38
rs974887925 Chr.22: 23070611-23070611 on Build GRCh38
rs1022714616 Chr.22: 23070513-23070513 on Build GRCh38
rs1022744375 Chr.22: 23070318-23070318 on Build GRCh38
rs545726926 Chr.22: 23070628-23070628 on Build GRCh38
rs944907658 Chr.22: 23070391-23070391 on Build GRCh38
rs1026894002 Chr.22: 23070265-23070265 on Build GRCh38
rs562696431 Chr.22: 23070634-23070634 on Build GRCh38
rs1036978746 Chr.22: 23070489-23070489 on Build GRCh38
rs569490501 Chr.22: 23070249-23070249 on Build GRCh38
rs912076748 Chr.22: 23070385-23070385 on Build GRCh38
rs885961 Chr.22: 23070418-23070418 on Build GRCh38
rs531520050 Chr.22: 23070654-23070654 on Build GRCh38
rs934839049 Chr.22: 23070238-23070238 on Build GRCh38
rs1038985423 Chr.22: 23070258-23070259 on Build GRCh38
rs371921422 Chr.22: 23070214-23070214 on Build GRCh38
rs915323063 Chr.22: 23070567-23070567 on Build GRCh38
rs979019625 Chr.22: 23070230-23070230 on Build GRCh38
rs969748170 Chr.22: 23070319-23070320 on Build GRCh38
rs966172118 Chr.22: 23070604-23070604 on Build GRCh38
rs952571921 Chr.22: 23070210-23070210 on Build GRCh38
rs1040635478 Chr.22: 23070241-23070258 on Build GRCh38
rs530661111 Chr.22: 23070211-23070211 on Build GRCh38
rs577246591 Chr.22: 23070519-23070519 on Build GRCh38
rs922006893 Chr.22: 23070612-23070612 on Build GRCh38
rs1004145574 Chr.22: 23070487-23070487 on Build GRCh38
rs895633319 Chr.22: 23070491-23070491 on Build GRCh38
rs890503408 Chr.22: 23070246-23070246 on Build GRCh38
rs930625816 Chr.22: 23070437-23070437 on Build GRCh38
rs574856830 Chr.22: 23070310-23070310 on Build GRCh38
rs956290119 Chr.22: 23070538-23070538 on Build GRCh38
rs1004008936 Chr.22: 23070266-23070266 on Build GRCh38
rs554053759 Chr.22: 23070519-23070520 on Build GRCh38
rs201366455 Chr.22: 23070600-23070601 on Build GRCh38
rs912182851 Chr.22: 23070601-23070601 on Build GRCh38
rs970221188 Chr.22: 23070523-23070524 on Build GRCh38
rs1014136390 Chr.22: 23070507-23070507 on Build GRCh38

View Full Product Details