Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197510793-197511287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
UniGene
Hs.274539
Gene Chromosome Location
Chr.3: 197509783-197573323 on Build GRCh38
Transcripts
NM_004051.4 NM_203314.2 NM_203315.2 XM_005269352.3 XM_005269355.4 XM_011513067.2 XM_017007007.1 XM_017007008.1 XM_017007009.1 XM_017007010.1 XM_017007011.1 XM_017007012.1 XM_017007014.1 XM_017007015.1

SNP Detail

SNP ID Chromosome Location
rs969724802 Chr.3: 197511042-197511042 on Build GRCh38
rs915557370 Chr.3: 197511025-197511025 on Build GRCh38
rs550112385 Chr.3: 197511100-197511100 on Build GRCh38
rs918698338 Chr.3: 197511171-197511171 on Build GRCh38
rs780020082 Chr.3: 197511054-197511054 on Build GRCh38
rs548816144 Chr.3: 197511203-197511203 on Build GRCh38
rs113076498 Chr.3: 197511126-197511126 on Build GRCh38
rs955850250 Chr.3: 197511131-197511131 on Build GRCh38
rs896421857 Chr.3: 197510935-197510935 on Build GRCh38
rs147847833 Chr.3: 197511158-197511158 on Build GRCh38
rs939994691 Chr.3: 197511092-197511092 on Build GRCh38
rs999426962 Chr.3: 197510976-197510976 on Build GRCh38
rs929947510 Chr.3: 197511199-197511199 on Build GRCh38
rs896006182 Chr.3: 197511261-197511261 on Build GRCh38
rs985731137 Chr.3: 197511137-197511137 on Build GRCh38
rs959743060 Chr.3: 197511016-197511016 on Build GRCh38
rs1007953083 Chr.3: 197511164-197511164 on Build GRCh38
rs983832940 Chr.3: 197511070-197511070 on Build GRCh38
rs141498037 Chr.3: 197510829-197510829 on Build GRCh38
rs530372855 Chr.3: 197511195-197511195 on Build GRCh38
rs1031876804 Chr.3: 197511003-197511003 on Build GRCh38
rs574685777 Chr.3: 197510910-197510910 on Build GRCh38
rs1007779051 Chr.3: 197510833-197510833 on Build GRCh38
rs1044996844 Chr.3: 197511262-197511262 on Build GRCh38
rs1043101016 Chr.3: 197511169-197511169 on Build GRCh38
rs903688830 Chr.3: 197511045-197511045 on Build GRCh38
rs1056377097 Chr.3: 197510897-197510898 on Build GRCh38
rs947927054 Chr.3: 197511250-197511250 on Build GRCh38
rs148912258 Chr.3: 197511210-197511210 on Build GRCh38
rs184062475 Chr.3: 197511103-197511103 on Build GRCh38
rs911343693 Chr.3: 197511153-197511153 on Build GRCh38
rs1022279230 Chr.3: 197510908-197510908 on Build GRCh38
rs111948864 Chr.3: 197511024-197511024 on Build GRCh38
rs918560819 Chr.3: 197510893-197510893 on Build GRCh38
rs1037265853 Chr.3: 197511096-197511096 on Build GRCh38
rs759519171 Chr.3: 197511063-197511063 on Build GRCh38
rs528449001 Chr.3: 197511055-197511055 on Build GRCh38
rs191824972 Chr.3: 197511010-197511010 on Build GRCh38
rs929976883 Chr.3: 197510894-197510894 on Build GRCh38
rs555925977 Chr.3: 197510835-197510835 on Build GRCh38
rs556904258 Chr.3: 197511085-197511085 on Build GRCh38
rs917838002 Chr.3: 197511213-197511213 on Build GRCh38
rs576776205 Chr.3: 197511228-197511228 on Build GRCh38
rs1012072962 Chr.3: 197510953-197510953 on Build GRCh38
rs1051809069 Chr.3: 197511142-197511142 on Build GRCh38
rs1038469955 Chr.3: 197510860-197510860 on Build GRCh38
rs999123672 Chr.3: 197511206-197511206 on Build GRCh38
rs904765026 Chr.3: 197511196-197511196 on Build GRCh38
rs969021099 Chr.3: 197510921-197510921 on Build GRCh38
rs1000537638 Chr.3: 197511046-197511046 on Build GRCh38

View Full Product Details