Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176388153-176388651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL10
Entrez Gene ID
285598
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
UniGene
Hs.424552
Gene Chromosome Location
Chr.5: 176365468-176415085 on Build GRCh38
Transcripts
XM_011534529.2 XM_011534530.2 NM_001256539.3 NM_001256540.3 NM_001291305.2 NM_001291307.2 NM_001291308.2 NM_001317975.1 NM_016391.7 NR_134316.1 NR_134318.1 XM_011534567.1

Gene Symbol
NOP16
Entrez Gene ID
51491
Gene Name
NOP16 nucleolar protein
Gene Aliases
HSPC111, HSPC185
UniGene
Hs.696283
Gene Chromosome Location
Chr.5: 176383937-176388927 on Build GRCh38
Transcripts
XM_011534529.2 XM_011534530.2 NM_001256539.3 NM_001256540.3 NM_001291305.2 NM_001291307.2 NM_001291308.2 NM_001317975.1 NM_016391.7 NR_134316.1 NR_134318.1 XM_011534567.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5565184 c.71G>T p.R24L Chr.5: 176388469-176388469 on Build GRCh38
3340327 c.35A>T p.K12M Chr.5: 176388505-176388505 on Build GRCh38
3373802 c.194C>T p.A65V Chr.5: 176388257-176388257 on Build GRCh38
7285712 c.14_15insA p.K7fs*41 Chr.5: 176388525-176388526 on Build GRCh38
737654 c.96G>A p.P32P Chr.5: 176388444-176388444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377455146 Chr.5: 176388516-176388516 on Build GRCh38
rs767669311 Chr.5: 176388212-176388212 on Build GRCh38
rs777675697 Chr.5: 176388424-176388424 on Build GRCh38
rs917097779 Chr.5: 176388259-176388259 on Build GRCh38
rs766742049 Chr.5: 176388594-176388594 on Build GRCh38
rs768780614 Chr.5: 176388275-176388275 on Build GRCh38
rs369929689 Chr.5: 176388231-176388231 on Build GRCh38
rs762362409 Chr.5: 176388187-176388187 on Build GRCh38
rs764643906 Chr.5: 176388357-176388357 on Build GRCh38
rs781235569 Chr.5: 176388588-176388588 on Build GRCh38
rs998458269 Chr.5: 176388366-176388366 on Build GRCh38
rs370995699 Chr.5: 176388468-176388468 on Build GRCh38
rs899794019 Chr.5: 176388356-176388356 on Build GRCh38
rs753723748 Chr.5: 176388575-176388575 on Build GRCh38
rs982616743 Chr.5: 176388617-176388617 on Build GRCh38
rs888548631 Chr.5: 176388304-176388304 on Build GRCh38
rs527818464 Chr.5: 176388560-176388560 on Build GRCh38
rs754010236 Chr.5: 176388622-176388622 on Build GRCh38
rs751258265 Chr.5: 176388488-176388488 on Build GRCh38
rs769713350 Chr.5: 176388305-176388305 on Build GRCh38
rs765915681 Chr.5: 176388199-176388201 on Build GRCh38
rs376729124 Chr.5: 176388414-176388414 on Build GRCh38
rs376625530 Chr.5: 176388257-176388257 on Build GRCh38
rs369903036 Chr.5: 176388260-176388260 on Build GRCh38
rs751172532 Chr.5: 176388411-176388411 on Build GRCh38
rs745451083 Chr.5: 176388174-176388174 on Build GRCh38
rs748124528 Chr.5: 176388415-176388415 on Build GRCh38
rs767155534 Chr.5: 176388510-176388510 on Build GRCh38
rs761435181 Chr.5: 176388308-176388308 on Build GRCh38
rs900895433 Chr.5: 176388196-176388196 on Build GRCh38
rs929780920 Chr.5: 176388289-176388289 on Build GRCh38
rs369167277 Chr.5: 176388322-176388322 on Build GRCh38
rs185134742 Chr.5: 176388217-176388217 on Build GRCh38
rs778703816 Chr.5: 176388580-176388580 on Build GRCh38
rs879207987 Chr.5: 176388598-176388598 on Build GRCh38
rs762354832 Chr.5: 176388461-176388461 on Build GRCh38
rs752662570 Chr.5: 176388525-176388525 on Build GRCh38
rs201019782 Chr.5: 176388299-176388299 on Build GRCh38
rs772025977 Chr.5: 176388321-176388321 on Build GRCh38
rs892709775 Chr.5: 176388372-176388372 on Build GRCh38
rs754267917 Chr.5: 176388240-176388240 on Build GRCh38
rs769429960 Chr.5: 176388419-176388419 on Build GRCh38
rs1026225217 Chr.5: 176388504-176388504 on Build GRCh38
rs759828112 Chr.5: 176388316-176388316 on Build GRCh38
rs768547595 Chr.5: 176388192-176388192 on Build GRCh38
rs775419543 Chr.5: 176388442-176388442 on Build GRCh38
rs754357508 Chr.5: 176388367-176388367 on Build GRCh38
rs777430631 Chr.5: 176388530-176388530 on Build GRCh38
rs975052787 Chr.5: 176388574-176388574 on Build GRCh38
rs766762264 Chr.5: 176388201-176388201 on Build GRCh38
rs575715231 Chr.5: 176388573-176388573 on Build GRCh38
rs747199026 Chr.5: 176388581-176388581 on Build GRCh38
rs149911333 Chr.5: 176388179-176388179 on Build GRCh38
rs190729915 Chr.5: 176388577-176388577 on Build GRCh38
rs201660459 Chr.5: 176388490-176388490 on Build GRCh38
rs758108765 Chr.5: 176388381-176388381 on Build GRCh38
rs557670787 Chr.5: 176388551-176388551 on Build GRCh38
rs144937482 Chr.5: 176388584-176388584 on Build GRCh38
rs374791604 Chr.5: 176388534-176388534 on Build GRCh38
rs374285141 Chr.5: 176388470-176388470 on Build GRCh38
rs761170714 Chr.5: 176388347-176388347 on Build GRCh38
rs749169901 Chr.5: 176388531-176388531 on Build GRCh38
rs17832192 Chr.5: 176388214-176388214 on Build GRCh38
rs774094666 Chr.5: 176388554-176388554 on Build GRCh38
rs759465541 Chr.5: 176388210-176388210 on Build GRCh38
rs372260739 Chr.5: 176388245-176388245 on Build GRCh38
rs775642185 Chr.5: 176388428-176388428 on Build GRCh38
rs768676655 Chr.5: 176388535-176388535 on Build GRCh38
rs768728192 Chr.5: 176388445-176388445 on Build GRCh38
rs750097354 Chr.5: 176388249-176388249 on Build GRCh38
rs750020555 Chr.5: 176388373-176388373 on Build GRCh38
rs776608916 Chr.5: 176388197-176388197 on Build GRCh38
rs564912122 Chr.5: 176388602-176388602 on Build GRCh38
rs35906642 Chr.5: 176388333-176388333 on Build GRCh38
rs937262782 Chr.5: 176388227-176388227 on Build GRCh38
rs779424803 Chr.5: 176388388-176388388 on Build GRCh38
rs777084499 Chr.5: 176388456-176388456 on Build GRCh38
rs371311461 Chr.5: 176388523-176388523 on Build GRCh38
rs372982357 Chr.5: 176388369-176388369 on Build GRCh38
rs765994335 Chr.5: 176388487-176388487 on Build GRCh38
rs756120217 Chr.5: 176388527-176388527 on Build GRCh38
rs951280748 Chr.5: 176388612-176388612 on Build GRCh38
rs115822936 Chr.5: 176388188-176388188 on Build GRCh38
rs141048893 Chr.5: 176388590-176388590 on Build GRCh38
rs755995657 Chr.5: 176388571-176388571 on Build GRCh38
rs767108492 Chr.5: 176388556-176388556 on Build GRCh38
rs367884738 Chr.5: 176388186-176388186 on Build GRCh38
rs760690134 Chr.5: 176388443-176388443 on Build GRCh38
rs774642064 Chr.5: 176388209-176388209 on Build GRCh38
rs748648726 Chr.5: 176388548-176388548 on Build GRCh38
rs372435052 Chr.5: 176388292-176388292 on Build GRCh38
rs776730232 Chr.5: 176388542-176388542 on Build GRCh38
rs746987248 Chr.5: 176388189-176388189 on Build GRCh38
rs892294018 Chr.5: 176388595-176388595 on Build GRCh38
rs763306414 Chr.5: 176388552-176388552 on Build GRCh38
rs752325489 Chr.5: 176388558-176388558 on Build GRCh38
rs772408909 Chr.5: 176388435-176388435 on Build GRCh38
rs753343413 Chr.5: 176388577-176388578 on Build GRCh38
rs941448956 Chr.5: 176388307-176388307 on Build GRCh38
rs756891138 Chr.5: 176388244-176388244 on Build GRCh38
rs766681494 Chr.5: 176388553-176388553 on Build GRCh38
rs767899190 Chr.5: 176388382-176388382 on Build GRCh38
rs540374183 Chr.5: 176388591-176388591 on Build GRCh38
rs200947536 Chr.5: 176388315-176388315 on Build GRCh38

View Full Product Details