Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30702662-30703172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MDC1
Entrez Gene ID
9656
Gene Name
mediator of DNA damage checkpoint 1
Gene Aliases
NFBD1
UniGene
Hs.653495
Gene Chromosome Location
Chr.6: 30699807-30717988 on Build GRCh38
Transcripts
NM_014641.2 XM_005249492.2 XM_005249493.3 XM_005249494.4 XM_005249497.3 XM_005249498.3 XM_011515001.2 XM_011515003.2 XM_011515004.2 XM_017011519.1 XM_017011520.1 XM_017011521.1 XM_017011522.1 NR_133647.1

Gene Symbol
MDC1-AS1
Entrez Gene ID
106478956
Gene Name
MDC1 antisense RNA 1
Gene Aliases
MDC1-AS
UniGene
Hs.708954
Gene Chromosome Location
Chr.6: 30703067-30713184 on Build GRCh38
Transcripts
NM_014641.2 XM_005249492.2 XM_005249493.3 XM_005249494.4 XM_005249497.3 XM_005249498.3 XM_011515001.2 XM_011515003.2 XM_011515004.2 XM_017011519.1 XM_017011520.1 XM_017011521.1 XM_017011522.1 NR_133647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
126799 c.5899G>T p.E1967* Chr.6: 30702844-30702844 on Build GRCh38
6710178 c.5953G>T p.D1985Y Chr.6: 30702790-30702790 on Build GRCh38
3861489 c.5798G>A p.R1933Q Chr.6: 30703171-30703171 on Build GRCh38
4701258 c.5955C>T p.D1985D Chr.6: 30702788-30702788 on Build GRCh38
6930754 c.5828G>C p.R1943P Chr.6: 30703141-30703141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149766760 Chr.6: 30703004-30703005 on Build GRCh38
rs191103790 Chr.6: 30702761-30702761 on Build GRCh38
rs148600920 Chr.6: 30703141-30703141 on Build GRCh38
rs147519338 Chr.6: 30702703-30702703 on Build GRCh38
rs767992636 Chr.6: 30703072-30703072 on Build GRCh38
rs746767199 Chr.6: 30702773-30702773 on Build GRCh38
rs774692071 Chr.6: 30703048-30703048 on Build GRCh38
rs957162456 Chr.6: 30702806-30702806 on Build GRCh38
rs114325637 Chr.6: 30702885-30702885 on Build GRCh38
rs1030301166 Chr.6: 30702784-30702784 on Build GRCh38
rs753325641 Chr.6: 30702688-30702688 on Build GRCh38
rs1032877192 Chr.6: 30702743-30702743 on Build GRCh38
rs779991732 Chr.6: 30703148-30703148 on Build GRCh38
rs372124226 Chr.6: 30702780-30702780 on Build GRCh38
rs960543389 Chr.6: 30702699-30702699 on Build GRCh38
rs149494987 Chr.6: 30702788-30702788 on Build GRCh38
rs755294233 Chr.6: 30702856-30702856 on Build GRCh38
rs756187041 Chr.6: 30703071-30703071 on Build GRCh38
rs757808811 Chr.6: 30702734-30702734 on Build GRCh38
rs766769094 Chr.6: 30703078-30703078 on Build GRCh38
rs116085079 Chr.6: 30703065-30703065 on Build GRCh38
rs774968134 Chr.6: 30703135-30703135 on Build GRCh38
rs751175105 Chr.6: 30702822-30702822 on Build GRCh38
rs902674659 Chr.6: 30703076-30703076 on Build GRCh38
rs759330464 Chr.6: 30702782-30702782 on Build GRCh38
rs756578505 Chr.6: 30702848-30702848 on Build GRCh38
rs762416337 Chr.6: 30703053-30703053 on Build GRCh38
rs758544306 Chr.6: 30703117-30703117 on Build GRCh38
rs749596006 Chr.6: 30702759-30702759 on Build GRCh38
rs781739100 Chr.6: 30702735-30702735 on Build GRCh38
rs367854484 Chr.6: 30702710-30702710 on Build GRCh38
rs771489509 Chr.6: 30703013-30703013 on Build GRCh38
rs759114870 Chr.6: 30702690-30702690 on Build GRCh38
rs757978653 Chr.6: 30702709-30702709 on Build GRCh38
rs748328915 Chr.6: 30702873-30702873 on Build GRCh38
rs770806736 Chr.6: 30702774-30702774 on Build GRCh38
rs371773610 Chr.6: 30703083-30703083 on Build GRCh38
rs745884792 Chr.6: 30702906-30702906 on Build GRCh38
rs764807291 Chr.6: 30702787-30702787 on Build GRCh38
rs780850494 Chr.6: 30702709-30702709 on Build GRCh38
rs769604487 Chr.6: 30702912-30702912 on Build GRCh38
rs936978976 Chr.6: 30702938-30702938 on Build GRCh38
rs779306353 Chr.6: 30702872-30702872 on Build GRCh38
rs781097847 Chr.6: 30703135-30703135 on Build GRCh38
rs994463741 Chr.6: 30702983-30702983 on Build GRCh38
rs560973662 Chr.6: 30702771-30702771 on Build GRCh38
rs565255371 Chr.6: 30703018-30703018 on Build GRCh38
rs756232485 Chr.6: 30702746-30702746 on Build GRCh38
rs916938881 Chr.6: 30703021-30703021 on Build GRCh38
rs979503750 Chr.6: 30702779-30702779 on Build GRCh38
rs996233394 Chr.6: 30703110-30703110 on Build GRCh38
rs768990928 Chr.6: 30702765-30702765 on Build GRCh38
rs769808635 Chr.6: 30703142-30703142 on Build GRCh38
rs769554361 Chr.6: 30702867-30702871 on Build GRCh38
rs749360038 Chr.6: 30702855-30702855 on Build GRCh38
rs560042770 Chr.6: 30702847-30702847 on Build GRCh38
rs745510219 Chr.6: 30702732-30702733 on Build GRCh38
rs368917248 Chr.6: 30702722-30702722 on Build GRCh38
rs909287967 Chr.6: 30702915-30702915 on Build GRCh38
rs763711633 Chr.6: 30702811-30702811 on Build GRCh38
rs755128553 Chr.6: 30703097-30703097 on Build GRCh38
rs780241407 Chr.6: 30702853-30702853 on Build GRCh38
rs983775258 Chr.6: 30702797-30702797 on Build GRCh38
rs773783519 Chr.6: 30702886-30702886 on Build GRCh38
rs2239886 Chr.6: 30702923-30702923 on Build GRCh38
rs376959883 Chr.6: 30703075-30703075 on Build GRCh38
rs367669411 Chr.6: 30703077-30703077 on Build GRCh38
rs779075020 Chr.6: 30703113-30703113 on Build GRCh38
rs897856130 Chr.6: 30702962-30702962 on Build GRCh38
rs371162703 Chr.6: 30702740-30702740 on Build GRCh38
rs140061594 Chr.6: 30702830-30702830 on Build GRCh38
rs558377335 Chr.6: 30702731-30702731 on Build GRCh38
rs774007940 Chr.6: 30703069-30703069 on Build GRCh38
rs762650225 Chr.6: 30702803-30702803 on Build GRCh38
rs779932059 Chr.6: 30702758-30702758 on Build GRCh38
rs747727839 Chr.6: 30702851-30702851 on Build GRCh38
rs767716856 Chr.6: 30702844-30702844 on Build GRCh38
rs752967444 Chr.6: 30703115-30703115 on Build GRCh38
rs377200740 Chr.6: 30703101-30703101 on Build GRCh38
rs770859881 Chr.6: 30702874-30702874 on Build GRCh38
rs1036753067 Chr.6: 30702937-30702937 on Build GRCh38
rs28994877 Chr.6: 30702842-30702842 on Build GRCh38

View Full Product Details