Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4803610-4804126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RADIL
Entrez Gene ID
55698
Gene Name
Rap associating with DIL domain
Gene Aliases
RASIP2
UniGene
Hs.667336
Gene Chromosome Location
Chr.7: 4799109-4883704 on Build GRCh38
Transcripts
NM_018059.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1622926 c.2331C>A p.V777V Chr.7: 4803714-4803714 on Build GRCh38
94324 c.2410C>T p.R804C Chr.7: 4803635-4803635 on Build GRCh38
42970 c.2369A>G p.N790S Chr.7: 4803676-4803676 on Build GRCh38
7266752 c.2329G>A p.V777I Chr.7: 4803716-4803716 on Build GRCh38
6378591 c.2384G>A p.S795N Chr.7: 4803661-4803661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746153607 Chr.7: 4803759-4803759 on Build GRCh38
rs768874838 Chr.7: 4803692-4803692 on Build GRCh38
rs1034059709 Chr.7: 4803642-4803642 on Build GRCh38
rs953606081 Chr.7: 4803828-4803828 on Build GRCh38
rs79438451 Chr.7: 4803950-4803950 on Build GRCh38
rs202242127 Chr.7: 4803704-4803704 on Build GRCh38
rs779691614 Chr.7: 4803702-4803702 on Build GRCh38
rs754205827 Chr.7: 4803911-4803911 on Build GRCh38
rs1007682303 Chr.7: 4804079-4804079 on Build GRCh38
rs770990661 Chr.7: 4803677-4803677 on Build GRCh38
rs986550749 Chr.7: 4803658-4803658 on Build GRCh38
rs777640931 Chr.7: 4803674-4803674 on Build GRCh38
rs578109505 Chr.7: 4803631-4803631 on Build GRCh38
rs145060414 Chr.7: 4803674-4803675 on Build GRCh38
rs377238550 Chr.7: 4803711-4803711 on Build GRCh38
rs926702230 Chr.7: 4803993-4803993 on Build GRCh38
rs776753560 Chr.7: 4803680-4803680 on Build GRCh38
rs747687776 Chr.7: 4804030-4804030 on Build GRCh38
rs760050745 Chr.7: 4803877-4803877 on Build GRCh38
rs915406991 Chr.7: 4804005-4804005 on Build GRCh38
rs989640359 Chr.7: 4804001-4804001 on Build GRCh38
rs750784730 Chr.7: 4803716-4803716 on Build GRCh38
rs754367652 Chr.7: 4803726-4803726 on Build GRCh38
rs549679178 Chr.7: 4803752-4803752 on Build GRCh38
rs868458528 Chr.7: 4803713-4803713 on Build GRCh38
rs532822283 Chr.7: 4803789-4803789 on Build GRCh38
rs766850553 Chr.7: 4803724-4803724 on Build GRCh38
rs778490995 Chr.7: 4803654-4803654 on Build GRCh38
rs1053840697 Chr.7: 4803844-4803844 on Build GRCh38
rs899589095 Chr.7: 4804063-4804063 on Build GRCh38
rs912118823 Chr.7: 4804035-4804035 on Build GRCh38
rs749636481 Chr.7: 4803852-4803853 on Build GRCh38
rs543235806 Chr.7: 4803689-4803689 on Build GRCh38
rs147281855 Chr.7: 4803953-4803953 on Build GRCh38
rs549202573 Chr.7: 4803906-4803906 on Build GRCh38
rs774392101 Chr.7: 4803702-4803702 on Build GRCh38
rs762334523 Chr.7: 4803948-4803948 on Build GRCh38
rs892337803 Chr.7: 4803696-4803696 on Build GRCh38
rs1030919081 Chr.7: 4804040-4804040 on Build GRCh38
rs949537251 Chr.7: 4803818-4803818 on Build GRCh38
rs576308664 Chr.7: 4804000-4804000 on Build GRCh38
rs754533817 Chr.7: 4803648-4803648 on Build GRCh38
rs925281476 Chr.7: 4803855-4803855 on Build GRCh38
rs866087736 Chr.7: 4803693-4803693 on Build GRCh38
rs528797277 Chr.7: 4803736-4803736 on Build GRCh38
rs118072475 Chr.7: 4804031-4804031 on Build GRCh38
rs982456254 Chr.7: 4803649-4803649 on Build GRCh38
rs373127909 Chr.7: 4803705-4803705 on Build GRCh38
rs897346031 Chr.7: 4803713-4803727 on Build GRCh38
rs367638850 Chr.7: 4803914-4803914 on Build GRCh38
rs1019465861 Chr.7: 4803782-4803783 on Build GRCh38
rs749561751 Chr.7: 4803872-4803872 on Build GRCh38
rs546102417 Chr.7: 4803951-4803951 on Build GRCh38
rs114077859 Chr.7: 4803806-4803806 on Build GRCh38
rs923248280 Chr.7: 4803638-4803638 on Build GRCh38
rs73312604 Chr.7: 4804046-4804046 on Build GRCh38
rs926697203 Chr.7: 4803823-4803823 on Build GRCh38
rs752237829 Chr.7: 4803735-4803735 on Build GRCh38
rs945655743 Chr.7: 4804014-4804014 on Build GRCh38
rs754445414 Chr.7: 4803729-4803729 on Build GRCh38
rs183365967 Chr.7: 4803815-4803815 on Build GRCh38
rs938303458 Chr.7: 4803724-4803724 on Build GRCh38
rs375363910 Chr.7: 4803989-4803989 on Build GRCh38
rs534919342 Chr.7: 4803915-4803915 on Build GRCh38
rs1011197678 Chr.7: 4803757-4803757 on Build GRCh38
rs913790003 Chr.7: 4804106-4804106 on Build GRCh38
rs764438311 Chr.7: 4803876-4803876 on Build GRCh38
rs149092116 Chr.7: 4804104-4804104 on Build GRCh38
rs975246439 Chr.7: 4803796-4803796 on Build GRCh38
rs187128776 Chr.7: 4803676-4803676 on Build GRCh38
rs988094576 Chr.7: 4803874-4803874 on Build GRCh38
rs765724494 Chr.7: 4803885-4803885 on Build GRCh38
rs766762839 Chr.7: 4803849-4803849 on Build GRCh38
rs986109842 Chr.7: 4803833-4803833 on Build GRCh38
rs540532070 Chr.7: 4804080-4804080 on Build GRCh38
rs773345389 Chr.7: 4803827-4803827 on Build GRCh38
rs879036204 Chr.7: 4803859-4803859 on Build GRCh38
rs757889879 Chr.7: 4803899-4803899 on Build GRCh38
rs117176565 Chr.7: 4803970-4803970 on Build GRCh38
rs545184177 Chr.7: 4803635-4803635 on Build GRCh38
rs376212483 Chr.7: 4804041-4804041 on Build GRCh38
rs982587946 Chr.7: 4803820-4803820 on Build GRCh38
rs892790107 Chr.7: 4803645-4803645 on Build GRCh38
rs780595219 Chr.7: 4803639-4803639 on Build GRCh38
rs752250560 Chr.7: 4803889-4803889 on Build GRCh38
rs376419252 Chr.7: 4803701-4803701 on Build GRCh38
rs574917682 Chr.7: 4803663-4803663 on Build GRCh38
rs117455509 Chr.7: 4803912-4803912 on Build GRCh38
rs778399445 Chr.7: 4803734-4803734 on Build GRCh38
rs769071529 Chr.7: 4803925-4803925 on Build GRCh38
rs898086227 Chr.7: 4804027-4804027 on Build GRCh38
rs997087173 Chr.7: 4803920-4803920 on Build GRCh38
rs753341166 Chr.7: 4803644-4803644 on Build GRCh38
rs1019872746 Chr.7: 4803758-4803758 on Build GRCh38
rs774813025 Chr.7: 4803932-4803932 on Build GRCh38
rs575729810 Chr.7: 4803971-4803971 on Build GRCh38
rs761856588 Chr.7: 4803814-4803814 on Build GRCh38
rs958047816 Chr.7: 4804103-4804103 on Build GRCh38
rs866040132 Chr.7: 4803634-4803634 on Build GRCh38
rs191399881 Chr.7: 4803777-4803777 on Build GRCh38
rs561405604 Chr.7: 4803717-4803717 on Build GRCh38
rs373542421 Chr.7: 4803666-4803666 on Build GRCh38
rs777358624 Chr.7: 4803746-4803746 on Build GRCh38
rs986659242 Chr.7: 4803673-4803673 on Build GRCh38
rs771698856 Chr.7: 4803672-4803672 on Build GRCh38
rs558923954 Chr.7: 4803781-4803781 on Build GRCh38
rs967059348 Chr.7: 4803671-4803671 on Build GRCh38
rs569802479 Chr.7: 4803807-4803807 on Build GRCh38
rs913717702 Chr.7: 4803901-4803901 on Build GRCh38
rs959448725 Chr.7: 4803994-4803994 on Build GRCh38
rs745740377 Chr.7: 4803958-4803958 on Build GRCh38
rs760980686 Chr.7: 4803841-4803841 on Build GRCh38
rs915498848 Chr.7: 4803853-4803853 on Build GRCh38
rs867779068 Chr.7: 4803665-4803665 on Build GRCh38
rs376449324 Chr.7: 4803959-4803959 on Build GRCh38
rs745827941 Chr.7: 4803800-4803800 on Build GRCh38
rs957932537 Chr.7: 4803864-4803864 on Build GRCh38

View Full Product Details