Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76327898-76328402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAG
Entrez Gene ID
7532
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma
Gene Aliases
14-3-3GAMMA, PPP1R170
UniGene
Hs.744840
Gene Chromosome Location
Chr.7: 76326791-76359025 on Build GRCh38
Transcripts
NM_012479.3

SNP Detail

SNP ID Chromosome Location
rs1001798872 Chr.7: 76328224-76328224 on Build GRCh38
rs766346896 Chr.7: 76328179-76328179 on Build GRCh38
rs1024541799 Chr.7: 76328201-76328201 on Build GRCh38
rs966333021 Chr.7: 76328347-76328347 on Build GRCh38
rs113115178 Chr.7: 76328291-76328291 on Build GRCh38
rs944543947 Chr.7: 76328115-76328115 on Build GRCh38
rs773189945 Chr.7: 76328061-76328061 on Build GRCh38
rs924537685 Chr.7: 76328122-76328122 on Build GRCh38
rs1046305 Chr.7: 76328282-76328282 on Build GRCh38
rs76772010 Chr.7: 76328295-76328295 on Build GRCh38
rs752769030 Chr.7: 76328210-76328210 on Build GRCh38
rs1027809041 Chr.7: 76328032-76328035 on Build GRCh38
rs980289148 Chr.7: 76328003-76328003 on Build GRCh38
rs1025011690 Chr.7: 76327942-76327942 on Build GRCh38
rs948510552 Chr.7: 76328232-76328232 on Build GRCh38
rs543267868 Chr.7: 76328017-76328017 on Build GRCh38
rs35854099 Chr.7: 76328208-76328209 on Build GRCh38
rs995081512 Chr.7: 76328335-76328335 on Build GRCh38
rs995427409 Chr.7: 76328032-76328034 on Build GRCh38
rs202098973 Chr.7: 76328243-76328243 on Build GRCh38
rs921735299 Chr.7: 76328373-76328373 on Build GRCh38
rs1034492068 Chr.7: 76328236-76328236 on Build GRCh38
rs900362979 Chr.7: 76328147-76328147 on Build GRCh38
rs1038815838 Chr.7: 76328123-76328123 on Build GRCh38
rs971586758 Chr.7: 76327987-76327987 on Build GRCh38
rs549613781 Chr.7: 76328258-76328258 on Build GRCh38
rs913194838 Chr.7: 76328060-76328060 on Build GRCh38
rs758577390 Chr.7: 76328229-76328229 on Build GRCh38
rs1046304 Chr.7: 76328327-76328327 on Build GRCh38
rs571539098 Chr.7: 76328354-76328354 on Build GRCh38
rs755108209 Chr.7: 76328349-76328350 on Build GRCh38
rs1048805708 Chr.7: 76328169-76328169 on Build GRCh38
rs967389277 Chr.7: 76328296-76328296 on Build GRCh38
rs990197499 Chr.7: 76328255-76328255 on Build GRCh38
rs529662682 Chr.7: 76328188-76328188 on Build GRCh38
rs1054721673 Chr.7: 76328187-76328187 on Build GRCh38
rs866088254 Chr.7: 76328355-76328355 on Build GRCh38
rs1004995252 Chr.7: 76328195-76328195 on Build GRCh38
rs554569680 Chr.7: 76327977-76327977 on Build GRCh38
rs1005101821 Chr.7: 76327930-76327930 on Build GRCh38
rs930356978 Chr.7: 76328166-76328166 on Build GRCh38
rs780397500 Chr.7: 76328028-76328031 on Build GRCh38
rs576145850 Chr.7: 76328000-76328000 on Build GRCh38
rs1003801483 Chr.7: 76327945-76327945 on Build GRCh38
rs776720587 Chr.7: 76328282-76328283 on Build GRCh38
rs906923211 Chr.7: 76328273-76328273 on Build GRCh38
rs559388991 Chr.7: 76328076-76328076 on Build GRCh38
rs75285465 Chr.7: 76328065-76328065 on Build GRCh38
rs935922211 Chr.7: 76328138-76328138 on Build GRCh38
rs376156449 Chr.7: 76328097-76328097 on Build GRCh38
rs978034128 Chr.7: 76328091-76328091 on Build GRCh38
rs912579030 Chr.7: 76328337-76328337 on Build GRCh38
rs977397920 Chr.7: 76328086-76328087 on Build GRCh38
rs201126549 Chr.7: 76328319-76328319 on Build GRCh38
rs191562756 Chr.7: 76327959-76327959 on Build GRCh38
rs565105497 Chr.7: 76328032-76328032 on Build GRCh38
rs1045465011 Chr.7: 76328236-76328236 on Build GRCh38
rs749725542 Chr.7: 76328336-76328336 on Build GRCh38
rs962297119 Chr.7: 76328024-76328027 on Build GRCh38
rs528457722 Chr.7: 76328059-76328059 on Build GRCh38
rs1028275057 Chr.7: 76328343-76328343 on Build GRCh38

View Full Product Details