Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 76329894-76330400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAG
Entrez Gene ID
7532
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma
Gene Aliases
14-3-3GAMMA, PPP1R170
UniGene
Hs.744840
Gene Chromosome Location
Chr.7: 76326791-76359025 on Build GRCh38
Transcripts
NM_012479.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3640643 c.427A>G p.R143G Chr.7: 76329894-76329894 on Build GRCh38
6228440 c.167G>A p.R56H Chr.7: 76330154-76330154 on Build GRCh38
3374675 c.418G>T p.G140* Chr.7: 76329903-76329903 on Build GRCh38
5997202 c.310C>T p.L104L Chr.7: 76330011-76330011 on Build GRCh38
5615869 c.133C>A p.L45M Chr.7: 76330188-76330188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201063817 Chr.7: 76330102-76330102 on Build GRCh38
rs765732943 Chr.7: 76330129-76330129 on Build GRCh38
rs183492561 Chr.7: 76330305-76330305 on Build GRCh38
rs759611694 Chr.7: 76330026-76330026 on Build GRCh38
rs187775113 Chr.7: 76330323-76330323 on Build GRCh38
rs148559105 Chr.7: 76330288-76330288 on Build GRCh38
rs778981755 Chr.7: 76329937-76329937 on Build GRCh38
rs745421481 Chr.7: 76330241-76330241 on Build GRCh38
rs997828550 Chr.7: 76330364-76330364 on Build GRCh38
rs998580355 Chr.7: 76330342-76330342 on Build GRCh38
rs1057055189 Chr.7: 76330272-76330272 on Build GRCh38
rs866370817 Chr.7: 76330158-76330158 on Build GRCh38
rs769682432 Chr.7: 76329999-76329999 on Build GRCh38
rs1048792928 Chr.7: 76330201-76330201 on Build GRCh38
rs543878282 Chr.7: 76330243-76330243 on Build GRCh38
rs764461978 Chr.7: 76330075-76330075 on Build GRCh38
rs762316167 Chr.7: 76330117-76330117 on Build GRCh38
rs770925257 Chr.7: 76330056-76330056 on Build GRCh38
rs900944525 Chr.7: 76330331-76330331 on Build GRCh38
rs774513890 Chr.7: 76330287-76330287 on Build GRCh38
rs745872119 Chr.7: 76329962-76329962 on Build GRCh38
rs775013704 Chr.7: 76330271-76330271 on Build GRCh38
rs780129232 Chr.7: 76329976-76329976 on Build GRCh38
rs758388896 Chr.7: 76330164-76330164 on Build GRCh38
rs1028516426 Chr.7: 76330365-76330365 on Build GRCh38
rs750576954 Chr.7: 76330155-76330155 on Build GRCh38
rs541111384 Chr.7: 76329925-76329925 on Build GRCh38
rs773103074 Chr.7: 76330004-76330004 on Build GRCh38
rs904357515 Chr.7: 76330341-76330341 on Build GRCh38
rs780366000 Chr.7: 76330167-76330167 on Build GRCh38
rs775844237 Chr.7: 76330259-76330259 on Build GRCh38
rs774070217 Chr.7: 76330057-76330057 on Build GRCh38
rs771049818 Chr.7: 76330214-76330214 on Build GRCh38
rs938754648 Chr.7: 76330096-76330096 on Build GRCh38
rs1055907096 Chr.7: 76330366-76330367 on Build GRCh38
rs760331444 Chr.7: 76330074-76330074 on Build GRCh38
rs976962773 Chr.7: 76329945-76329945 on Build GRCh38
rs1037532353 Chr.7: 76330070-76330070 on Build GRCh38
rs144143195 Chr.7: 76329970-76329970 on Build GRCh38
rs769855430 Chr.7: 76330124-76330124 on Build GRCh38
rs762781768 Chr.7: 76330005-76330005 on Build GRCh38
rs900832043 Chr.7: 76330358-76330358 on Build GRCh38
rs1012851921 Chr.7: 76330289-76330289 on Build GRCh38
rs749090391 Chr.7: 76330207-76330207 on Build GRCh38
rs757708675 Chr.7: 76329931-76329931 on Build GRCh38
rs778692127 Chr.7: 76330231-76330231 on Build GRCh38
rs751763117 Chr.7: 76330168-76330168 on Build GRCh38
rs760330709 Chr.7: 76330275-76330275 on Build GRCh38
rs78533137 Chr.7: 76330348-76330348 on Build GRCh38
rs748026712 Chr.7: 76329982-76329982 on Build GRCh38
rs761972667 Chr.7: 76330306-76330306 on Build GRCh38
rs758954176 Chr.7: 76330064-76330064 on Build GRCh38
rs984280298 Chr.7: 76330003-76330003 on Build GRCh38
rs150943178 Chr.7: 76330183-76330183 on Build GRCh38
rs569904540 Chr.7: 76330069-76330069 on Build GRCh38
rs1006743271 Chr.7: 76330285-76330285 on Build GRCh38
rs1057100383 Chr.7: 76330101-76330101 on Build GRCh38
rs777725046 Chr.7: 76330198-76330198 on Build GRCh38
rs921555508 Chr.7: 76329952-76329952 on Build GRCh38
rs768234902 Chr.7: 76330284-76330284 on Build GRCh38
rs1022312062 Chr.7: 76330376-76330376 on Build GRCh38

View Full Product Details