Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90384580-90385122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP10
Entrez Gene ID
85865
Gene Name
GTP binding protein 10
Gene Aliases
ObgH2, UG0751c10
UniGene
Hs.593547
Gene Chromosome Location
Chr.7: 90346665-90391455 on Build GRCh38
Transcripts
NM_001042717.2 NM_033107.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903635 c.1089G>A p.W363* Chr.7: 90385079-90385079 on Build GRCh38
6333050 c.997G>T p.E333* Chr.7: 90384987-90384987 on Build GRCh38
5903634 c.1090A>C p.I364L Chr.7: 90385080-90385080 on Build GRCh38
4807457 c.921A>T p.E307D Chr.7: 90384911-90384911 on Build GRCh38
1092827 c.997G>A p.E333K Chr.7: 90384987-90384987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781172282 Chr.7: 90384941-90384941 on Build GRCh38
rs901594076 Chr.7: 90384817-90384817 on Build GRCh38
rs756608010 Chr.7: 90384901-90384901 on Build GRCh38
rs758529195 Chr.7: 90385071-90385071 on Build GRCh38
rs551715366 Chr.7: 90385063-90385063 on Build GRCh38
rs113724934 Chr.7: 90384690-90384690 on Build GRCh38
rs796986401 Chr.7: 90384603-90384605 on Build GRCh38
rs757620202 Chr.7: 90385054-90385054 on Build GRCh38
rs999573164 Chr.7: 90384948-90384948 on Build GRCh38
rs533190751 Chr.7: 90385037-90385037 on Build GRCh38
rs991284170 Chr.7: 90385007-90385007 on Build GRCh38
rs1013337297 Chr.7: 90384628-90384631 on Build GRCh38
rs17866390 Chr.7: 90384733-90384733 on Build GRCh38
rs778015258 Chr.7: 90384910-90384910 on Build GRCh38
rs543613888 Chr.7: 90384744-90384744 on Build GRCh38
rs114771596 Chr.7: 90384850-90384850 on Build GRCh38
rs762523127 Chr.7: 90384855-90384855 on Build GRCh38
rs774027772 Chr.7: 90385009-90385009 on Build GRCh38
rs768603331 Chr.7: 90384841-90384841 on Build GRCh38
rs767125180 Chr.7: 90385035-90385035 on Build GRCh38
rs772885312 Chr.7: 90384959-90384959 on Build GRCh38
rs17869967 Chr.7: 90384826-90384826 on Build GRCh38
rs754166184 Chr.7: 90384924-90384924 on Build GRCh38
rs745885335 Chr.7: 90384910-90384910 on Build GRCh38
rs267601622 Chr.7: 90384957-90384957 on Build GRCh38
rs559724937 Chr.7: 90384861-90384861 on Build GRCh38
rs748062317 Chr.7: 90384942-90384942 on Build GRCh38
rs1013827347 Chr.7: 90384998-90384998 on Build GRCh38
rs776527786 Chr.7: 90384849-90384849 on Build GRCh38
rs35157693 Chr.7: 90384773-90384773 on Build GRCh38
rs965927782 Chr.7: 90384783-90384783 on Build GRCh38
rs9886362 Chr.7: 90384797-90384797 on Build GRCh38
rs745997084 Chr.7: 90384936-90384936 on Build GRCh38
rs1050282799 Chr.7: 90384848-90384848 on Build GRCh38
rs969059913 Chr.7: 90384651-90384651 on Build GRCh38
rs765986130 Chr.7: 90384857-90384857 on Build GRCh38
rs374680644 Chr.7: 90385042-90385042 on Build GRCh38
rs1026586790 Chr.7: 90384699-90384699 on Build GRCh38
rs201866556 Chr.7: 90385066-90385066 on Build GRCh38
rs770957088 Chr.7: 90384880-90384883 on Build GRCh38
rs762765294 Chr.7: 90384975-90384975 on Build GRCh38
rs67290421 Chr.7: 90384603-90384603 on Build GRCh38
rs1040440902 Chr.7: 90384808-90384808 on Build GRCh38
rs754176936 Chr.7: 90385048-90385048 on Build GRCh38
rs996293216 Chr.7: 90384825-90384825 on Build GRCh38
rs747023041 Chr.7: 90385091-90385091 on Build GRCh38
rs768182387 Chr.7: 90384865-90384865 on Build GRCh38
rs753255397 Chr.7: 90384872-90384872 on Build GRCh38
rs780535460 Chr.7: 90384869-90384869 on Build GRCh38
rs781318104 Chr.7: 90384909-90384910 on Build GRCh38
rs55762592 Chr.7: 90384605-90384605 on Build GRCh38
rs747529954 Chr.7: 90384869-90384872 on Build GRCh38
rs377658515 Chr.7: 90384935-90384935 on Build GRCh38
rs577546129 Chr.7: 90384609-90384609 on Build GRCh38
rs904970826 Chr.7: 90385099-90385099 on Build GRCh38
rs772107421 Chr.7: 90384938-90384938 on Build GRCh38
rs17863999 Chr.7: 90385094-90385094 on Build GRCh38
rs769647269 Chr.7: 90385092-90385092 on Build GRCh38
rs370884460 Chr.7: 90384986-90384986 on Build GRCh38
rs17863998 Chr.7: 90384603-90384603 on Build GRCh38
rs951677286 Chr.7: 90384729-90384729 on Build GRCh38
rs564641476 Chr.7: 90384731-90384731 on Build GRCh38
rs751059492 Chr.7: 90384860-90384860 on Build GRCh38
rs769091506 Chr.7: 90385017-90385017 on Build GRCh38
rs764624426 Chr.7: 90385043-90385043 on Build GRCh38
rs747347928 Chr.7: 90385004-90385004 on Build GRCh38
rs778794316 Chr.7: 90384634-90384634 on Build GRCh38
rs912734167 Chr.7: 90384781-90384781 on Build GRCh38
rs780119986 Chr.7: 90385086-90385086 on Build GRCh38
rs930895862 Chr.7: 90384665-90384665 on Build GRCh38
rs757453617 Chr.7: 90384925-90384925 on Build GRCh38
rs769659138 Chr.7: 90384954-90384954 on Build GRCh38
rs561943567 Chr.7: 90384746-90384746 on Build GRCh38

View Full Product Details