Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127600112-127600604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCN3
Entrez Gene ID
29999
Gene Name
fascin actin-bundling protein 3
Gene Aliases
-
UniGene
Hs.712692
Gene Chromosome Location
Chr.7: 127593635-127601797 on Build GRCh38
Transcripts
NM_020369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207598 c.1355C>T p.A452V Chr.7: 127600257-127600257 on Build GRCh38
2768623 c.1489G>T p.E497* Chr.7: 127600391-127600391 on Build GRCh38
5798613 c.1448T>G p.L483R Chr.7: 127600350-127600350 on Build GRCh38
3877873 c.1406C>T p.A469V Chr.7: 127600308-127600308 on Build GRCh38
4568838 c.1353T>C p.F451F Chr.7: 127600255-127600255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764322176 Chr.7: 127600303-127600303 on Build GRCh38
rs756196787 Chr.7: 127600423-127600423 on Build GRCh38
rs759185719 Chr.7: 127600354-127600354 on Build GRCh38
rs940438297 Chr.7: 127600445-127600445 on Build GRCh38
rs777925123 Chr.7: 127600243-127600243 on Build GRCh38
rs771126835 Chr.7: 127600197-127600198 on Build GRCh38
rs976329610 Chr.7: 127600382-127600382 on Build GRCh38
rs778874738 Chr.7: 127600233-127600233 on Build GRCh38
rs749716490 Chr.7: 127600274-127600274 on Build GRCh38
rs749595538 Chr.7: 127600318-127600318 on Build GRCh38
rs755319552 Chr.7: 127600438-127600438 on Build GRCh38
rs56827675 Chr.7: 127600162-127600162 on Build GRCh38
rs773941062 Chr.7: 127600178-127600178 on Build GRCh38
rs756565313 Chr.7: 127600273-127600273 on Build GRCh38
rs750486392 Chr.7: 127600422-127600422 on Build GRCh38
rs994550883 Chr.7: 127600176-127600176 on Build GRCh38
rs190031771 Chr.7: 127600157-127600157 on Build GRCh38
rs747101519 Chr.7: 127600247-127600247 on Build GRCh38
rs776250456 Chr.7: 127600347-127600347 on Build GRCh38
rs938306028 Chr.7: 127600519-127600519 on Build GRCh38
rs759840258 Chr.7: 127600166-127600166 on Build GRCh38
rs371822748 Chr.7: 127600414-127600414 on Build GRCh38
rs909393226 Chr.7: 127600444-127600444 on Build GRCh38
rs371780754 Chr.7: 127600424-127600424 on Build GRCh38
rs36022783 Chr.7: 127600205-127600206 on Build GRCh38
rs374297879 Chr.7: 127600305-127600305 on Build GRCh38
rs944960943 Chr.7: 127600206-127600206 on Build GRCh38
rs766279413 Chr.7: 127600198-127600198 on Build GRCh38
rs765307108 Chr.7: 127600296-127600296 on Build GRCh38
rs1003821282 Chr.7: 127600523-127600523 on Build GRCh38
rs757323520 Chr.7: 127600308-127600308 on Build GRCh38
rs776662325 Chr.7: 127600291-127600291 on Build GRCh38
rs934178700 Chr.7: 127600324-127600324 on Build GRCh38
rs781219378 Chr.7: 127600343-127600359 on Build GRCh38
rs370947261 Chr.7: 127600264-127600264 on Build GRCh38
rs772752415 Chr.7: 127600282-127600282 on Build GRCh38
rs763831426 Chr.7: 127600391-127600391 on Build GRCh38
rs557976681 Chr.7: 127600499-127600499 on Build GRCh38
rs898800257 Chr.7: 127600442-127600442 on Build GRCh38
rs775585547 Chr.7: 127600383-127600383 on Build GRCh38
rs932229806 Chr.7: 127600434-127600434 on Build GRCh38
rs898955706 Chr.7: 127600168-127600168 on Build GRCh38
rs767195797 Chr.7: 127600181-127600181 on Build GRCh38
rs1038689047 Chr.7: 127600141-127600141 on Build GRCh38
rs754778455 Chr.7: 127600222-127600222 on Build GRCh38
rs751760556 Chr.7: 127600406-127600406 on Build GRCh38
rs761428106 Chr.7: 127600179-127600179 on Build GRCh38
rs768904777 Chr.7: 127600328-127600328 on Build GRCh38
rs753723468 Chr.7: 127600203-127600203 on Build GRCh38
rs1049262761 Chr.7: 127600270-127600270 on Build GRCh38
rs747100059 Chr.7: 127600155-127600155 on Build GRCh38
rs1039113614 Chr.7: 127600428-127600428 on Build GRCh38
rs772880479 Chr.7: 127600329-127600329 on Build GRCh38
rs201485979 Chr.7: 127600290-127600290 on Build GRCh38
rs966477707 Chr.7: 127600360-127600360 on Build GRCh38
rs77295263 Chr.7: 127600186-127600186 on Build GRCh38
rs200283525 Chr.7: 127600315-127600315 on Build GRCh38
rs746662602 Chr.7: 127600340-127600340 on Build GRCh38
rs770152969 Chr.7: 127600171-127600171 on Build GRCh38
rs12540741 Chr.7: 127600174-127600174 on Build GRCh38
rs539623514 Chr.7: 127600346-127600346 on Build GRCh38
rs1036554947 Chr.7: 127600495-127600495 on Build GRCh38
rs968137787 Chr.7: 127600312-127600312 on Build GRCh38
rs758328839 Chr.7: 127600302-127600302 on Build GRCh38
rs202126167 Chr.7: 127600238-127600238 on Build GRCh38
rs746408667 Chr.7: 127600271-127600271 on Build GRCh38
rs1011330055 Chr.7: 127600304-127600304 on Build GRCh38
rs765253862 Chr.7: 127600363-127600363 on Build GRCh38
rs35223517 Chr.7: 127600276-127600276 on Build GRCh38
rs540487214 Chr.7: 127600532-127600532 on Build GRCh38
rs376116499 Chr.7: 127600431-127600431 on Build GRCh38
rs748586755 Chr.7: 127600447-127600447 on Build GRCh38
rs760160400 Chr.7: 127600187-127600187 on Build GRCh38
rs758665140 Chr.7: 127600239-127600239 on Build GRCh38
rs146183506 Chr.7: 127600294-127600294 on Build GRCh38
rs181240283 Chr.7: 127600314-127600314 on Build GRCh38
rs984929687 Chr.7: 127600439-127600439 on Build GRCh38
rs770932411 Chr.7: 127600248-127600248 on Build GRCh38
rs1003359604 Chr.7: 127600231-127600231 on Build GRCh38
rs149244100 Chr.7: 127600283-127600283 on Build GRCh38
rs144723488 Chr.7: 127600518-127600518 on Build GRCh38
rs1035378075 Chr.7: 127600232-127600232 on Build GRCh38
rs779125397 Chr.7: 127600446-127600446 on Build GRCh38
rs745997244 Chr.7: 127600349-127600350 on Build GRCh38
rs762744878 Chr.7: 127600384-127600384 on Build GRCh38
rs141291522 Chr.7: 127600334-127600334 on Build GRCh38
rs745982988 Chr.7: 127600311-127600311 on Build GRCh38
rs1047534216 Chr.7: 127600479-127600479 on Build GRCh38
rs75021202 Chr.7: 127600301-127600301 on Build GRCh38
rs200461532 Chr.7: 127600309-127600309 on Build GRCh38
rs777889013 Chr.7: 127600154-127600154 on Build GRCh38
rs760165475 Chr.7: 127600284-127600284 on Build GRCh38

View Full Product Details