Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139258398-139258894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBN2
Entrez Gene ID
254048
Gene Name
ubinuclein 2
Gene Aliases
-
UniGene
Hs.153458
Gene Chromosome Location
Chr.7: 139229565-139329421 on Build GRCh38
Transcripts
NM_173569.3 XM_005250249.3 XM_006715916.3 XM_011516002.2 XM_011516003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4507281 c.492C>T p.R164R Chr.7: 139258565-139258565 on Build GRCh38
6108953 c.548C>T p.P183L Chr.7: 139258621-139258621 on Build GRCh38
5979451 c.770T>C p.I257T Chr.7: 139258594-139258594 on Build GRCh38
6331556 c.531C>T p.N177N Chr.7: 139258604-139258604 on Build GRCh38
4401806 c.681C>T p.P227P Chr.7: 139258505-139258505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936965639 Chr.7: 139258457-139258457 on Build GRCh38
rs565617282 Chr.7: 139258683-139258683 on Build GRCh38
rs116050227 Chr.7: 139258461-139258461 on Build GRCh38
rs202078043 Chr.7: 139258584-139258584 on Build GRCh38
rs758495168 Chr.7: 139258666-139258666 on Build GRCh38
rs767937287 Chr.7: 139258621-139258621 on Build GRCh38
rs774176180 Chr.7: 139258600-139258600 on Build GRCh38
rs367911012 Chr.7: 139258505-139258505 on Build GRCh38
rs779086058 Chr.7: 139258557-139258557 on Build GRCh38
rs374061713 Chr.7: 139258465-139258465 on Build GRCh38
rs925281242 Chr.7: 139258423-139258423 on Build GRCh38
rs762756180 Chr.7: 139258470-139258470 on Build GRCh38
rs775086345 Chr.7: 139258468-139258468 on Build GRCh38
rs745835487 Chr.7: 139258575-139258575 on Build GRCh38
rs748782406 Chr.7: 139258591-139258591 on Build GRCh38
rs757108976 Chr.7: 139258564-139258564 on Build GRCh38
rs531397231 Chr.7: 139258462-139258462 on Build GRCh38
rs769147515 Chr.7: 139258468-139258469 on Build GRCh38
rs766858739 Chr.7: 139258633-139258633 on Build GRCh38
rs780803183 Chr.7: 139258570-139258570 on Build GRCh38
rs149834036 Chr.7: 139258541-139258541 on Build GRCh38
rs774724044 Chr.7: 139258581-139258583 on Build GRCh38
rs925063884 Chr.7: 139258711-139258711 on Build GRCh38
rs866516362 Chr.7: 139258588-139258588 on Build GRCh38
rs558096441 Chr.7: 139258737-139258737 on Build GRCh38
rs985690770 Chr.7: 139258733-139258733 on Build GRCh38
rs896868203 Chr.7: 139258508-139258508 on Build GRCh38
rs1041249804 Chr.7: 139258786-139258786 on Build GRCh38
rs537504288 Chr.7: 139258816-139258816 on Build GRCh38
rs767731256 Chr.7: 139258514-139258514 on Build GRCh38
rs993583960 Chr.7: 139258864-139258864 on Build GRCh38
rs762199095 Chr.7: 139258587-139258589 on Build GRCh38
rs759582689 Chr.7: 139258638-139258638 on Build GRCh38
rs972513909 Chr.7: 139258429-139258429 on Build GRCh38
rs576366810 Chr.7: 139258758-139258758 on Build GRCh38
rs935077484 Chr.7: 139258718-139258718 on Build GRCh38
rs1044918742 Chr.7: 139258769-139258769 on Build GRCh38
rs896126115 Chr.7: 139258724-139258724 on Build GRCh38
rs768460300 Chr.7: 139258594-139258594 on Build GRCh38
rs766483908 Chr.7: 139258544-139258544 on Build GRCh38
rs765262596 Chr.7: 139258648-139258648 on Build GRCh38
rs767796665 Chr.7: 139258676-139258676 on Build GRCh38
rs1006042251 Chr.7: 139258805-139258805 on Build GRCh38
rs547224611 Chr.7: 139258664-139258664 on Build GRCh38
rs146366044 Chr.7: 139258589-139258589 on Build GRCh38
rs897927773 Chr.7: 139258863-139258863 on Build GRCh38
rs556798660 Chr.7: 139258419-139258419 on Build GRCh38
rs949348177 Chr.7: 139258735-139258735 on Build GRCh38
rs763821056 Chr.7: 139258500-139258500 on Build GRCh38
rs568196064 Chr.7: 139258556-139258556 on Build GRCh38
rs936663129 Chr.7: 139258726-139258726 on Build GRCh38
rs376185098 Chr.7: 139258460-139258460 on Build GRCh38
rs796374592 Chr.7: 139258530-139258530 on Build GRCh38
rs368951598 Chr.7: 139258616-139258616 on Build GRCh38
rs1026690363 Chr.7: 139258580-139258580 on Build GRCh38
rs1016046784 Chr.7: 139258808-139258808 on Build GRCh38
rs746848105 Chr.7: 139258563-139258563 on Build GRCh38
rs750960706 Chr.7: 139258524-139258524 on Build GRCh38
rs978918881 Chr.7: 139258703-139258703 on Build GRCh38
rs112375645 Chr.7: 139258721-139258721 on Build GRCh38
rs773382013 Chr.7: 139258622-139258622 on Build GRCh38
rs753910923 Chr.7: 139258547-139258547 on Build GRCh38
rs753968828 Chr.7: 139258636-139258636 on Build GRCh38
rs910740851 Chr.7: 139258551-139258551 on Build GRCh38
rs34615984 Chr.7: 139258444-139258445 on Build GRCh38
rs371819353 Chr.7: 139258539-139258539 on Build GRCh38
rs747701222 Chr.7: 139258702-139258702 on Build GRCh38
rs74531371 Chr.7: 139258676-139258676 on Build GRCh38
rs905040940 Chr.7: 139258804-139258804 on Build GRCh38
rs966855992 Chr.7: 139258698-139258699 on Build GRCh38
rs746794797 Chr.7: 139258451-139258451 on Build GRCh38
rs763540799 Chr.7: 139258449-139258449 on Build GRCh38
rs761310556 Chr.7: 139258625-139258625 on Build GRCh38

View Full Product Details