Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41541668-41542067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS4
Entrez Gene ID
84296
Gene Name
GINS complex subunit 4
Gene Aliases
SLD5
UniGene
Hs.656996
Gene Chromosome Location
Chr.8: 41529206-41545046 on Build GRCh38
Transcripts
NM_032336.2 XM_005273659.3 NR_125824.1

Gene Symbol
LOC102723729
Entrez Gene ID
102723729
Gene Name
uncharacterized LOC102723729
Gene Aliases
-
UniGene
Hs.586384
Gene Chromosome Location
Chr.8: 41534305-41578368 on Build GRCh38
Transcripts
NM_032336.2 XM_005273659.3 NR_125824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454155 c.583G>A p.V195M Chr.8: 41541998-41541998 on Build GRCh38
3900156 c.582C>T p.Y194Y Chr.8: 41541997-41541997 on Build GRCh38
3900155 c.537A>C p.Q179H Chr.8: 41541861-41541861 on Build GRCh38
1457064 c.546A>G p.I182M Chr.8: 41541870-41541870 on Build GRCh38
6561477 c.589G>A p.D197N Chr.8: 41542004-41542004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148743285 Chr.8: 41541998-41541998 on Build GRCh38
rs769176951 Chr.8: 41541762-41541762 on Build GRCh38
rs779799341 Chr.8: 41541904-41541904 on Build GRCh38
rs201868897 Chr.8: 41541908-41541908 on Build GRCh38
rs556997519 Chr.8: 41541856-41541856 on Build GRCh38
rs769636118 Chr.8: 41542027-41542027 on Build GRCh38
rs749910879 Chr.8: 41541796-41541796 on Build GRCh38
rs1005122408 Chr.8: 41541704-41541704 on Build GRCh38
rs781762102 Chr.8: 41541892-41541892 on Build GRCh38
rs577571618 Chr.8: 41541926-41541926 on Build GRCh38
rs911885221 Chr.8: 41541970-41541970 on Build GRCh38
rs565761441 Chr.8: 41541746-41541746 on Build GRCh38
rs370288384 Chr.8: 41541784-41541784 on Build GRCh38
rs553503109 Chr.8: 41541942-41541942 on Build GRCh38
rs142942452 Chr.8: 41541828-41541828 on Build GRCh38
rs760616817 Chr.8: 41541935-41541935 on Build GRCh38
rs748396243 Chr.8: 41541766-41541766 on Build GRCh38
rs374202550 Chr.8: 41541909-41541909 on Build GRCh38
rs1031135106 Chr.8: 41541880-41541880 on Build GRCh38
rs772826423 Chr.8: 41542033-41542033 on Build GRCh38
rs765295271 Chr.8: 41541995-41541995 on Build GRCh38
rs377034608 Chr.8: 41541834-41541834 on Build GRCh38
rs747997128 Chr.8: 41542025-41542025 on Build GRCh38
rs78860704 Chr.8: 41541940-41541940 on Build GRCh38
rs897377255 Chr.8: 41541962-41541962 on Build GRCh38
rs141417166 Chr.8: 41542018-41542018 on Build GRCh38
rs532286065 Chr.8: 41541696-41541696 on Build GRCh38
rs776544104 Chr.8: 41541950-41541950 on Build GRCh38
rs759545722 Chr.8: 41541976-41541982 on Build GRCh38
rs141719341 Chr.8: 41541835-41541835 on Build GRCh38
rs569569552 Chr.8: 41541695-41541695 on Build GRCh38
rs898982205 Chr.8: 41541981-41541981 on Build GRCh38
rs931993947 Chr.8: 41541692-41541692 on Build GRCh38
rs112065135 Chr.8: 41541974-41541974 on Build GRCh38
rs534849373 Chr.8: 41541772-41541772 on Build GRCh38
rs372866931 Chr.8: 41541773-41541773 on Build GRCh38
rs989133354 Chr.8: 41541771-41541771 on Build GRCh38
rs750387861 Chr.8: 41541832-41541833 on Build GRCh38
rs371910436 Chr.8: 41541955-41541955 on Build GRCh38
rs755486248 Chr.8: 41541871-41541871 on Build GRCh38
rs117165584 Chr.8: 41541794-41541794 on Build GRCh38
rs79350355 Chr.8: 41541978-41541978 on Build GRCh38
rs144307110 Chr.8: 41541857-41541857 on Build GRCh38
rs372934804 Chr.8: 41541928-41541928 on Build GRCh38
rs771025991 Chr.8: 41541925-41541925 on Build GRCh38
rs569175425 Chr.8: 41541700-41541700 on Build GRCh38
rs775426528 Chr.8: 41541792-41541792 on Build GRCh38
rs767309887 Chr.8: 41541790-41541792 on Build GRCh38
rs756526940 Chr.8: 41541991-41541991 on Build GRCh38
rs753568737 Chr.8: 41541801-41541801 on Build GRCh38
rs1055455430 Chr.8: 41542007-41542007 on Build GRCh38
rs891279705 Chr.8: 41541927-41541927 on Build GRCh38
rs753126972 Chr.8: 41541983-41541983 on Build GRCh38
rs777955735 Chr.8: 41541992-41541992 on Build GRCh38
rs768180460 Chr.8: 41541982-41541982 on Build GRCh38
rs185706457 Chr.8: 41541954-41541954 on Build GRCh38
rs764808238 Chr.8: 41541765-41541765 on Build GRCh38
rs995779354 Chr.8: 41541964-41541964 on Build GRCh38
rs763136903 Chr.8: 41541780-41541780 on Build GRCh38
rs781536483 Chr.8: 41541972-41541972 on Build GRCh38
rs749526438 Chr.8: 41541997-41541997 on Build GRCh38
rs989545816 Chr.8: 41541921-41541921 on Build GRCh38
rs774118928 Chr.8: 41542040-41542040 on Build GRCh38
rs192899689 Chr.8: 41541830-41541830 on Build GRCh38
rs778109760 Chr.8: 41541913-41541913 on Build GRCh38
rs779090249 Chr.8: 41542015-41542015 on Build GRCh38
rs749418771 Chr.8: 41541920-41541920 on Build GRCh38
rs772107093 Chr.8: 41542021-41542021 on Build GRCh38
rs750122115 Chr.8: 41541969-41541969 on Build GRCh38
rs759171041 Chr.8: 41542045-41542045 on Build GRCh38
rs546011448 Chr.8: 41542035-41542035 on Build GRCh38
rs766205000 Chr.8: 41542039-41542039 on Build GRCh38
rs776678766 Chr.8: 41542024-41542024 on Build GRCh38

View Full Product Details