Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 51319442-51319977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXDNL
Entrez Gene ID
137902
Gene Name
peroxidasin like
Gene Aliases
PMR1, PRM1, VPO2
UniGene
Hs.444882
Gene Chromosome Location
Chr.8: 51319575-51811031 on Build GRCh38
Transcripts
NM_144651.4 XM_005251168.3 XM_006716420.3 XM_011517456.2 XM_011517457.2 XM_011517458.2 XM_017013041.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2718295 c.4372G>A p.D1458N Chr.8: 51319911-51319911 on Build GRCh38
6113735 c.4387C>A p.R1463S Chr.8: 51319896-51319896 on Build GRCh38
2718294 c.4388G>T p.R1463L Chr.8: 51319895-51319895 on Build GRCh38
2718298 c.4334C>G p.T1445S Chr.8: 51319949-51319949 on Build GRCh38
2718296 c.4369A>C p.S1457R Chr.8: 51319914-51319914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956127802 Chr.8: 51319651-51319651 on Build GRCh38
rs889540286 Chr.8: 51319512-51319512 on Build GRCh38
rs1000613312 Chr.8: 51319622-51319622 on Build GRCh38
rs555206034 Chr.8: 51319673-51319673 on Build GRCh38
rs914252016 Chr.8: 51319925-51319925 on Build GRCh38
rs918312411 Chr.8: 51319777-51319777 on Build GRCh38
rs372092860 Chr.8: 51319852-51319852 on Build GRCh38
rs573435731 Chr.8: 51319690-51319690 on Build GRCh38
rs1016200605 Chr.8: 51319739-51319739 on Build GRCh38
rs1042363036 Chr.8: 51319945-51319945 on Build GRCh38
rs62508843 Chr.8: 51319670-51319670 on Build GRCh38
rs912096030 Chr.8: 51319835-51319835 on Build GRCh38
rs776133432 Chr.8: 51319896-51319896 on Build GRCh38
rs918338202 Chr.8: 51319521-51319521 on Build GRCh38
rs918478186 Chr.8: 51319744-51319744 on Build GRCh38
rs868763792 Chr.8: 51319901-51319901 on Build GRCh38
rs929445557 Chr.8: 51319526-51319526 on Build GRCh38
rs187618172 Chr.8: 51319557-51319557 on Build GRCh38
rs762818923 Chr.8: 51319938-51319938 on Build GRCh38
rs1054108562 Chr.8: 51319546-51319546 on Build GRCh38
rs369513519 Chr.8: 51319895-51319895 on Build GRCh38
rs769760381 Chr.8: 51319856-51319856 on Build GRCh38
rs779892019 Chr.8: 51319949-51319949 on Build GRCh38
rs774749299 Chr.8: 51319887-51319887 on Build GRCh38
rs116291347 Chr.8: 51319741-51319741 on Build GRCh38
rs984787500 Chr.8: 51319730-51319730 on Build GRCh38
rs140109714 Chr.8: 51319743-51319743 on Build GRCh38
rs942376537 Chr.8: 51319519-51319519 on Build GRCh38
rs764894926 Chr.8: 51319932-51319932 on Build GRCh38
rs544181382 Chr.8: 51319769-51319769 on Build GRCh38
rs759933244 Chr.8: 51319889-51319889 on Build GRCh38
rs770423376 Chr.8: 51319953-51319953 on Build GRCh38
rs1052704 Chr.8: 51319927-51319927 on Build GRCh38
rs1051296863 Chr.8: 51319466-51319466 on Build GRCh38
rs372589323 Chr.8: 51319923-51319923 on Build GRCh38
rs986642915 Chr.8: 51319805-51319805 on Build GRCh38
rs779907994 Chr.8: 51319848-51319848 on Build GRCh38
rs199528987 Chr.8: 51319952-51319952 on Build GRCh38
rs958776367 Chr.8: 51319715-51319715 on Build GRCh38
rs868014103 Chr.8: 51319779-51319779 on Build GRCh38
rs571322831 Chr.8: 51319535-51319535 on Build GRCh38
rs962371217 Chr.8: 51319754-51319754 on Build GRCh38
rs753425862 Chr.8: 51319926-51319926 on Build GRCh38
rs557396651 Chr.8: 51319647-51319647 on Build GRCh38
rs375457259 Chr.8: 51319879-51319879 on Build GRCh38
rs559922943 Chr.8: 51319947-51319947 on Build GRCh38
rs760370959 Chr.8: 51319942-51319943 on Build GRCh38
rs540345882 Chr.8: 51319701-51319701 on Build GRCh38
rs1047871507 Chr.8: 51319808-51319808 on Build GRCh38
rs761300116 Chr.8: 51319910-51319910 on Build GRCh38
rs771229142 Chr.8: 51319885-51319885 on Build GRCh38
rs142250763 Chr.8: 51319820-51319820 on Build GRCh38
rs369555697 Chr.8: 51319948-51319948 on Build GRCh38
rs750088139 Chr.8: 51319939-51319939 on Build GRCh38
rs267601939 Chr.8: 51319922-51319922 on Build GRCh38
rs893413839 Chr.8: 51319575-51319575 on Build GRCh38
rs890689119 Chr.8: 51319574-51319574 on Build GRCh38
rs149627511 Chr.8: 51319656-51319656 on Build GRCh38
rs747020324 Chr.8: 51319597-51319597 on Build GRCh38
rs773399170 Chr.8: 51319862-51319862 on Build GRCh38
rs944874217 Chr.8: 51319870-51319870 on Build GRCh38
rs372262910 Chr.8: 51319832-51319832 on Build GRCh38

View Full Product Details