Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66871409-66871959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCMDC2
Entrez Gene ID
157777
Gene Name
minichromosome maintenance domain containing 2
Gene Aliases
C8orf45
UniGene
Hs.437006
Gene Chromosome Location
Chr.8: 66870749-66926436 on Build GRCh38
Transcripts
NM_001136160.1 NM_001136161.1 NM_173518.4 XM_005251174.2 XM_006716427.3 XM_006716429.2 XM_006716433.3 XM_011517467.2 XM_011517468.2 XM_011517469.2 XM_017013140.1

SNP Detail

SNP ID Chromosome Location
rs188734068 Chr.8: 66871831-66871831 on Build GRCh38
rs997098532 Chr.8: 66871806-66871806 on Build GRCh38
rs1045205036 Chr.8: 66871718-66871718 on Build GRCh38
rs889811080 Chr.8: 66871530-66871530 on Build GRCh38
rs746946269 Chr.8: 66871584-66871584 on Build GRCh38
rs897150606 Chr.8: 66871922-66871922 on Build GRCh38
rs930791376 Chr.8: 66871454-66871454 on Build GRCh38
rs961334935 Chr.8: 66871877-66871877 on Build GRCh38
rs973737979 Chr.8: 66871878-66871878 on Build GRCh38
rs889850981 Chr.8: 66871842-66871842 on Build GRCh38
rs934835337 Chr.8: 66871440-66871440 on Build GRCh38
rs780946799 Chr.8: 66871803-66871803 on Build GRCh38
rs566087708 Chr.8: 66871637-66871637 on Build GRCh38
rs1050282841 Chr.8: 66871788-66871788 on Build GRCh38
rs561057007 Chr.8: 66871481-66871481 on Build GRCh38
rs527708858 Chr.8: 66871575-66871575 on Build GRCh38
rs777252485 Chr.8: 66871644-66871644 on Build GRCh38
rs754809990 Chr.8: 66871702-66871702 on Build GRCh38
rs532418480 Chr.8: 66871577-66871577 on Build GRCh38
rs73262317 Chr.8: 66871519-66871519 on Build GRCh38
rs993846448 Chr.8: 66871933-66871933 on Build GRCh38
rs368364828 Chr.8: 66871918-66871918 on Build GRCh38
rs922784037 Chr.8: 66871755-66871755 on Build GRCh38
rs904762876 Chr.8: 66871435-66871435 on Build GRCh38
rs80098250 Chr.8: 66871729-66871729 on Build GRCh38
rs532536836 Chr.8: 66871648-66871648 on Build GRCh38
rs193265393 Chr.8: 66871640-66871640 on Build GRCh38
rs926217253 Chr.8: 66871699-66871699 on Build GRCh38
rs76665003 Chr.8: 66871572-66871572 on Build GRCh38
rs1045725454 Chr.8: 66871477-66871477 on Build GRCh38
rs1005558455 Chr.8: 66871869-66871869 on Build GRCh38
rs568318334 Chr.8: 66871740-66871740 on Build GRCh38
rs780098247 Chr.8: 66871574-66871574 on Build GRCh38
rs993016014 Chr.8: 66871615-66871615 on Build GRCh38
rs1056432942 Chr.8: 66871761-66871761 on Build GRCh38
rs186622650 Chr.8: 66871566-66871566 on Build GRCh38
rs114144116 Chr.8: 66871479-66871479 on Build GRCh38
rs934949923 Chr.8: 66871720-66871720 on Build GRCh38
rs998713979 Chr.8: 66871517-66871517 on Build GRCh38
rs748395330 Chr.8: 66871872-66871872 on Build GRCh38
rs866505157 Chr.8: 66871757-66871757 on Build GRCh38
rs959882961 Chr.8: 66871611-66871611 on Build GRCh38
rs903915015 Chr.8: 66871799-66871799 on Build GRCh38
rs397939776 Chr.8: 66871914-66871915 on Build GRCh38
rs77837973 Chr.8: 66871478-66871478 on Build GRCh38
rs892461004 Chr.8: 66871772-66871772 on Build GRCh38
rs974235634 Chr.8: 66871655-66871655 on Build GRCh38
rs1028141371 Chr.8: 66871824-66871824 on Build GRCh38
rs1010066027 Chr.8: 66871538-66871538 on Build GRCh38
rs1022164125 Chr.8: 66871550-66871550 on Build GRCh38
rs183716895 Chr.8: 66871682-66871682 on Build GRCh38
rs1009536878 Chr.8: 66871782-66871782 on Build GRCh38
rs955260919 Chr.8: 66871621-66871621 on Build GRCh38
rs955312892 Chr.8: 66871866-66871866 on Build GRCh38
rs55682741 Chr.8: 66871610-66871610 on Build GRCh38
rs1016208215 Chr.8: 66871567-66871567 on Build GRCh38

View Full Product Details