Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22927501-22928005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEBP4
Entrez Gene ID
157310
Gene Name
phosphatidylethanolamine binding protein 4
Gene Aliases
CORK-1, CORK1, GWTM1933, HEL-S-300, PEBP-4, PRO4408, hPEBP4
UniGene
Hs.491242
Gene Chromosome Location
Chr.8: 22713251-22941095 on Build GRCh38
Transcripts
NM_144962.2 XM_011544414.2 XM_017013103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3765399 c.69C>T p.D23D Chr.8: 22927646-22927646 on Build GRCh38
6743841 c.103C>A p.L35I Chr.8: 22927612-22927612 on Build GRCh38
2786441 c.28G>A p.A10T Chr.8: 22927687-22927687 on Build GRCh38
750060 c.24C>A p.V8V Chr.8: 22927691-22927691 on Build GRCh38
5948283 c.34C>T p.L12L Chr.8: 22927681-22927681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201758779 Chr.8: 22927736-22927736 on Build GRCh38
rs371970174 Chr.8: 22927604-22927604 on Build GRCh38
rs973761781 Chr.8: 22927793-22927793 on Build GRCh38
rs191595252 Chr.8: 22927916-22927916 on Build GRCh38
rs73218763 Chr.8: 22927534-22927534 on Build GRCh38
rs770187168 Chr.8: 22927537-22927537 on Build GRCh38
rs981529498 Chr.8: 22927586-22927586 on Build GRCh38
rs770071784 Chr.8: 22927765-22927765 on Build GRCh38
rs554335329 Chr.8: 22927914-22927914 on Build GRCh38
rs899586369 Chr.8: 22927637-22927637 on Build GRCh38
rs546399691 Chr.8: 22927744-22927744 on Build GRCh38
rs554372055 Chr.8: 22927666-22927666 on Build GRCh38
rs777017102 Chr.8: 22927710-22927710 on Build GRCh38
rs753893491 Chr.8: 22927861-22927861 on Build GRCh38
rs200783083 Chr.8: 22927584-22927584 on Build GRCh38
rs546935000 Chr.8: 22927840-22927840 on Build GRCh38
rs1043098115 Chr.8: 22927796-22927796 on Build GRCh38
rs751355034 Chr.8: 22927790-22927790 on Build GRCh38
rs1042303373 Chr.8: 22927905-22927905 on Build GRCh38
rs921961659 Chr.8: 22927867-22927867 on Build GRCh38
rs747534801 Chr.8: 22927694-22927694 on Build GRCh38
rs757416949 Chr.8: 22927741-22927741 on Build GRCh38
rs778845368 Chr.8: 22927673-22927673 on Build GRCh38
rs769403186 Chr.8: 22927791-22927791 on Build GRCh38
rs746027406 Chr.8: 22927680-22927680 on Build GRCh38
rs951224700 Chr.8: 22927822-22927822 on Build GRCh38
rs368497848 Chr.8: 22927823-22927823 on Build GRCh38
rs1027495509 Chr.8: 22927886-22927886 on Build GRCh38
rs763960082 Chr.8: 22927730-22927730 on Build GRCh38
rs932092060 Chr.8: 22927722-22927722 on Build GRCh38
rs1002525783 Chr.8: 22927766-22927766 on Build GRCh38
rs577911332 Chr.8: 22927745-22927745 on Build GRCh38
rs780823329 Chr.8: 22927603-22927603 on Build GRCh38
rs181166548 Chr.8: 22927749-22927749 on Build GRCh38
rs1026696343 Chr.8: 22927801-22927801 on Build GRCh38
rs767425972 Chr.8: 22927653-22927653 on Build GRCh38
rs760122549 Chr.8: 22927559-22927559 on Build GRCh38
rs117342433 Chr.8: 22927825-22927825 on Build GRCh38
rs772801097 Chr.8: 22927634-22927634 on Build GRCh38
rs907448919 Chr.8: 22927944-22927944 on Build GRCh38
rs762569301 Chr.8: 22927713-22927713 on Build GRCh38
rs142439943 Chr.8: 22927669-22927669 on Build GRCh38
rs376279710 Chr.8: 22927750-22927750 on Build GRCh38
rs755926565 Chr.8: 22927660-22927660 on Build GRCh38
rs1054618991 Chr.8: 22927739-22927739 on Build GRCh38
rs376727009 Chr.8: 22927643-22927643 on Build GRCh38
rs766962600 Chr.8: 22927717-22927717 on Build GRCh38
rs202161701 Chr.8: 22927629-22927629 on Build GRCh38
rs774531378 Chr.8: 22927627-22927627 on Build GRCh38
rs778087882 Chr.8: 22927611-22927611 on Build GRCh38
rs772048457 Chr.8: 22927689-22927689 on Build GRCh38
rs190294068 Chr.8: 22927656-22927656 on Build GRCh38
rs369274882 Chr.8: 22927720-22927720 on Build GRCh38
rs760738425 Chr.8: 22927727-22927727 on Build GRCh38
rs2466225 Chr.8: 22927873-22927873 on Build GRCh38
rs576056387 Chr.8: 22927890-22927890 on Build GRCh38
rs183564569 Chr.8: 22927943-22927943 on Build GRCh38
rs754855160 Chr.8: 22927648-22927648 on Build GRCh38
rs749546056 Chr.8: 22927612-22927612 on Build GRCh38
rs771150371 Chr.8: 22927557-22927557 on Build GRCh38
rs760962272 Chr.8: 22927630-22927630 on Build GRCh38
rs543218718 Chr.8: 22927778-22927778 on Build GRCh38
rs531060762 Chr.8: 22927761-22927761 on Build GRCh38
rs11547477 Chr.8: 22927691-22927691 on Build GRCh38
rs1023844290 Chr.8: 22927864-22927864 on Build GRCh38
rs550198602 Chr.8: 22927874-22927874 on Build GRCh38
rs185312207 Chr.8: 22927785-22927785 on Build GRCh38
rs17685420 Chr.8: 22927888-22927888 on Build GRCh38
rs761353296 Chr.8: 22927687-22927687 on Build GRCh38
rs755981471 Chr.8: 22927610-22927610 on Build GRCh38
rs992288873 Chr.8: 22927701-22927701 on Build GRCh38
rs1039567856 Chr.8: 22927788-22927788 on Build GRCh38
rs765119945 Chr.8: 22927714-22927714 on Build GRCh38
rs992231655 Chr.8: 22927554-22927554 on Build GRCh38
rs762621694 Chr.8: 22927640-22927640 on Build GRCh38
rs974476138 Chr.8: 22927733-22927733 on Build GRCh38
rs757843846 Chr.8: 22927596-22927596 on Build GRCh38
rs538633446 Chr.8: 22927600-22927600 on Build GRCh38
rs1003571112 Chr.8: 22927957-22927957 on Build GRCh38
rs369536370 Chr.8: 22927628-22927628 on Build GRCh38
rs148015046 Chr.8: 22927599-22927599 on Build GRCh38
rs2466224 Chr.8: 22927848-22927848 on Build GRCh38
rs768967211 Chr.8: 22927698-22927698 on Build GRCh38
rs566428762 Chr.8: 22927976-22927977 on Build GRCh38
rs1013698721 Chr.8: 22927854-22927854 on Build GRCh38
rs2175192 Chr.8: 22927646-22927646 on Build GRCh38
rs545439608 Chr.8: 22927747-22927747 on Build GRCh38
rs1020962939 Chr.8: 22927568-22927568 on Build GRCh38
rs199555529 Chr.8: 22927631-22927631 on Build GRCh38
rs754083252 Chr.8: 22927667-22927667 on Build GRCh38
rs79977603 Chr.8: 22927891-22927891 on Build GRCh38
rs773731388 Chr.8: 22927952-22927952 on Build GRCh38
rs373579924 Chr.8: 22927560-22927560 on Build GRCh38
rs774675048 Chr.8: 22927558-22927558 on Build GRCh38
rs916682579 Chr.8: 22927556-22927556 on Build GRCh38
rs902877531 Chr.8: 22927816-22927816 on Build GRCh38
rs749318675 Chr.8: 22927541-22927541 on Build GRCh38
rs377109138 Chr.8: 22927622-22927622 on Build GRCh38
rs761442342 Chr.8: 22927562-22927562 on Build GRCh38
rs761168066 Chr.8: 22927966-22927966 on Build GRCh38
rs750326624 Chr.8: 22927743-22927743 on Build GRCh38
rs750080189 Chr.8: 22927594-22927594 on Build GRCh38

View Full Product Details