Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23316866-23317369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOXL2
Entrez Gene ID
4017
Gene Name
lysyl oxidase like 2
Gene Aliases
LOR2, WS9-14
UniGene
Hs.626637
Gene Chromosome Location
Chr.8: 23296897-23404209 on Build GRCh38
Transcripts
NM_002318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
150468 c.1599C>T p.G533G Chr.8: 23316986-23316986 on Build GRCh38
7215570 c.1617C>T p.A539A Chr.8: 23316968-23316968 on Build GRCh38
3647364 c.1567C>T p.H523Y Chr.8: 23317018-23317018 on Build GRCh38
1635724 c.1581C>T p.D527D Chr.8: 23317004-23317004 on Build GRCh38
6837215 c.1471-1G>A p.? Chr.8: 23317115-23317115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051062398 Chr.8: 23317233-23317233 on Build GRCh38
rs757624681 Chr.8: 23317244-23317244 on Build GRCh38
rs755229911 Chr.8: 23317050-23317050 on Build GRCh38
rs145002005 Chr.8: 23317003-23317003 on Build GRCh38
rs926306789 Chr.8: 23317346-23317346 on Build GRCh38
rs113598291 Chr.8: 23317140-23317141 on Build GRCh38
rs78917444 Chr.8: 23317155-23317155 on Build GRCh38
rs752353125 Chr.8: 23316950-23316950 on Build GRCh38
rs748190912 Chr.8: 23316995-23316995 on Build GRCh38
rs768445976 Chr.8: 23317117-23317117 on Build GRCh38
rs767095447 Chr.8: 23316944-23316944 on Build GRCh38
rs572475264 Chr.8: 23317337-23317337 on Build GRCh38
rs146017667 Chr.8: 23317028-23317028 on Build GRCh38
rs868434696 Chr.8: 23317006-23317006 on Build GRCh38
rs1009744912 Chr.8: 23317252-23317252 on Build GRCh38
rs778888581 Chr.8: 23316974-23316974 on Build GRCh38
rs370872248 Chr.8: 23317096-23317096 on Build GRCh38
rs142665521 Chr.8: 23317013-23317013 on Build GRCh38
rs778640877 Chr.8: 23316907-23316907 on Build GRCh38
rs201353287 Chr.8: 23316987-23316987 on Build GRCh38
rs76278588 Chr.8: 23317312-23317312 on Build GRCh38
rs765119814 Chr.8: 23317039-23317039 on Build GRCh38
rs939159318 Chr.8: 23316977-23316977 on Build GRCh38
rs756177095 Chr.8: 23317066-23317066 on Build GRCh38
rs75209794 Chr.8: 23317311-23317311 on Build GRCh38
rs140003064 Chr.8: 23317160-23317160 on Build GRCh38
rs754579112 Chr.8: 23317058-23317062 on Build GRCh38
rs757130776 Chr.8: 23316906-23316906 on Build GRCh38
rs1025116445 Chr.8: 23317267-23317267 on Build GRCh38
rs778145993 Chr.8: 23317072-23317072 on Build GRCh38
rs891221139 Chr.8: 23317022-23317022 on Build GRCh38
rs748246154 Chr.8: 23317062-23317062 on Build GRCh38
rs149587418 Chr.8: 23316985-23316985 on Build GRCh38
rs753760992 Chr.8: 23316903-23316903 on Build GRCh38
rs748952661 Chr.8: 23316969-23316970 on Build GRCh38
rs368479016 Chr.8: 23316930-23316930 on Build GRCh38
rs771328795 Chr.8: 23317093-23317093 on Build GRCh38
rs776384340 Chr.8: 23317129-23317129 on Build GRCh38
rs772234310 Chr.8: 23316910-23316910 on Build GRCh38
rs146073991 Chr.8: 23317044-23317044 on Build GRCh38
rs1804833 Chr.8: 23317075-23317075 on Build GRCh38
rs754916263 Chr.8: 23316896-23316896 on Build GRCh38
rs918582603 Chr.8: 23317180-23317180 on Build GRCh38
rs772586986 Chr.8: 23317145-23317145 on Build GRCh38
rs1018879026 Chr.8: 23317037-23317037 on Build GRCh38
rs369018704 Chr.8: 23316967-23316967 on Build GRCh38
rs1001557890 Chr.8: 23317222-23317222 on Build GRCh38
rs13250587 Chr.8: 23317235-23317235 on Build GRCh38
rs778493926 Chr.8: 23317272-23317272 on Build GRCh38
rs141290588 Chr.8: 23317049-23317049 on Build GRCh38
rs761360325 Chr.8: 23317029-23317029 on Build GRCh38
rs766269460 Chr.8: 23317147-23317147 on Build GRCh38
rs116512927 Chr.8: 23316920-23316920 on Build GRCh38
rs749744310 Chr.8: 23317091-23317091 on Build GRCh38
rs781365662 Chr.8: 23317056-23317056 on Build GRCh38
rs373023981 Chr.8: 23316973-23316973 on Build GRCh38
rs750210282 Chr.8: 23316979-23316979 on Build GRCh38
rs530034771 Chr.8: 23316919-23316919 on Build GRCh38
rs202166801 Chr.8: 23317021-23317021 on Build GRCh38
rs79320561 Chr.8: 23317310-23317310 on Build GRCh38
rs778425980 Chr.8: 23317126-23317130 on Build GRCh38
rs919120758 Chr.8: 23317137-23317137 on Build GRCh38
rs764748451 Chr.8: 23317143-23317143 on Build GRCh38
rs755567371 Chr.8: 23317206-23317206 on Build GRCh38
rs893330013 Chr.8: 23317287-23317287 on Build GRCh38
rs771258066 Chr.8: 23317012-23317012 on Build GRCh38
rs1034701107 Chr.8: 23317342-23317342 on Build GRCh38
rs199878615 Chr.8: 23316935-23316935 on Build GRCh38
rs372762004 Chr.8: 23317136-23317136 on Build GRCh38
rs536996583 Chr.8: 23317242-23317242 on Build GRCh38
rs762369102 Chr.8: 23316927-23316927 on Build GRCh38
rs35951329 Chr.8: 23317310-23317312 on Build GRCh38
rs745645431 Chr.8: 23316909-23316909 on Build GRCh38
rs979549805 Chr.8: 23316999-23316999 on Build GRCh38
rs3824213 Chr.8: 23316912-23316912 on Build GRCh38
rs562648730 Chr.8: 23317279-23317279 on Build GRCh38
rs138796129 Chr.8: 23317004-23317004 on Build GRCh38
rs374553547 Chr.8: 23317097-23317097 on Build GRCh38
rs369368043 Chr.8: 23317308-23317309 on Build GRCh38
rs1019391924 Chr.8: 23317243-23317243 on Build GRCh38
rs538881351 Chr.8: 23317172-23317172 on Build GRCh38
rs746213240 Chr.8: 23317105-23317105 on Build GRCh38
rs7357444 Chr.8: 23316968-23316968 on Build GRCh38
rs200898540 Chr.8: 23317143-23317144 on Build GRCh38
rs370168554 Chr.8: 23316936-23316936 on Build GRCh38
rs750311862 Chr.8: 23317042-23317042 on Build GRCh38
rs377292868 Chr.8: 23317312-23317313 on Build GRCh38
rs201434218 Chr.8: 23317146-23317146 on Build GRCh38
rs747178789 Chr.8: 23316991-23316991 on Build GRCh38
rs3736016 Chr.8: 23316986-23316986 on Build GRCh38
rs767755976 Chr.8: 23317163-23317163 on Build GRCh38
rs758143352 Chr.8: 23317043-23317043 on Build GRCh38
rs760460697 Chr.8: 23317020-23317020 on Build GRCh38
rs142667936 Chr.8: 23317303-23317303 on Build GRCh38
rs777139811 Chr.8: 23316921-23316921 on Build GRCh38
rs149160173 Chr.8: 23316992-23316992 on Build GRCh38

View Full Product Details