Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144506395-144506900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPT
Entrez Gene ID
2875
Gene Name
glutamic--pyruvic transaminase
Gene Aliases
AAT1, ALT1, GPT1
UniGene
Hs.103502
Gene Chromosome Location
Chr.8: 144502217-144507174 on Build GRCh38
Transcripts
NM_005309.2 XM_011516993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639253 c.1262C>T p.P421L Chr.8: 144506631-144506631 on Build GRCh38
6674701 c.1356G>T p.V452V Chr.8: 144506799-144506799 on Build GRCh38
5016240 c.1324_1325insGT p.L443fs*27 Chr.8: 144506767-144506768 on Build GRCh38
7418393 c.1190T>C p.F397S Chr.8: 144506559-144506559 on Build GRCh38
6315111 c.1311G>T p.M437I Chr.8: 144506754-144506754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376189057 Chr.8: 144506855-144506855 on Build GRCh38
rs755869216 Chr.8: 144506689-144506689 on Build GRCh38
rs976061797 Chr.8: 144506514-144506514 on Build GRCh38
rs750427788 Chr.8: 144506599-144506599 on Build GRCh38
rs774335721 Chr.8: 144506484-144506484 on Build GRCh38
rs370730581 Chr.8: 144506468-144506468 on Build GRCh38
rs749288207 Chr.8: 144506853-144506853 on Build GRCh38
rs1036245021 Chr.8: 144506610-144506610 on Build GRCh38
rs554475955 Chr.8: 144506692-144506692 on Build GRCh38
rs760475837 Chr.8: 144506694-144506694 on Build GRCh38
rs367895411 Chr.8: 144506460-144506460 on Build GRCh38
rs764565994 Chr.8: 144506687-144506688 on Build GRCh38
rs751685146 Chr.8: 144506700-144506700 on Build GRCh38
rs748817984 Chr.8: 144506675-144506675 on Build GRCh38
rs759784022 Chr.8: 144506870-144506870 on Build GRCh38
rs751767383 Chr.8: 144506663-144506663 on Build GRCh38
rs775104723 Chr.8: 144506636-144506636 on Build GRCh38
rs537094982 Chr.8: 144506706-144506706 on Build GRCh38
rs750175698 Chr.8: 144506844-144506844 on Build GRCh38
rs760687865 Chr.8: 144506499-144506501 on Build GRCh38
rs146152870 Chr.8: 144506714-144506714 on Build GRCh38
rs745741227 Chr.8: 144506749-144506749 on Build GRCh38
rs1053275309 Chr.8: 144506709-144506709 on Build GRCh38
rs777931588 Chr.8: 144506856-144506856 on Build GRCh38
rs897755799 Chr.8: 144506618-144506618 on Build GRCh38
rs769471662 Chr.8: 144506555-144506555 on Build GRCh38
rs200263358 Chr.8: 144506872-144506872 on Build GRCh38
rs768436371 Chr.8: 144506755-144506757 on Build GRCh38
rs767839047 Chr.8: 144506755-144506755 on Build GRCh38
rs759126477 Chr.8: 144506465-144506465 on Build GRCh38
rs779496876 Chr.8: 144506808-144506808 on Build GRCh38
rs756004927 Chr.8: 144506850-144506850 on Build GRCh38
rs766308413 Chr.8: 144506647-144506647 on Build GRCh38
rs372280992 Chr.8: 144506587-144506587 on Build GRCh38
rs776606424 Chr.8: 144506748-144506748 on Build GRCh38
rs371318945 Chr.8: 144506843-144506843 on Build GRCh38
rs774129797 Chr.8: 144506584-144506584 on Build GRCh38
rs528675744 Chr.8: 144506684-144506692 on Build GRCh38
rs777058322 Chr.8: 144506548-144506548 on Build GRCh38
rs748417596 Chr.8: 144506511-144506511 on Build GRCh38
rs779845527 Chr.8: 144506688-144506689 on Build GRCh38
rs760002275 Chr.8: 144506822-144506822 on Build GRCh38
rs768299034 Chr.8: 144506637-144506637 on Build GRCh38
rs773547243 Chr.8: 144506816-144506816 on Build GRCh38
rs779122855 Chr.8: 144506428-144506428 on Build GRCh38
rs756130692 Chr.8: 144506421-144506421 on Build GRCh38
rs201282843 Chr.8: 144506842-144506842 on Build GRCh38
rs771805043 Chr.8: 144506641-144506641 on Build GRCh38
rs369157167 Chr.8: 144506715-144506715 on Build GRCh38
rs202022536 Chr.8: 144506669-144506669 on Build GRCh38
rs200853809 Chr.8: 144506562-144506562 on Build GRCh38
rs761903768 Chr.8: 144506702-144506702 on Build GRCh38
rs748082273 Chr.8: 144506830-144506830 on Build GRCh38
rs368684614 Chr.8: 144506624-144506624 on Build GRCh38
rs200508853 Chr.8: 144506649-144506649 on Build GRCh38
rs751337590 Chr.8: 144506490-144506490 on Build GRCh38
rs796637610 Chr.8: 144506687-144506688 on Build GRCh38
rs376611071 Chr.8: 144506604-144506604 on Build GRCh38
rs371146513 Chr.8: 144506772-144506772 on Build GRCh38
rs763055722 Chr.8: 144506697-144506697 on Build GRCh38
rs757182382 Chr.8: 144506491-144506491 on Build GRCh38
rs764369102 Chr.8: 144506657-144506657 on Build GRCh38
rs779825822 Chr.8: 144506851-144506851 on Build GRCh38
rs758578129 Chr.8: 144506619-144506619 on Build GRCh38
rs746798773 Chr.8: 144506739-144506739 on Build GRCh38
rs750038133 Chr.8: 144506794-144506794 on Build GRCh38
rs934861641 Chr.8: 144506708-144506708 on Build GRCh38
rs752135706 Chr.8: 144506781-144506781 on Build GRCh38
rs771044777 Chr.8: 144506860-144506860 on Build GRCh38
rs780720685 Chr.8: 144506672-144506672 on Build GRCh38
rs371985904 Chr.8: 144506871-144506871 on Build GRCh38
rs763542493 Chr.8: 144506841-144506841 on Build GRCh38
rs747338893 Chr.8: 144506540-144506540 on Build GRCh38
rs766420184 Chr.8: 144506515-144506523 on Build GRCh38
rs781699796 Chr.8: 144506631-144506631 on Build GRCh38
rs554139775 Chr.8: 144506769-144506769 on Build GRCh38
rs548905016 Chr.8: 144506678-144506678 on Build GRCh38
rs374357547 Chr.8: 144506742-144506742 on Build GRCh38
rs767361154 Chr.8: 144506492-144506492 on Build GRCh38
rs953448807 Chr.8: 144506729-144506729 on Build GRCh38
rs756626466 Chr.8: 144506671-144506671 on Build GRCh38
rs750967411 Chr.8: 144506664-144506664 on Build GRCh38
rs753857754 Chr.8: 144506493-144506493 on Build GRCh38
rs199690645 Chr.8: 144506767-144506767 on Build GRCh38
rs776865752 Chr.8: 144506651-144506655 on Build GRCh38
rs747697160 Chr.8: 144506725-144506725 on Build GRCh38
rs774156720 Chr.8: 144506768-144506778 on Build GRCh38
rs761970551 Chr.8: 144506754-144506754 on Build GRCh38
rs374893047 Chr.8: 144506426-144506426 on Build GRCh38
rs763689807 Chr.8: 144506580-144506580 on Build GRCh38
rs201844010 Chr.8: 144506867-144506867 on Build GRCh38
rs778598396 Chr.8: 144506681-144506681 on Build GRCh38
rs375588825 Chr.8: 144506498-144506498 on Build GRCh38
rs758784417 Chr.8: 144506684-144506685 on Build GRCh38
rs765419964 Chr.8: 144506679-144506679 on Build GRCh38
rs755111201 Chr.8: 144506507-144506507 on Build GRCh38
rs564073131 Chr.8: 144506640-144506640 on Build GRCh38
rs764126314 Chr.8: 144506698-144506698 on Build GRCh38
rs954142136 Chr.8: 144506445-144506446 on Build GRCh38
rs768545254 Chr.8: 144506460-144506465 on Build GRCh38
rs147998249 Chr.8: 144506797-144506797 on Build GRCh38
rs540894994 Chr.8: 144506467-144506467 on Build GRCh38
rs757878396 Chr.8: 144506840-144506840 on Build GRCh38
rs762997607 Chr.8: 144506654-144506654 on Build GRCh38
rs149616184 Chr.8: 144506539-144506539 on Build GRCh38
rs990450500 Chr.8: 144506497-144506497 on Build GRCh38
rs771785463 Chr.8: 144506690-144506690 on Build GRCh38
rs776136592 Chr.8: 144506461-144506461 on Build GRCh38
rs757626184 Chr.8: 144506626-144506626 on Build GRCh38
rs760932712 Chr.8: 144506818-144506818 on Build GRCh38
rs750763247 Chr.8: 144506710-144506710 on Build GRCh38
rs773933970 Chr.8: 144506639-144506639 on Build GRCh38
rs765657098 Chr.8: 144506832-144506832 on Build GRCh38
rs775560041 Chr.8: 144506805-144506805 on Build GRCh38
rs777370445 Chr.8: 144506785-144506785 on Build GRCh38
rs143462595 Chr.8: 144506768-144506768 on Build GRCh38
rs574293673 Chr.8: 144506793-144506793 on Build GRCh38
rs545775484 Chr.8: 144506634-144506634 on Build GRCh38
rs761195757 Chr.8: 144506459-144506459 on Build GRCh38
rs745364481 Chr.8: 144506549-144506549 on Build GRCh38
rs552732658 Chr.8: 144506484-144506484 on Build GRCh38
rs1027351811 Chr.8: 144506661-144506661 on Build GRCh38
rs370247961 Chr.8: 144506424-144506424 on Build GRCh38
rs747704575 Chr.8: 144506685-144506685 on Build GRCh38
rs781650289 Chr.8: 144506668-144506668 on Build GRCh38
rs778159097 Chr.8: 144506527-144506527 on Build GRCh38
rs773568283 Chr.8: 144506449-144506449 on Build GRCh38
rs765787079 Chr.8: 144506779-144506779 on Build GRCh38
rs753283113 Chr.8: 144506838-144506838 on Build GRCh38
rs767478951 Chr.8: 144506596-144506596 on Build GRCh38
rs766902019 Chr.8: 144506770-144506770 on Build GRCh38
rs577262676 Chr.8: 144506432-144506432 on Build GRCh38
rs530290663 Chr.8: 144506670-144506670 on Build GRCh38
rs771407331 Chr.8: 144506547-144506547 on Build GRCh38
rs368335519 Chr.8: 144506757-144506757 on Build GRCh38
rs201688778 Chr.8: 144506425-144506425 on Build GRCh38
rs756749551 Chr.8: 144506688-144506688 on Build GRCh38
rs772924716 Chr.8: 144506691-144506691 on Build GRCh38
rs778244305 Chr.8: 144506722-144506722 on Build GRCh38
rs752805176 Chr.8: 144506617-144506617 on Build GRCh38
rs777867324 Chr.8: 144506621-144506621 on Build GRCh38
rs747098200 Chr.8: 144506859-144506859 on Build GRCh38
rs768119857 Chr.8: 144506679-144506679 on Build GRCh38
rs139040022 Chr.8: 144506512-144506512 on Build GRCh38
rs777459970 Chr.8: 144506735-144506735 on Build GRCh38
rs766711055 Chr.8: 144506821-144506821 on Build GRCh38
rs762814084 Chr.8: 144506469-144506469 on Build GRCh38
rs769804699 Chr.8: 144506750-144506750 on Build GRCh38
rs991975762 Chr.8: 144506623-144506638 on Build GRCh38
rs761664060 Chr.8: 144506753-144506753 on Build GRCh38
rs1001681116 Chr.8: 144506482-144506482 on Build GRCh38
rs986654850 Chr.8: 144506450-144506450 on Build GRCh38
rs558794924 Chr.8: 144506430-144506430 on Build GRCh38
rs141505249 Chr.8: 144506731-144506731 on Build GRCh38
rs528169048 Chr.8: 144506644-144506644 on Build GRCh38
rs773012570 Chr.8: 144506643-144506643 on Build GRCh38
rs759703057 Chr.8: 144506671-144506697 on Build GRCh38
rs762649719 Chr.8: 144506572-144506572 on Build GRCh38
rs755373318 Chr.8: 144506778-144506778 on Build GRCh38
rs961203940 Chr.8: 144506635-144506635 on Build GRCh38
rs770942460 Chr.8: 144506867-144506868 on Build GRCh38
rs772319250 Chr.8: 144506809-144506809 on Build GRCh38
rs56010190 Chr.8: 144506688-144506688 on Build GRCh38
rs374457286 Chr.8: 144506784-144506784 on Build GRCh38
rs963210352 Chr.8: 144506740-144506740 on Build GRCh38

View Full Product Details