Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173886203-173886715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB37
Entrez Gene ID
84614
Gene Name
zinc finger and BTB domain containing 37
Gene Aliases
D430004I08Rik, ZNF908
UniGene
Hs.668497
Gene Chromosome Location
Chr.1: 173868095-173891122 on Build GRCh38
Transcripts
NM_001122770.1 XM_005245546.2 XM_006711578.3 XM_017002556.1 XM_017002557.1

SNP Detail

SNP ID Chromosome Location
rs894481876 Chr.1: 173886291-173886291 on Build GRCh38
rs773583917 Chr.1: 173886585-173886585 on Build GRCh38
rs987389506 Chr.1: 173886512-173886512 on Build GRCh38
rs556450792 Chr.1: 173886473-173886473 on Build GRCh38
rs966458399 Chr.1: 173886556-173886556 on Build GRCh38
rs965668140 Chr.1: 173886608-173886608 on Build GRCh38
rs941386893 Chr.1: 173886502-173886502 on Build GRCh38
rs35967651 Chr.1: 173886507-173886507 on Build GRCh38
rs888577691 Chr.1: 173886230-173886230 on Build GRCh38
rs983442174 Chr.1: 173886643-173886643 on Build GRCh38
rs556121907 Chr.1: 173886244-173886244 on Build GRCh38
rs752636855 Chr.1: 173886395-173886395 on Build GRCh38
rs894450783 Chr.1: 173886530-173886530 on Build GRCh38
rs1017969314 Chr.1: 173886523-173886523 on Build GRCh38
rs769635136 Chr.1: 173886586-173886586 on Build GRCh38
rs777594233 Chr.1: 173886307-173886307 on Build GRCh38
rs1024240507 Chr.1: 173886306-173886306 on Build GRCh38
rs957433476 Chr.1: 173886552-173886552 on Build GRCh38
rs1055677979 Chr.1: 173886418-173886418 on Build GRCh38
rs200589937 Chr.1: 173886667-173886667 on Build GRCh38
rs910407701 Chr.1: 173886233-173886233 on Build GRCh38
rs559015193 Chr.1: 173886548-173886548 on Build GRCh38
rs192676478 Chr.1: 173886602-173886602 on Build GRCh38
rs941522033 Chr.1: 173886689-173886689 on Build GRCh38
rs369134250 Chr.1: 173886501-173886501 on Build GRCh38
rs746704088 Chr.1: 173886323-173886323 on Build GRCh38
rs769647666 Chr.1: 173886449-173886449 on Build GRCh38
rs975762334 Chr.1: 173886610-173886610 on Build GRCh38
rs756276321 Chr.1: 173886593-173886595 on Build GRCh38
rs1047262008 Chr.1: 173886493-173886493 on Build GRCh38
rs144527009 Chr.1: 173886346-173886346 on Build GRCh38
rs1032810564 Chr.1: 173886549-173886549 on Build GRCh38
rs192599843 Chr.1: 173886314-173886314 on Build GRCh38
rs922838065 Chr.1: 173886611-173886611 on Build GRCh38
rs903137559 Chr.1: 173886539-173886539 on Build GRCh38
rs749472596 Chr.1: 173886617-173886617 on Build GRCh38
rs906749511 Chr.1: 173886399-173886399 on Build GRCh38
rs4650988 Chr.1: 173886427-173886427 on Build GRCh38
rs989233662 Chr.1: 173886294-173886294 on Build GRCh38
rs1034318327 Chr.1: 173886290-173886290 on Build GRCh38
rs561379066 Chr.1: 173886402-173886403 on Build GRCh38
rs1014155907 Chr.1: 173886295-173886295 on Build GRCh38
rs967602504 Chr.1: 173886315-173886315 on Build GRCh38
rs189270488 Chr.1: 173886417-173886417 on Build GRCh38
rs977724741 Chr.1: 173886574-173886574 on Build GRCh38
rs1055906070 Chr.1: 173886511-173886511 on Build GRCh38
rs548865020 Chr.1: 173886416-173886416 on Build GRCh38
rs1029623328 Chr.1: 173886499-173886499 on Build GRCh38
rs536106631 Chr.1: 173886458-173886458 on Build GRCh38
rs1005959890 Chr.1: 173886289-173886289 on Build GRCh38
rs567448433 Chr.1: 173886437-173886437 on Build GRCh38
rs1042079614 Chr.1: 173886372-173886372 on Build GRCh38
rs184397709 Chr.1: 173886406-173886406 on Build GRCh38
rs1045658420 Chr.1: 173886538-173886538 on Build GRCh38
rs545388095 Chr.1: 173886527-173886527 on Build GRCh38
rs945022146 Chr.1: 173886334-173886337 on Build GRCh38
rs943222815 Chr.1: 173886269-173886269 on Build GRCh38
rs576413825 Chr.1: 173886481-173886481 on Build GRCh38
rs550975814 Chr.1: 173886350-173886350 on Build GRCh38
rs944390750 Chr.1: 173886455-173886455 on Build GRCh38
rs941534056 Chr.1: 173886660-173886660 on Build GRCh38
rs770671938 Chr.1: 173886438-173886438 on Build GRCh38
rs747954972 Chr.1: 173886442-173886442 on Build GRCh38
rs922904347 Chr.1: 173886479-173886479 on Build GRCh38
rs1014293420 Chr.1: 173886535-173886535 on Build GRCh38

View Full Product Details